Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 52083135-52083636 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTF3L4
Entrez Gene ID
91408
Gene Name
basic transcription factor 3 like 4
Gene Aliases
-
UniGene
Hs.429839
Gene Chromosome Location
Chr.1: 52056185-52090716 on Build GRCh38
Transcripts
NM_001136497.2 NM_001243767.1 NM_152265.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6898186 c.245C>T p.T82I Chr.1: 52083416-52083416 on Build GRCh38
4490253 c.360C>T p.F120F Chr.1: 52083531-52083531 on Build GRCh38
6242287 c.367_368insA p.V124fs*5 Chr.1: 52083538-52083539 on Build GRCh38
910613 c.238G>T p.A80S Chr.1: 52083409-52083409 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs189016234 Chr.1: 52083167-52083167 on Build GRCh38
rs929177124 Chr.1: 52083204-52083204 on Build GRCh38
rs984618703 Chr.1: 52083593-52083593 on Build GRCh38
rs760217720 Chr.1: 52083522-52083522 on Build GRCh38
rs776829117 Chr.1: 52083448-52083448 on Build GRCh38
rs750052703 Chr.1: 52083489-52083489 on Build GRCh38
rs1012649392 Chr.1: 52083493-52083493 on Build GRCh38
rs778924068 Chr.1: 52083554-52083554 on Build GRCh38
rs151133552 Chr.1: 52083246-52083246 on Build GRCh38
rs34672243 Chr.1: 52083243-52083244 on Build GRCh38
rs140100882 Chr.1: 52083285-52083285 on Build GRCh38
rs926387478 Chr.1: 52083235-52083235 on Build GRCh38
rs991179316 Chr.1: 52083161-52083161 on Build GRCh38
rs558995370 Chr.1: 52083231-52083231 on Build GRCh38
rs778169535 Chr.1: 52083575-52083575 on Build GRCh38
rs755889313 Chr.1: 52083318-52083318 on Build GRCh38
rs74083206 Chr.1: 52083589-52083589 on Build GRCh38
rs909994603 Chr.1: 52083220-52083220 on Build GRCh38
rs370518293 Chr.1: 52083196-52083196 on Build GRCh38
rs760136849 Chr.1: 52083455-52083455 on Build GRCh38
rs773857304 Chr.1: 52083482-52083482 on Build GRCh38
rs758888096 Chr.1: 52083586-52083589 on Build GRCh38
rs573379408 Chr.1: 52083197-52083197 on Build GRCh38
rs746212891 Chr.1: 52083592-52083592 on Build GRCh38
rs1056700745 Chr.1: 52083263-52083263 on Build GRCh38
rs1033163721 Chr.1: 52083463-52083463 on Build GRCh38
rs771286463 Chr.1: 52083435-52083435 on Build GRCh38
rs554880992 Chr.1: 52083177-52083177 on Build GRCh38
rs35771250 Chr.1: 52083203-52083204 on Build GRCh38
rs146227724 Chr.1: 52083535-52083535 on Build GRCh38
rs747295467 Chr.1: 52083578-52083578 on Build GRCh38
rs1019154161 Chr.1: 52083186-52083186 on Build GRCh38
rs1051816299 Chr.1: 52083301-52083301 on Build GRCh38
rs1039698308 Chr.1: 52083236-52083236 on Build GRCh38
rs758498923 Chr.1: 52083574-52083574 on Build GRCh38
rs946449141 Chr.1: 52083225-52083225 on Build GRCh38
rs915535573 Chr.1: 52083174-52083174 on Build GRCh38
rs754853340 Chr.1: 52083371-52083371 on Build GRCh38
rs747353221 Chr.1: 52083428-52083428 on Build GRCh38
rs761256803 Chr.1: 52083483-52083483 on Build GRCh38
rs753858242 Chr.1: 52083360-52083360 on Build GRCh38
rs771036630 Chr.1: 52083587-52083587 on Build GRCh38
rs957089362 Chr.1: 52083159-52083159 on Build GRCh38
rs150363662 Chr.1: 52083536-52083536 on Build GRCh38
rs756934677 Chr.1: 52083289-52083289 on Build GRCh38
rs779889062 Chr.1: 52083330-52083330 on Build GRCh38
rs767179583 Chr.1: 52083484-52083484 on Build GRCh38
rs1048879767 Chr.1: 52083207-52083207 on Build GRCh38
rs953499609 Chr.1: 52083171-52083171 on Build GRCh38
rs1022633881 Chr.1: 52083521-52083521 on Build GRCh38
rs999045629 Chr.1: 52083416-52083416 on Build GRCh38
rs939965698 Chr.1: 52083278-52083280 on Build GRCh38
rs369805791 Chr.1: 52083573-52083573 on Build GRCh38
rs564581518 Chr.1: 52083203-52083203 on Build GRCh38
rs770271523 Chr.1: 52083478-52083478 on Build GRCh38
rs768875398 Chr.1: 52083260-52083260 on Build GRCh38
rs917836581 Chr.1: 52083264-52083264 on Build GRCh38
rs534158538 Chr.1: 52083215-52083215 on Build GRCh38
rs199657688 Chr.1: 52083402-52083402 on Build GRCh38
rs755036545 Chr.1: 52083545-52083545 on Build GRCh38
rs779195605 Chr.1: 52083374-52083374 on Build GRCh38

View Full Product Details