Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 52353457-52353957 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CC2D1B
Entrez Gene ID
200014
Gene Name
coiled-coil and C2 domain containing 1B
Gene Aliases
-
UniGene
Hs.591451
Gene Chromosome Location
Chr.1: 52348437-52366291 on Build GRCh38
Transcripts
NM_032449.2 XM_005270590.2 XM_005270591.2 XM_005270593.2 XM_006710423.2 XM_017000559.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4608286 c.2550G>T p.W850C Chr.1: 52353546-52353546 on Build GRCh38
3985127 c.2540C>G p.T847S Chr.1: 52353556-52353556 on Build GRCh38
5556077 c.1818G>T p.G606G Chr.1: 52353620-52353620 on Build GRCh38
1247406 c.2527G>A p.V843M Chr.1: 52353569-52353569 on Build GRCh38
6063724 c.1835+1G>T p.? Chr.1: 52353602-52353602 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746892769 Chr.1: 52353620-52353620 on Build GRCh38
rs762339491 Chr.1: 52353633-52353633 on Build GRCh38
rs763364108 Chr.1: 52353499-52353499 on Build GRCh38
rs201374933 Chr.1: 52353496-52353496 on Build GRCh38
rs765132573 Chr.1: 52353603-52353603 on Build GRCh38
rs746023729 Chr.1: 52353566-52353566 on Build GRCh38
rs1018203082 Chr.1: 52353887-52353887 on Build GRCh38
rs986300698 Chr.1: 52353617-52353617 on Build GRCh38
rs373167954 Chr.1: 52353630-52353630 on Build GRCh38
rs1026779246 Chr.1: 52353846-52353847 on Build GRCh38
rs758899419 Chr.1: 52353908-52353908 on Build GRCh38
rs377744839 Chr.1: 52353542-52353542 on Build GRCh38
rs549212022 Chr.1: 52353874-52353874 on Build GRCh38
rs777595405 Chr.1: 52353619-52353619 on Build GRCh38
rs11804966 Chr.1: 52353829-52353829 on Build GRCh38
rs764806198 Chr.1: 52353602-52353602 on Build GRCh38
rs775703581 Chr.1: 52353578-52353578 on Build GRCh38
rs41294498 Chr.1: 52353858-52353858 on Build GRCh38
rs771150022 Chr.1: 52353774-52353777 on Build GRCh38
rs763563610 Chr.1: 52353580-52353580 on Build GRCh38
rs767111281 Chr.1: 52353503-52353503 on Build GRCh38
rs1015533207 Chr.1: 52353791-52353791 on Build GRCh38
rs770626996 Chr.1: 52353654-52353654 on Build GRCh38
rs750984681 Chr.1: 52353662-52353662 on Build GRCh38
rs776801789 Chr.1: 52353485-52353485 on Build GRCh38
rs763388606 Chr.1: 52353773-52353773 on Build GRCh38
rs770950405 Chr.1: 52353557-52353557 on Build GRCh38
rs751075716 Chr.1: 52353718-52353718 on Build GRCh38
rs760045420 Chr.1: 52353617-52353618 on Build GRCh38
rs528222929 Chr.1: 52353718-52353718 on Build GRCh38
rs776262672 Chr.1: 52353659-52353659 on Build GRCh38
rs1055371434 Chr.1: 52353714-52353714 on Build GRCh38
rs575473780 Chr.1: 52353616-52353616 on Build GRCh38
rs368931438 Chr.1: 52353596-52353596 on Build GRCh38
rs903680750 Chr.1: 52353736-52353736 on Build GRCh38
rs557312416 Chr.1: 52353588-52353588 on Build GRCh38
rs903974743 Chr.1: 52353684-52353684 on Build GRCh38
rs922953510 Chr.1: 52353925-52353925 on Build GRCh38
rs766157337 Chr.1: 52353515-52353515 on Build GRCh38
rs753488099 Chr.1: 52353516-52353516 on Build GRCh38
rs776853274 Chr.1: 52353563-52353563 on Build GRCh38
rs770210325 Chr.1: 52353570-52353570 on Build GRCh38
rs143725276 Chr.1: 52353581-52353581 on Build GRCh38
rs749877534 Chr.1: 52353510-52353510 on Build GRCh38
rs755869037 Chr.1: 52353512-52353512 on Build GRCh38
rs896811482 Chr.1: 52353816-52353816 on Build GRCh38
rs1015685827 Chr.1: 52353523-52353523 on Build GRCh38
rs750910682 Chr.1: 52353913-52353914 on Build GRCh38
rs778571580 Chr.1: 52353529-52353529 on Build GRCh38
rs576157797 Chr.1: 52353849-52353850 on Build GRCh38
rs547072569 Chr.1: 52353930-52353930 on Build GRCh38
rs144481098 Chr.1: 52353621-52353621 on Build GRCh38
rs765528560 Chr.1: 52353488-52353488 on Build GRCh38
rs561018851 Chr.1: 52353701-52353701 on Build GRCh38
rs554425856 Chr.1: 52353843-52353843 on Build GRCh38
rs564687531 Chr.1: 52353814-52353814 on Build GRCh38
rs937826393 Chr.1: 52353702-52353702 on Build GRCh38
rs767981801 Chr.1: 52353674-52353675 on Build GRCh38
rs376482808 Chr.1: 52353685-52353685 on Build GRCh38
rs945384041 Chr.1: 52353635-52353635 on Build GRCh38
rs532138414 Chr.1: 52353821-52353821 on Build GRCh38
rs758249382 Chr.1: 52353536-52353536 on Build GRCh38
rs374120552 Chr.1: 52353579-52353579 on Build GRCh38
rs953799439 Chr.1: 52353854-52353854 on Build GRCh38
rs749644041 Chr.1: 52353623-52353623 on Build GRCh38
rs142628405 Chr.1: 52353904-52353904 on Build GRCh38
rs945158057 Chr.1: 52353928-52353928 on Build GRCh38
rs971490233 Chr.1: 52353583-52353584 on Build GRCh38
rs1000659872 Chr.1: 52353750-52353750 on Build GRCh38
rs759305230 Chr.1: 52353671-52353671 on Build GRCh38
rs754829407 Chr.1: 52353519-52353519 on Build GRCh38
rs774861740 Chr.1: 52353618-52353618 on Build GRCh38
rs1039992887 Chr.1: 52353658-52353658 on Build GRCh38
rs769012646 Chr.1: 52353624-52353624 on Build GRCh38
rs746145085 Chr.1: 52353622-52353622 on Build GRCh38
rs41294496 Chr.1: 52353618-52353618 on Build GRCh38
rs748178136 Chr.1: 52353534-52353534 on Build GRCh38
rs765925293 Chr.1: 52353592-52353592 on Build GRCh38
rs540216462 Chr.1: 52353801-52353801 on Build GRCh38
rs753616552 Chr.1: 52353593-52353593 on Build GRCh38
rs951077285 Chr.1: 52353707-52353707 on Build GRCh38
rs190292882 Chr.1: 52353909-52353909 on Build GRCh38
rs777950699 Chr.1: 52353541-52353541 on Build GRCh38

View Full Product Details