Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 52355060-52355560 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CC2D1B
Entrez Gene ID
200014
Gene Name
coiled-coil and C2 domain containing 1B
Gene Aliases
-
UniGene
Hs.591451
Gene Chromosome Location
Chr.1: 52348437-52366291 on Build GRCh38
Transcripts
NM_032449.2 XM_005270590.2 XM_005270591.2 XM_005270593.2 XM_006710423.2 XM_017000559.1

SNP Detail

SNP ID Chromosome Location
rs566793831 Chr.1: 52355277-52355277 on Build GRCh38
rs766321768 Chr.1: 52355442-52355442 on Build GRCh38
rs377556369 Chr.1: 52355433-52355433 on Build GRCh38
rs989566952 Chr.1: 52355460-52355460 on Build GRCh38
rs149059637 Chr.1: 52355083-52355083 on Build GRCh38
rs761241297 Chr.1: 52355426-52355426 on Build GRCh38
rs906755549 Chr.1: 52355322-52355322 on Build GRCh38
rs371748647 Chr.1: 52355243-52355243 on Build GRCh38
rs141635623 Chr.1: 52355443-52355443 on Build GRCh38
rs748201880 Chr.1: 52355382-52355382 on Build GRCh38
rs776959494 Chr.1: 52355399-52355399 on Build GRCh38
rs996607937 Chr.1: 52355372-52355372 on Build GRCh38
rs573524481 Chr.1: 52355477-52355477 on Build GRCh38
rs888503957 Chr.1: 52355111-52355111 on Build GRCh38
rs533726575 Chr.1: 52355336-52355336 on Build GRCh38
rs1030993771 Chr.1: 52355453-52355453 on Build GRCh38
rs771129921 Chr.1: 52355379-52355380 on Build GRCh38
rs75157276 Chr.1: 52355370-52355370 on Build GRCh38
rs565441302 Chr.1: 52355518-52355518 on Build GRCh38
rs751003738 Chr.1: 52355354-52355354 on Build GRCh38
rs551107083 Chr.1: 52355108-52355108 on Build GRCh38
rs374603660 Chr.1: 52355111-52355111 on Build GRCh38
rs192052757 Chr.1: 52355379-52355379 on Build GRCh38
rs994090610 Chr.1: 52355097-52355097 on Build GRCh38
rs753439785 Chr.1: 52355245-52355245 on Build GRCh38
rs939602884 Chr.1: 52355513-52355513 on Build GRCh38
rs371062800 Chr.1: 52355364-52355364 on Build GRCh38
rs752616510 Chr.1: 52355469-52355469 on Build GRCh38
rs954889424 Chr.1: 52355176-52355176 on Build GRCh38
rs951138553 Chr.1: 52355092-52355092 on Build GRCh38
rs559007372 Chr.1: 52355244-52355244 on Build GRCh38
rs776015720 Chr.1: 52355417-52355417 on Build GRCh38
rs746241634 Chr.1: 52355480-52355480 on Build GRCh38
rs772624108 Chr.1: 52355256-52355256 on Build GRCh38
rs553592121 Chr.1: 52355166-52355166 on Build GRCh38
rs774773453 Chr.1: 52355492-52355493 on Build GRCh38
rs79146324 Chr.1: 52355360-52355360 on Build GRCh38
rs554939521 Chr.1: 52355471-52355471 on Build GRCh38
rs565113251 Chr.1: 52355497-52355518 on Build GRCh38
rs903084433 Chr.1: 52355420-52355420 on Build GRCh38
rs771765313 Chr.1: 52355497-52355497 on Build GRCh38
rs925626806 Chr.1: 52355221-52355221 on Build GRCh38
rs915679579 Chr.1: 52355251-52355251 on Build GRCh38
rs6588428 Chr.1: 52355133-52355133 on Build GRCh38
rs74080943 Chr.1: 52355431-52355431 on Build GRCh38
rs555860653 Chr.1: 52355385-52355385 on Build GRCh38
rs6690342 Chr.1: 52355180-52355180 on Build GRCh38
rs771344611 Chr.1: 52355386-52355386 on Build GRCh38
rs971587394 Chr.1: 52355512-52355512 on Build GRCh38
rs777821519 Chr.1: 52355476-52355476 on Build GRCh38
rs1047029094 Chr.1: 52355359-52355359 on Build GRCh38
rs771111700 Chr.1: 52355478-52355478 on Build GRCh38
rs1047059700 Chr.1: 52355102-52355102 on Build GRCh38
rs956814246 Chr.1: 52355459-52355461 on Build GRCh38
rs767068004 Chr.1: 52355430-52355430 on Build GRCh38
rs778732748 Chr.1: 52355466-52355466 on Build GRCh38
rs922511529 Chr.1: 52355173-52355173 on Build GRCh38
rs754760107 Chr.1: 52355458-52355458 on Build GRCh38
rs1005363222 Chr.1: 52355116-52355117 on Build GRCh38
rs967294504 Chr.1: 52355207-52355207 on Build GRCh38
rs772136891 Chr.1: 52355384-52355384 on Build GRCh38
rs1046708253 Chr.1: 52355333-52355333 on Build GRCh38
rs977055854 Chr.1: 52355211-52355211 on Build GRCh38
rs375520962 Chr.1: 52355280-52355280 on Build GRCh38
rs777923871 Chr.1: 52355350-52355351 on Build GRCh38
rs928187444 Chr.1: 52355299-52355299 on Build GRCh38
rs1005612207 Chr.1: 52355122-52355122 on Build GRCh38
rs770136017 Chr.1: 52355481-52355481 on Build GRCh38
rs755975631 Chr.1: 52355367-52355367 on Build GRCh38
rs1016708561 Chr.1: 52355127-52355127 on Build GRCh38
rs982287996 Chr.1: 52355537-52355537 on Build GRCh38
rs998463666 Chr.1: 52355167-52355167 on Build GRCh38
rs987467819 Chr.1: 52355177-52355177 on Build GRCh38
rs948244041 Chr.1: 52355252-52355252 on Build GRCh38
rs975306450 Chr.1: 52355150-52355150 on Build GRCh38
rs963902095 Chr.1: 52355148-52355148 on Build GRCh38
rs764427021 Chr.1: 52355527-52355527 on Build GRCh38
rs935390054 Chr.1: 52355224-52355224 on Build GRCh38
rs746069218 Chr.1: 52355493-52355493 on Build GRCh38
rs1022710476 Chr.1: 52355503-52355503 on Build GRCh38
rs1003636744 Chr.1: 52355331-52355331 on Build GRCh38
rs754845040 Chr.1: 52355373-52355373 on Build GRCh38
rs182473142 Chr.1: 52355482-52355482 on Build GRCh38
rs765695799 Chr.1: 52355405-52355405 on Build GRCh38
rs749293974 Chr.1: 52355371-52355371 on Build GRCh38
rs762458453 Chr.1: 52355358-52355358 on Build GRCh38
rs749316468 Chr.1: 52355359-52355360 on Build GRCh38
rs760131080 Chr.1: 52355401-52355401 on Build GRCh38
rs886885041 Chr.1: 52355361-52355362 on Build GRCh38
rs930242473 Chr.1: 52355329-52355329 on Build GRCh38

View Full Product Details