Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 53213978-53214512 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf123
Entrez Gene ID
54987
Gene Name
chromosome 1 open reading frame 123
Gene Aliases
-
UniGene
Hs.525391
Gene Chromosome Location
Chr.1: 53214099-53220639 on Build GRCh38
Transcripts
NM_001304759.1 NM_001304760.1 NM_017887.2 NM_000098.2 XM_005270484.1

Gene Symbol
CPT2
Entrez Gene ID
1376
Gene Name
carnitine palmitoyltransferase 2
Gene Aliases
CPT1, CPTASE, IIAE4
UniGene
Hs.713535
Gene Chromosome Location
Chr.1: 53196429-53214197 on Build GRCh38
Transcripts
NM_001304759.1 NM_001304760.1 NM_017887.2 NM_000098.2 XM_005270484.1

SNP Detail

SNP ID Chromosome Location
rs1037360749 Chr.1: 53214353-53214353 on Build GRCh38
rs574256743 Chr.1: 53214112-53214112 on Build GRCh38
rs540664995 Chr.1: 53214275-53214275 on Build GRCh38
rs369718292 Chr.1: 53214092-53214092 on Build GRCh38
rs1056438 Chr.1: 53214206-53214206 on Build GRCh38
rs969148073 Chr.1: 53214429-53214429 on Build GRCh38
rs1056875315 Chr.1: 53214333-53214333 on Build GRCh38
rs905729083 Chr.1: 53214170-53214170 on Build GRCh38
rs918359221 Chr.1: 53214023-53214023 on Build GRCh38
rs1019670024 Chr.1: 53214278-53214278 on Build GRCh38
rs1049983412 Chr.1: 53214381-53214381 on Build GRCh38
rs1055553519 Chr.1: 53214039-53214039 on Build GRCh38
rs549416028 Chr.1: 53214414-53214414 on Build GRCh38
rs897350965 Chr.1: 53214089-53214089 on Build GRCh38
rs1027209249 Chr.1: 53214094-53214094 on Build GRCh38
rs947169860 Chr.1: 53214145-53214145 on Build GRCh38
rs1048595305 Chr.1: 53214384-53214384 on Build GRCh38
rs936592573 Chr.1: 53214315-53214315 on Build GRCh38
rs930102958 Chr.1: 53214380-53214380 on Build GRCh38
rs17848487 Chr.1: 53214284-53214284 on Build GRCh38
rs1008242595 Chr.1: 53214277-53214277 on Build GRCh38
rs925274669 Chr.1: 53214034-53214034 on Build GRCh38
rs768410018 Chr.1: 53214347-53214347 on Build GRCh38
rs751241039 Chr.1: 53214212-53214212 on Build GRCh38
rs532772009 Chr.1: 53214372-53214372 on Build GRCh38
rs867768853 Chr.1: 53214137-53214137 on Build GRCh38
rs990827214 Chr.1: 53214325-53214325 on Build GRCh38
rs529023047 Chr.1: 53214439-53214439 on Build GRCh38
rs914596553 Chr.1: 53214300-53214300 on Build GRCh38
rs564148316 Chr.1: 53214348-53214348 on Build GRCh38
rs925178794 Chr.1: 53214314-53214314 on Build GRCh38
rs543259286 Chr.1: 53214149-53214149 on Build GRCh38
rs1046944805 Chr.1: 53214356-53214356 on Build GRCh38
rs907627156 Chr.1: 53214345-53214345 on Build GRCh38
rs1028433098 Chr.1: 53214247-53214247 on Build GRCh38
rs1046555099 Chr.1: 53214064-53214064 on Build GRCh38
rs949533362 Chr.1: 53214061-53214061 on Build GRCh38
rs996193058 Chr.1: 53214244-53214244 on Build GRCh38
rs927095044 Chr.1: 53214319-53214319 on Build GRCh38
rs1016229189 Chr.1: 53214228-53214228 on Build GRCh38
rs927209370 Chr.1: 53214447-53214447 on Build GRCh38
rs761214937 Chr.1: 53214487-53214487 on Build GRCh38
rs956146402 Chr.1: 53214409-53214409 on Build GRCh38
rs976277044 Chr.1: 53214232-53214232 on Build GRCh38
rs966545633 Chr.1: 53214280-53214280 on Build GRCh38
rs1008358748 Chr.1: 53214383-53214383 on Build GRCh38
rs895197584 Chr.1: 53214053-53214053 on Build GRCh38
rs1056425 Chr.1: 53214418-53214418 on Build GRCh38
rs779777403 Chr.1: 53214323-53214323 on Build GRCh38
rs886046410 Chr.1: 53214034-53214034 on Build GRCh38
rs1030104832 Chr.1: 53214250-53214250 on Build GRCh38
rs886046411 Chr.1: 53214195-53214196 on Build GRCh38
rs751539798 Chr.1: 53214006-53214006 on Build GRCh38
rs977908377 Chr.1: 53214306-53214306 on Build GRCh38
rs549196653 Chr.1: 53214479-53214479 on Build GRCh38
rs374495416 Chr.1: 53214093-53214094 on Build GRCh38
rs769556721 Chr.1: 53214296-53214296 on Build GRCh38
rs979993634 Chr.1: 53214436-53214436 on Build GRCh38
rs889922003 Chr.1: 53214382-53214382 on Build GRCh38
rs557500034 Chr.1: 53214005-53214005 on Build GRCh38
rs936646936 Chr.1: 53214035-53214035 on Build GRCh38
rs954234560 Chr.1: 53214260-53214260 on Build GRCh38
rs749276489 Chr.1: 53214346-53214346 on Build GRCh38
rs1010771301 Chr.1: 53214412-53214412 on Build GRCh38
rs14829 Chr.1: 53214202-53214202 on Build GRCh38

View Full Product Details