Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 53218335-53218848 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf123
Entrez Gene ID
54987
Gene Name
chromosome 1 open reading frame 123
Gene Aliases
-
UniGene
Hs.525391
Gene Chromosome Location
Chr.1: 53214099-53220639 on Build GRCh38
Transcripts
NM_001304759.1 NM_001304760.1 NM_017887.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7358216 c.187A>G p.M63V Chr.1: 53218456-53218456 on Build GRCh38
5846973 c.229+1G>A p.? Chr.1: 53218413-53218413 on Build GRCh38
3490738 c.186C>T p.S62S Chr.1: 53218457-53218457 on Build GRCh38
3490737 c.224C>T p.S75F Chr.1: 53218419-53218419 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs770439959 Chr.1: 53218411-53218411 on Build GRCh38
rs200900856 Chr.1: 53218528-53218528 on Build GRCh38
rs895736408 Chr.1: 53218717-53218717 on Build GRCh38
rs189510666 Chr.1: 53218473-53218473 on Build GRCh38
rs756188383 Chr.1: 53218387-53218387 on Build GRCh38
rs779495665 Chr.1: 53218483-53218483 on Build GRCh38
rs570402850 Chr.1: 53218785-53218785 on Build GRCh38
rs922331624 Chr.1: 53218748-53218748 on Build GRCh38
rs762965134 Chr.1: 53218438-53218438 on Build GRCh38
rs34035770 Chr.1: 53218762-53218763 on Build GRCh38
rs555568109 Chr.1: 53218552-53218552 on Build GRCh38
rs199913224 Chr.1: 53218403-53218403 on Build GRCh38
rs994956102 Chr.1: 53218757-53218757 on Build GRCh38
rs373880930 Chr.1: 53218538-53218538 on Build GRCh38
rs201416768 Chr.1: 53218500-53218500 on Build GRCh38
rs987767998 Chr.1: 53218773-53218773 on Build GRCh38
rs3736118 Chr.1: 53218652-53218652 on Build GRCh38
rs11546995 Chr.1: 53218444-53218444 on Build GRCh38
rs763801420 Chr.1: 53218541-53218542 on Build GRCh38
rs1014231359 Chr.1: 53218728-53218728 on Build GRCh38
rs533734605 Chr.1: 53218730-53218730 on Build GRCh38
rs1001380444 Chr.1: 53218699-53218699 on Build GRCh38
rs145021398 Chr.1: 53218418-53218418 on Build GRCh38
rs767565531 Chr.1: 53218461-53218461 on Build GRCh38
rs145124211 Chr.1: 53218462-53218462 on Build GRCh38
rs767341598 Chr.1: 53218818-53218818 on Build GRCh38
rs113038301 Chr.1: 53218374-53218374 on Build GRCh38
rs188449206 Chr.1: 53218816-53218816 on Build GRCh38
rs750456320 Chr.1: 53218381-53218381 on Build GRCh38
rs112877831 Chr.1: 53218772-53218772 on Build GRCh38
rs1034243075 Chr.1: 53218707-53218707 on Build GRCh38
rs780171532 Chr.1: 53218397-53218397 on Build GRCh38
rs1003488750 Chr.1: 53218596-53218596 on Build GRCh38
rs776437572 Chr.1: 53218414-53218414 on Build GRCh38
rs146859972 Chr.1: 53218630-53218630 on Build GRCh38
rs143898173 Chr.1: 53218615-53218615 on Build GRCh38
rs891529846 Chr.1: 53218625-53218625 on Build GRCh38
rs761694363 Chr.1: 53218460-53218460 on Build GRCh38
rs761644990 Chr.1: 53218823-53218826 on Build GRCh38
rs375523271 Chr.1: 53218410-53218410 on Build GRCh38
rs745315369 Chr.1: 53218415-53218415 on Build GRCh38
rs576038288 Chr.1: 53218398-53218398 on Build GRCh38
rs1016642760 Chr.1: 53218653-53218653 on Build GRCh38
rs1025887952 Chr.1: 53218598-53218598 on Build GRCh38
rs547950194 Chr.1: 53218645-53218645 on Build GRCh38
rs920814813 Chr.1: 53218424-53218424 on Build GRCh38
rs1053883708 Chr.1: 53218527-53218527 on Build GRCh38
rs775007939 Chr.1: 53218372-53218372 on Build GRCh38
rs781510282 Chr.1: 53218375-53218375 on Build GRCh38
rs755553635 Chr.1: 53218399-53218399 on Build GRCh38
rs562231137 Chr.1: 53218471-53218471 on Build GRCh38
rs113531134 Chr.1: 53218482-53218482 on Build GRCh38
rs963647597 Chr.1: 53218671-53218671 on Build GRCh38
rs552195065 Chr.1: 53218604-53218604 on Build GRCh38
rs945855800 Chr.1: 53218636-53218636 on Build GRCh38
rs141056335 Chr.1: 53218456-53218456 on Build GRCh38
rs757183059 Chr.1: 53218735-53218735 on Build GRCh38
rs145613337 Chr.1: 53218463-53218463 on Build GRCh38
rs1012254961 Chr.1: 53218534-53218534 on Build GRCh38
rs368031769 Chr.1: 53218761-53218761 on Build GRCh38
rs748574766 Chr.1: 53218407-53218407 on Build GRCh38
rs906441119 Chr.1: 53218556-53218556 on Build GRCh38
rs904383095 Chr.1: 53218695-53218695 on Build GRCh38
rs760163030 Chr.1: 53218473-53218473 on Build GRCh38
rs769621068 Chr.1: 53218539-53218539 on Build GRCh38
rs755399907 Chr.1: 53218474-53218474 on Build GRCh38
rs775112921 Chr.1: 53218540-53218540 on Build GRCh38
rs191880219 Chr.1: 53218546-53218546 on Build GRCh38
rs776233308 Chr.1: 53218822-53218831 on Build GRCh38
rs149527063 Chr.1: 53218470-53218470 on Build GRCh38
rs142980878 Chr.1: 53218434-53218434 on Build GRCh38
rs112803850 Chr.1: 53218413-53218413 on Build GRCh38

View Full Product Details