Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 53464530-53465035 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMRTB1
Entrez Gene ID
63948
Gene Name
DMRT like family B with proline rich C-terminal 1
Gene Aliases
-
UniGene
Hs.131654
Gene Chromosome Location
Chr.1: 53459399-53467488 on Build GRCh38
Transcripts
NM_033067.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3805567 c.927G>A p.L309L Chr.1: 53464813-53464813 on Build GRCh38
910790 c.954G>T p.E318D Chr.1: 53464840-53464840 on Build GRCh38
191539 c.897G>A p.P299P Chr.1: 53464783-53464783 on Build GRCh38
3786246 c.933C>T p.V311V Chr.1: 53464819-53464819 on Build GRCh38
6126935 c.789C>A p.Y263* Chr.1: 53464675-53464675 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs925213296 Chr.1: 53464976-53464976 on Build GRCh38
rs964733737 Chr.1: 53464941-53464941 on Build GRCh38
rs765508334 Chr.1: 53464596-53464597 on Build GRCh38
rs748264748 Chr.1: 53464858-53464859 on Build GRCh38
rs761941427 Chr.1: 53464716-53464716 on Build GRCh38
rs758295903 Chr.1: 53464705-53464706 on Build GRCh38
rs990630561 Chr.1: 53464990-53464990 on Build GRCh38
rs576142948 Chr.1: 53464741-53464741 on Build GRCh38
rs752152791 Chr.1: 53464612-53464612 on Build GRCh38
rs867923942 Chr.1: 53464791-53464791 on Build GRCh38
rs910229403 Chr.1: 53464934-53464934 on Build GRCh38
rs748875770 Chr.1: 53464624-53464624 on Build GRCh38
rs867000359 Chr.1: 53464729-53464729 on Build GRCh38
rs756777522 Chr.1: 53464746-53464746 on Build GRCh38
rs547919415 Chr.1: 53464566-53464566 on Build GRCh38
rs868663808 Chr.1: 53464688-53464688 on Build GRCh38
rs771652627 Chr.1: 53464646-53464646 on Build GRCh38
rs935441013 Chr.1: 53464817-53464817 on Build GRCh38
rs545617032 Chr.1: 53464722-53464722 on Build GRCh38
rs542149843 Chr.1: 53464816-53464816 on Build GRCh38
rs371372052 Chr.1: 53464854-53464854 on Build GRCh38
rs762915457 Chr.1: 53464683-53464683 on Build GRCh38
rs544399051 Chr.1: 53464999-53465000 on Build GRCh38
rs777039064 Chr.1: 53464798-53464798 on Build GRCh38
rs201126524 Chr.1: 53464692-53464692 on Build GRCh38
rs745627508 Chr.1: 53464710-53464710 on Build GRCh38
rs201967120 Chr.1: 53464622-53464622 on Build GRCh38
rs994180118 Chr.1: 53464887-53464887 on Build GRCh38
rs986079062 Chr.1: 53464915-53464915 on Build GRCh38
rs773661120 Chr.1: 53464780-53464780 on Build GRCh38
rs748199064 Chr.1: 53464777-53464777 on Build GRCh38
rs527679471 Chr.1: 53464564-53464564 on Build GRCh38
rs752444740 Chr.1: 53464694-53464694 on Build GRCh38
rs746978994 Chr.1: 53464774-53464775 on Build GRCh38
rs887977388 Chr.1: 53464911-53464911 on Build GRCh38
rs768532791 Chr.1: 53464783-53464784 on Build GRCh38
rs376960740 Chr.1: 53464873-53464873 on Build GRCh38
rs377154853 Chr.1: 53464690-53464690 on Build GRCh38
rs745464968 Chr.1: 53464696-53464698 on Build GRCh38
rs112444758 Chr.1: 53464723-53464723 on Build GRCh38
rs142464747 Chr.1: 53464700-53464700 on Build GRCh38
rs1017648105 Chr.1: 53464939-53464939 on Build GRCh38
rs769757100 Chr.1: 53464712-53464712 on Build GRCh38
rs147771576 Chr.1: 53464689-53464689 on Build GRCh38
rs978441980 Chr.1: 53464971-53464971 on Build GRCh38
rs761203609 Chr.1: 53464783-53464783 on Build GRCh38
rs372673074 Chr.1: 53464611-53464611 on Build GRCh38
rs147307088 Chr.1: 53464678-53464683 on Build GRCh38
rs367594464 Chr.1: 53464631-53464631 on Build GRCh38
rs776578628 Chr.1: 53464674-53464674 on Build GRCh38
rs779643202 Chr.1: 53464775-53464780 on Build GRCh38
rs375544915 Chr.1: 53464678-53464686 on Build GRCh38
rs533549553 Chr.1: 53464693-53464693 on Build GRCh38
rs1001307444 Chr.1: 53464743-53464743 on Build GRCh38
rs959601177 Chr.1: 53464771-53464771 on Build GRCh38
rs746862743 Chr.1: 53464702-53464702 on Build GRCh38
rs752254162 Chr.1: 53464738-53464738 on Build GRCh38
rs748529063 Chr.1: 53464680-53464681 on Build GRCh38
rs749416642 Chr.1: 53464774-53464774 on Build GRCh38
rs958147356 Chr.1: 53464989-53464989 on Build GRCh38
rs992612102 Chr.1: 53464592-53464592 on Build GRCh38
rs756088048 Chr.1: 53464871-53464871 on Build GRCh38
rs150466113 Chr.1: 53464713-53464713 on Build GRCh38
rs943002021 Chr.1: 53464966-53464966 on Build GRCh38
rs770136132 Chr.1: 53464680-53464680 on Build GRCh38
rs770976644 Chr.1: 53464705-53464705 on Build GRCh38
rs943514 Chr.1: 53464878-53464878 on Build GRCh38
rs763770460 Chr.1: 53464822-53464822 on Build GRCh38
rs764254629 Chr.1: 53464684-53464684 on Build GRCh38
rs773403546 Chr.1: 53464625-53464626 on Build GRCh38
rs1042581326 Chr.1: 53464634-53464634 on Build GRCh38
rs769817994 Chr.1: 53464860-53464861 on Build GRCh38
rs752706098 Chr.1: 53464596-53464596 on Build GRCh38
rs113052809 Chr.1: 53464877-53464877 on Build GRCh38
rs745818002 Chr.1: 53464753-53464753 on Build GRCh38
rs773003448 Chr.1: 53464655-53464655 on Build GRCh38
rs566504124 Chr.1: 53464678-53464680 on Build GRCh38
rs371858797 Chr.1: 53465002-53465002 on Build GRCh38
rs750317515 Chr.1: 53464864-53464864 on Build GRCh38
rs758082200 Chr.1: 53464740-53464740 on Build GRCh38
rs778754413 Chr.1: 53464686-53464687 on Build GRCh38
rs766790398 Chr.1: 53464735-53464735 on Build GRCh38
rs774506748 Chr.1: 53464714-53464714 on Build GRCh38
rs930332604 Chr.1: 53464664-53464664 on Build GRCh38
rs781345887 Chr.1: 53464708-53464708 on Build GRCh38
rs150471942 Chr.1: 53464904-53464904 on Build GRCh38
rs754640485 Chr.1: 53464629-53464629 on Build GRCh38
rs748393160 Chr.1: 53464896-53464896 on Build GRCh38
rs1024990718 Chr.1: 53464782-53464782 on Build GRCh38
rs779900864 Chr.1: 53464758-53464758 on Build GRCh38
rs766613980 Chr.1: 53464677-53464678 on Build GRCh38
rs866110277 Chr.1: 53464770-53464770 on Build GRCh38
rs530331864 Chr.1: 53464683-53464684 on Build GRCh38
rs768912441 Chr.1: 53464776-53464776 on Build GRCh38
rs140725492 Chr.1: 53464630-53464630 on Build GRCh38
rs149517536 Chr.1: 53464668-53464668 on Build GRCh38
rs1047964135 Chr.1: 53464908-53464908 on Build GRCh38
rs151293595 Chr.1: 53464701-53464701 on Build GRCh38
rs573937523 Chr.1: 53464949-53464949 on Build GRCh38
rs763332208 Chr.1: 53464676-53464696 on Build GRCh38
rs865871439 Chr.1: 53464550-53464550 on Build GRCh38
rs749107239 Chr.1: 53464876-53464876 on Build GRCh38
rs769670161 Chr.1: 53464755-53464755 on Build GRCh38
rs758570286 Chr.1: 53464610-53464610 on Build GRCh38
rs1030267116 Chr.1: 53464964-53464964 on Build GRCh38
rs775817795 Chr.1: 53464681-53464681 on Build GRCh38
rs767163641 Chr.1: 53464860-53464860 on Build GRCh38
rs961546083 Chr.1: 53464792-53464792 on Build GRCh38
rs976006371 Chr.1: 53464590-53464590 on Build GRCh38
rs776625008 Chr.1: 53464786-53464787 on Build GRCh38
rs75818218 Chr.1: 53464679-53464679 on Build GRCh38

View Full Product Details