Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 53904527-53905029 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIO1
Entrez Gene ID
1733
Gene Name
deiodinase, iodothyronine type I
Gene Aliases
5DI, TXDI1
UniGene
Hs.251415
Gene Chromosome Location
Chr.1: 53894187-53911086 on Build GRCh38
Transcripts
NM_000792.6 NM_001039715.2 NM_001039716.2 NM_001324316.1 NM_213593.4 NR_136692.1 NR_136693.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4008906 c.369T>C p.S123S Chr.1: 53904697-53904697 on Build GRCh38
464769 c.441A>G p.A147A Chr.1: 53904769-53904769 on Build GRCh38
3490791 c.351G>A p.L117L Chr.1: 53904679-53904679 on Build GRCh38
122575 c.371G>T p.C124F Chr.1: 53904699-53904699 on Build GRCh38
3805573 c.426C>T p.D142D Chr.1: 53904754-53904754 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs878912273 Chr.1: 53904562-53904562 on Build GRCh38
rs2294511 Chr.1: 53904591-53904591 on Build GRCh38
rs768451416 Chr.1: 53904632-53904632 on Build GRCh38
rs776034416 Chr.1: 53904820-53904820 on Build GRCh38
rs560696782 Chr.1: 53904990-53904990 on Build GRCh38
rs985063642 Chr.1: 53904870-53904870 on Build GRCh38
rs1034381796 Chr.1: 53904642-53904642 on Build GRCh38
rs748072763 Chr.1: 53904773-53904773 on Build GRCh38
rs766074470 Chr.1: 53904837-53904837 on Build GRCh38
rs3215500 Chr.1: 53904562-53904563 on Build GRCh38
rs764544360 Chr.1: 53904826-53904826 on Build GRCh38
rs776783102 Chr.1: 53904706-53904706 on Build GRCh38
rs535371743 Chr.1: 53904572-53904572 on Build GRCh38
rs776536362 Chr.1: 53904790-53904790 on Build GRCh38
rs770983208 Chr.1: 53904685-53904685 on Build GRCh38
rs186903408 Chr.1: 53904802-53904802 on Build GRCh38
rs373695562 Chr.1: 53904821-53904821 on Build GRCh38
rs377232338 Chr.1: 53904752-53904752 on Build GRCh38
rs115485347 Chr.1: 53904555-53904555 on Build GRCh38
rs1040809340 Chr.1: 53904909-53904909 on Build GRCh38
rs891575596 Chr.1: 53904935-53904935 on Build GRCh38
rs771898897 Chr.1: 53904680-53904680 on Build GRCh38
rs992943113 Chr.1: 53904691-53904691 on Build GRCh38
rs960368620 Chr.1: 53904670-53904670 on Build GRCh38
rs746883577 Chr.1: 53904638-53904638 on Build GRCh38
rs1026531701 Chr.1: 53904555-53904555 on Build GRCh38
rs758185036 Chr.1: 53904774-53904774 on Build GRCh38
rs770068603 Chr.1: 53904760-53904760 on Build GRCh38
rs542947113 Chr.1: 53904877-53904877 on Build GRCh38
rs980900190 Chr.1: 53904581-53904581 on Build GRCh38
rs747297274 Chr.1: 53904681-53904681 on Build GRCh38
rs572396863 Chr.1: 53904884-53904884 on Build GRCh38
rs397805052 Chr.1: 53904567-53904568 on Build GRCh38
rs994994129 Chr.1: 53904963-53904963 on Build GRCh38
rs184097906 Chr.1: 53904779-53904779 on Build GRCh38
rs779517591 Chr.1: 53904639-53904642 on Build GRCh38
rs199606717 Chr.1: 53904764-53904764 on Build GRCh38
rs755891659 Chr.1: 53904753-53904753 on Build GRCh38
rs1043272119 Chr.1: 53905001-53905001 on Build GRCh38
rs765805689 Chr.1: 53904717-53904717 on Build GRCh38
rs995088845 Chr.1: 53904549-53904550 on Build GRCh38
rs775669000 Chr.1: 53904723-53904723 on Build GRCh38
rs763387241 Chr.1: 53904751-53904751 on Build GRCh38
rs759895018 Chr.1: 53904716-53904716 on Build GRCh38
rs961202045 Chr.1: 53904770-53904770 on Build GRCh38
rs1053666077 Chr.1: 53904629-53904629 on Build GRCh38
rs541233180 Chr.1: 53904634-53904634 on Build GRCh38
rs145706521 Chr.1: 53904676-53904676 on Build GRCh38
rs745984372 Chr.1: 53904798-53904798 on Build GRCh38
rs2294512 Chr.1: 53904633-53904633 on Build GRCh38
rs936475148 Chr.1: 53904608-53904608 on Build GRCh38
rs114936164 Chr.1: 53904549-53904549 on Build GRCh38
rs970488887 Chr.1: 53904563-53904563 on Build GRCh38
rs753417809 Chr.1: 53904763-53904763 on Build GRCh38
rs899337792 Chr.1: 53904916-53904916 on Build GRCh38
rs886670448 Chr.1: 53904998-53904998 on Build GRCh38
rs774873570 Chr.1: 53904835-53904835 on Build GRCh38
rs771110226 Chr.1: 53904665-53904665 on Build GRCh38
rs376673558 Chr.1: 53904836-53904836 on Build GRCh38
rs779932337 Chr.1: 53904978-53904978 on Build GRCh38
rs771144639 Chr.1: 53904789-53904789 on Build GRCh38
rs778884582 Chr.1: 53904765-53904765 on Build GRCh38
rs776543126 Chr.1: 53904855-53904858 on Build GRCh38
rs746028830 Chr.1: 53904667-53904667 on Build GRCh38
rs775343578 Chr.1: 53904882-53904882 on Build GRCh38

View Full Product Details