Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 54239032-54239529 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SSBP3
Entrez Gene ID
23648
Gene Name
single stranded DNA binding protein 3
Gene Aliases
CSDP, SSDP, SSDP1
UniGene
Hs.476706
Gene Chromosome Location
Chr.1: 54225431-54413479 on Build GRCh38
Transcripts
NM_001009955.3 NM_018070.4 NM_145716.3 XM_006710545.3 XM_017000897.1 XM_017000898.1 XM_017000899.1 NR_103541.1

Gene Symbol
SSBP3-AS1
Entrez Gene ID
619518
Gene Name
SSBP3 antisense RNA 1
Gene Aliases
C1orf191, MST128, MSTP128
UniGene
Hs.591438
Gene Chromosome Location
Chr.1: 54236443-54239063 on Build GRCh38
Transcripts
NM_001009955.3 NM_018070.4 NM_145716.3 XM_006710545.3 XM_017000897.1 XM_017000898.1 XM_017000899.1 NR_103541.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5772271 c.927+3A>G p.? Chr.1: 54239126-54239126 on Build GRCh38
7392061 c.830G>A p.G277E Chr.1: 54239145-54239145 on Build GRCh38
6263042 c.904C>A p.P302T Chr.1: 54239152-54239152 on Build GRCh38
5053591 c.803A>G p.Y268C Chr.1: 54239172-54239172 on Build GRCh38
4511852 c.882C>T p.I294I Chr.1: 54239174-54239174 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1055521490 Chr.1: 54239456-54239456 on Build GRCh38
rs200509133 Chr.1: 54239227-54239227 on Build GRCh38
rs371468900 Chr.1: 54239434-54239434 on Build GRCh38
rs913152380 Chr.1: 54239387-54239387 on Build GRCh38
rs758827003 Chr.1: 54239058-54239058 on Build GRCh38
rs1053058656 Chr.1: 54239488-54239488 on Build GRCh38
rs778255331 Chr.1: 54239061-54239061 on Build GRCh38
rs761806342 Chr.1: 54239093-54239093 on Build GRCh38
rs60925522 Chr.1: 54239156-54239156 on Build GRCh38
rs747606477 Chr.1: 54239234-54239234 on Build GRCh38
rs531170206 Chr.1: 54239467-54239467 on Build GRCh38
rs866825963 Chr.1: 54239208-54239208 on Build GRCh38
rs1029374971 Chr.1: 54239293-54239293 on Build GRCh38
rs776985241 Chr.1: 54239184-54239184 on Build GRCh38
rs775698542 Chr.1: 54239128-54239129 on Build GRCh38
rs545963869 Chr.1: 54239145-54239145 on Build GRCh38
rs752620786 Chr.1: 54239365-54239365 on Build GRCh38
rs138868760 Chr.1: 54239412-54239412 on Build GRCh38
rs760460209 Chr.1: 54239158-54239158 on Build GRCh38
rs775038905 Chr.1: 54239070-54239070 on Build GRCh38
rs767205540 Chr.1: 54239095-54239095 on Build GRCh38
rs943875425 Chr.1: 54239352-54239352 on Build GRCh38
rs375329751 Chr.1: 54239203-54239203 on Build GRCh38
rs760833387 Chr.1: 54239232-54239233 on Build GRCh38
rs918637498 Chr.1: 54239087-54239087 on Build GRCh38
rs768151058 Chr.1: 54239144-54239144 on Build GRCh38
rs139009912 Chr.1: 54239132-54239132 on Build GRCh38
rs770636084 Chr.1: 54239160-54239160 on Build GRCh38
rs148337946 Chr.1: 54239194-54239194 on Build GRCh38
rs953658670 Chr.1: 54239322-54239322 on Build GRCh38
rs895995117 Chr.1: 54239465-54239465 on Build GRCh38
rs754676836 Chr.1: 54239226-54239226 on Build GRCh38
rs765382620 Chr.1: 54239198-54239198 on Build GRCh38
rs768171576 Chr.1: 54239091-54239091 on Build GRCh38
rs748751158 Chr.1: 54239072-54239072 on Build GRCh38
rs533940033 Chr.1: 54239129-54239129 on Build GRCh38
rs1041008869 Chr.1: 54239378-54239378 on Build GRCh38
rs189006328 Chr.1: 54239418-54239418 on Build GRCh38
rs745483736 Chr.1: 54239063-54239063 on Build GRCh38
rs904150484 Chr.1: 54239423-54239423 on Build GRCh38
rs904264211 Chr.1: 54239060-54239060 on Build GRCh38
rs201671456 Chr.1: 54239211-54239211 on Build GRCh38
rs370171475 Chr.1: 54239130-54239130 on Build GRCh38
rs377053655 Chr.1: 54239103-54239103 on Build GRCh38
rs1015163127 Chr.1: 54239478-54239478 on Build GRCh38
rs753476139 Chr.1: 54239223-54239223 on Build GRCh38
rs142781098 Chr.1: 54239499-54239499 on Build GRCh38
rs898088605 Chr.1: 54239480-54239480 on Build GRCh38
rs962584603 Chr.1: 54239409-54239409 on Build GRCh38
rs564506877 Chr.1: 54239417-54239417 on Build GRCh38
rs777699898 Chr.1: 54239240-54239240 on Build GRCh38
rs367861609 Chr.1: 54239205-54239205 on Build GRCh38
rs992195397 Chr.1: 54239457-54239457 on Build GRCh38
rs201180203 Chr.1: 54239209-54239209 on Build GRCh38
rs757589176 Chr.1: 54239138-54239138 on Build GRCh38
rs376946839 Chr.1: 54239122-54239122 on Build GRCh38
rs375752123 Chr.1: 54239248-54239248 on Build GRCh38
rs551985363 Chr.1: 54239433-54239433 on Build GRCh38
rs755038737 Chr.1: 54239120-54239120 on Build GRCh38
rs748057892 Chr.1: 54239150-54239150 on Build GRCh38
rs759798595 Chr.1: 54239251-54239251 on Build GRCh38
rs763906319 Chr.1: 54239473-54239473 on Build GRCh38
rs562186917 Chr.1: 54239466-54239466 on Build GRCh38
rs760416321 Chr.1: 54239105-54239105 on Build GRCh38
rs754019495 Chr.1: 54239119-54239119 on Build GRCh38
rs997919030 Chr.1: 54239288-54239288 on Build GRCh38
rs376505077 Chr.1: 54239241-54239241 on Build GRCh38
rs1006809485 Chr.1: 54239356-54239356 on Build GRCh38
rs369737592 Chr.1: 54239067-54239067 on Build GRCh38
rs766571764 Chr.1: 54239115-54239115 on Build GRCh38
rs771494482 Chr.1: 54239235-54239235 on Build GRCh38
rs548079391 Chr.1: 54239476-54239476 on Build GRCh38
rs748945116 Chr.1: 54239143-54239143 on Build GRCh38
rs747361260 Chr.1: 54239133-54239133 on Build GRCh38
rs772004232 Chr.1: 54239151-54239151 on Build GRCh38
rs776615115 Chr.1: 54239250-54239250 on Build GRCh38
rs201358342 Chr.1: 54239104-54239104 on Build GRCh38
rs527910634 Chr.1: 54239280-54239280 on Build GRCh38
rs781476632 Chr.1: 54239218-54239218 on Build GRCh38
rs779702779 Chr.1: 54239141-54239141 on Build GRCh38

View Full Product Details