Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 54807441-54807946 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LEXM
Entrez Gene ID
163747
Gene Name
lymphocyte expansion molecule
Gene Aliases
C1orf177, LEM
UniGene
Hs.376018
Gene Chromosome Location
Chr.1: 54806063-54842264 on Build GRCh38
Transcripts
NM_001110533.1 NM_152607.2 XM_017000480.1 XM_017000481.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3751314 c.376G>T p.G126C Chr.1: 54807907-54807907 on Build GRCh38
6322042 c.413G>T p.R138L Chr.1: 54807944-54807944 on Build GRCh38
4841721 c.252G>A p.L84L Chr.1: 54807585-54807585 on Build GRCh38
4667347 c.295A>G p.T99A Chr.1: 54807628-54807628 on Build GRCh38
4008989 c.332C>T p.A111V Chr.1: 54807665-54807665 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768042866 Chr.1: 54807871-54807875 on Build GRCh38
rs745841866 Chr.1: 54807921-54807921 on Build GRCh38
rs12404626 Chr.1: 54807713-54807713 on Build GRCh38
rs759026106 Chr.1: 54807487-54807487 on Build GRCh38
rs558450851 Chr.1: 54807661-54807661 on Build GRCh38
rs752970377 Chr.1: 54807639-54807639 on Build GRCh38
rs773135491 Chr.1: 54807536-54807536 on Build GRCh38
rs757647863 Chr.1: 54807575-54807575 on Build GRCh38
rs114632512 Chr.1: 54807486-54807486 on Build GRCh38
rs559366610 Chr.1: 54807816-54807816 on Build GRCh38
rs775806461 Chr.1: 54807722-54807722 on Build GRCh38
rs902580928 Chr.1: 54807549-54807549 on Build GRCh38
rs754055679 Chr.1: 54807553-54807553 on Build GRCh38
rs1014104519 Chr.1: 54807758-54807758 on Build GRCh38
rs372048254 Chr.1: 54807593-54807593 on Build GRCh38
rs192746435 Chr.1: 54807765-54807765 on Build GRCh38
rs373507359 Chr.1: 54807904-54807904 on Build GRCh38
rs753560945 Chr.1: 54807524-54807524 on Build GRCh38
rs1014819055 Chr.1: 54807771-54807771 on Build GRCh38
rs767508565 Chr.1: 54807535-54807535 on Build GRCh38
rs1041230634 Chr.1: 54807910-54807910 on Build GRCh38
rs760402084 Chr.1: 54807538-54807538 on Build GRCh38
rs549458783 Chr.1: 54807469-54807469 on Build GRCh38
rs757887341 Chr.1: 54807675-54807675 on Build GRCh38
rs766179749 Chr.1: 54807552-54807552 on Build GRCh38
rs996599094 Chr.1: 54807875-54807875 on Build GRCh38
rs765447741 Chr.1: 54807638-54807638 on Build GRCh38
rs577954332 Chr.1: 54807678-54807678 on Build GRCh38
rs775905023 Chr.1: 54807875-54807876 on Build GRCh38
rs557397842 Chr.1: 54807693-54807693 on Build GRCh38
rs908414336 Chr.1: 54807694-54807694 on Build GRCh38
rs535179886 Chr.1: 54807490-54807490 on Build GRCh38
rs1027680150 Chr.1: 54807877-54807877 on Build GRCh38
rs919965012 Chr.1: 54807817-54807817 on Build GRCh38
rs377396512 Chr.1: 54807510-54807510 on Build GRCh38
rs1042566302 Chr.1: 54807777-54807777 on Build GRCh38
rs778894499 Chr.1: 54807479-54807479 on Build GRCh38
rs377604678 Chr.1: 54807729-54807729 on Build GRCh38
rs961982077 Chr.1: 54807775-54807775 on Build GRCh38
rs781449937 Chr.1: 54807580-54807580 on Build GRCh38
rs985916415 Chr.1: 54807837-54807837 on Build GRCh38
rs752310766 Chr.1: 54807903-54807903 on Build GRCh38
rs907787777 Chr.1: 54807502-54807502 on Build GRCh38
rs555514148 Chr.1: 54807528-54807528 on Build GRCh38
rs778369186 Chr.1: 54807687-54807687 on Build GRCh38
rs766169578 Chr.1: 54807632-54807632 on Build GRCh38
rs747818355 Chr.1: 54807692-54807692 on Build GRCh38
rs778359724 Chr.1: 54807565-54807565 on Build GRCh38
rs773905978 Chr.1: 54807603-54807603 on Build GRCh38
rs202224828 Chr.1: 54807682-54807682 on Build GRCh38
rs756382202 Chr.1: 54807883-54807883 on Build GRCh38
rs370704363 Chr.1: 54807558-54807558 on Build GRCh38
rs758971123 Chr.1: 54807564-54807564 on Build GRCh38
rs947012447 Chr.1: 54807764-54807764 on Build GRCh38
rs112841482 Chr.1: 54807630-54807630 on Build GRCh38
rs761239382 Chr.1: 54807910-54807910 on Build GRCh38
rs751747228 Chr.1: 54807849-54807849 on Build GRCh38
rs759844289 Chr.1: 54807704-54807705 on Build GRCh38
rs671022 Chr.1: 54807749-54807749 on Build GRCh38
rs774745558 Chr.1: 54807668-54807669 on Build GRCh38
rs201438425 Chr.1: 54807683-54807683 on Build GRCh38
rs369452055 Chr.1: 54807633-54807633 on Build GRCh38
rs780935770 Chr.1: 54807911-54807911 on Build GRCh38
rs376513670 Chr.1: 54807615-54807615 on Build GRCh38
rs146088179 Chr.1: 54807556-54807556 on Build GRCh38
rs142483178 Chr.1: 54807631-54807631 on Build GRCh38
rs762404687 Chr.1: 54807865-54807865 on Build GRCh38
rs376208136 Chr.1: 54807846-54807846 on Build GRCh38
rs752779858 Chr.1: 54807560-54807560 on Build GRCh38
rs9782980 Chr.1: 54807907-54807907 on Build GRCh38
rs187251221 Chr.1: 54807468-54807468 on Build GRCh38
rs747446097 Chr.1: 54807491-54807491 on Build GRCh38
rs770834399 Chr.1: 54807712-54807712 on Build GRCh38
rs747533556 Chr.1: 54807571-54807571 on Build GRCh38
rs766173655 Chr.1: 54807495-54807495 on Build GRCh38
rs267598656 Chr.1: 54807622-54807622 on Build GRCh38
rs952820811 Chr.1: 54807825-54807825 on Build GRCh38
rs373134177 Chr.1: 54807721-54807721 on Build GRCh38
rs574133995 Chr.1: 54807708-54807708 on Build GRCh38
rs750950498 Chr.1: 54807873-54807873 on Build GRCh38
rs748915104 Chr.1: 54807523-54807523 on Build GRCh38
rs1009830017 Chr.1: 54807637-54807637 on Build GRCh38
rs771109663 Chr.1: 54807663-54807666 on Build GRCh38
rs1046035833 Chr.1: 54807475-54807475 on Build GRCh38
rs868510092 Chr.1: 54807908-54807908 on Build GRCh38
rs777301046 Chr.1: 54807697-54807697 on Build GRCh38
rs368525051 Chr.1: 54807629-54807629 on Build GRCh38
rs764292945 Chr.1: 54807666-54807666 on Build GRCh38
rs369155828 Chr.1: 54807872-54807872 on Build GRCh38
rs777255972 Chr.1: 54807906-54807906 on Build GRCh38
rs540966717 Chr.1: 54807778-54807778 on Build GRCh38
rs764246262 Chr.1: 54807731-54807731 on Build GRCh38
rs748948493 Chr.1: 54807591-54807591 on Build GRCh38
rs761794966 Chr.1: 54807531-54807531 on Build GRCh38
rs1019048339 Chr.1: 54807819-54807819 on Build GRCh38
rs752073834 Chr.1: 54807671-54807671 on Build GRCh38
rs1021024984 Chr.1: 54807539-54807539 on Build GRCh38
rs902716003 Chr.1: 54807784-54807784 on Build GRCh38
rs944230387 Chr.1: 54807619-54807619 on Build GRCh38
rs759793816 Chr.1: 54807636-54807636 on Build GRCh38
rs780494871 Chr.1: 54807897-54807897 on Build GRCh38

View Full Product Details