Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 54851356-54851852 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHCR24
Entrez Gene ID
1718
Gene Name
24-dehydrocholesterol reductase
Gene Aliases
DCE, Nbla03646, SELADIN1, seladin-1
UniGene
Hs.498727
Gene Chromosome Location
Chr.1: 54849627-54887248 on Build GRCh38
Transcripts
NM_014762.3

SNP Detail

SNP ID Chromosome Location
rs950769698 Chr.1: 54851650-54851650 on Build GRCh38
rs150093568 Chr.1: 54851548-54851548 on Build GRCh38
rs74072035 Chr.1: 54851515-54851515 on Build GRCh38
rs929289591 Chr.1: 54851401-54851401 on Build GRCh38
rs974912054 Chr.1: 54851733-54851733 on Build GRCh38
rs370735535 Chr.1: 54851484-54851484 on Build GRCh38
rs1023385388 Chr.1: 54851589-54851589 on Build GRCh38
rs922164561 Chr.1: 54851742-54851742 on Build GRCh38
rs915909499 Chr.1: 54851496-54851496 on Build GRCh38
rs925164869 Chr.1: 54851516-54851516 on Build GRCh38
rs1003495409 Chr.1: 54851622-54851622 on Build GRCh38
rs988089845 Chr.1: 54851660-54851660 on Build GRCh38
rs1050208266 Chr.1: 54851558-54851558 on Build GRCh38
rs1032075334 Chr.1: 54851553-54851553 on Build GRCh38
rs57714270 Chr.1: 54851656-54851656 on Build GRCh38
rs552112143 Chr.1: 54851411-54851411 on Build GRCh38
rs1013010146 Chr.1: 54851811-54851811 on Build GRCh38
rs576670614 Chr.1: 54851753-54851753 on Build GRCh38
rs886046417 Chr.1: 54851799-54851799 on Build GRCh38
rs897525701 Chr.1: 54851784-54851784 on Build GRCh38
rs372344030 Chr.1: 54851580-54851580 on Build GRCh38
rs648804 Chr.1: 54851771-54851771 on Build GRCh38
rs900765940 Chr.1: 54851464-54851464 on Build GRCh38
rs539554514 Chr.1: 54851760-54851760 on Build GRCh38
rs895976219 Chr.1: 54851810-54851810 on Build GRCh38
rs1023108786 Chr.1: 54851812-54851812 on Build GRCh38
rs970637658 Chr.1: 54851380-54851380 on Build GRCh38
rs923094960 Chr.1: 54851679-54851679 on Build GRCh38
rs56279713 Chr.1: 54851752-54851752 on Build GRCh38
rs569294492 Chr.1: 54851424-54851424 on Build GRCh38
rs1023035010 Chr.1: 54851614-54851614 on Build GRCh38
rs767726171 Chr.1: 54851611-54851611 on Build GRCh38
rs935279982 Chr.1: 54851539-54851539 on Build GRCh38
rs192813255 Chr.1: 54851559-54851559 on Build GRCh38
rs928691646 Chr.1: 54851395-54851395 on Build GRCh38
rs1037383604 Chr.1: 54851581-54851581 on Build GRCh38
rs933162457 Chr.1: 54851688-54851688 on Build GRCh38
rs9436941 Chr.1: 54851445-54851445 on Build GRCh38
rs762917924 Chr.1: 54851557-54851557 on Build GRCh38
rs1057340141 Chr.1: 54851809-54851809 on Build GRCh38
rs898034075 Chr.1: 54851586-54851586 on Build GRCh38
rs902212193 Chr.1: 54851547-54851547 on Build GRCh38
rs752767678 Chr.1: 54851822-54851822 on Build GRCh38
rs888764317 Chr.1: 54851570-54851570 on Build GRCh38
rs963425278 Chr.1: 54851732-54851732 on Build GRCh38
rs764347154 Chr.1: 54851390-54851390 on Build GRCh38
rs1050157177 Chr.1: 54851723-54851725 on Build GRCh38
rs765112724 Chr.1: 54851426-54851426 on Build GRCh38
rs970963332 Chr.1: 54851606-54851606 on Build GRCh38
rs935016967 Chr.1: 54851819-54851819 on Build GRCh38
rs116289873 Chr.1: 54851502-54851502 on Build GRCh38
rs756116799 Chr.1: 54851675-54851675 on Build GRCh38
rs761848598 Chr.1: 54851697-54851699 on Build GRCh38
rs558278303 Chr.1: 54851785-54851785 on Build GRCh38
rs187950024 Chr.1: 54851391-54851391 on Build GRCh38
rs751850671 Chr.1: 54851410-54851410 on Build GRCh38
rs886046416 Chr.1: 54851725-54851725 on Build GRCh38
rs1037741650 Chr.1: 54851777-54851777 on Build GRCh38
rs962222013 Chr.1: 54851422-54851422 on Build GRCh38
rs545646276 Chr.1: 54851655-54851655 on Build GRCh38
rs576893984 Chr.1: 54851613-54851613 on Build GRCh38
rs184125615 Chr.1: 54851633-54851633 on Build GRCh38

View Full Product Details