Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 54871258-54871757 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHCR24
Entrez Gene ID
1718
Gene Name
24-dehydrocholesterol reductase
Gene Aliases
DCE, Nbla03646, SELADIN1, seladin-1
UniGene
Hs.498727
Gene Chromosome Location
Chr.1: 54849627-54887248 on Build GRCh38
Transcripts
NM_014762.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4755619 c.747G>T p.E249D Chr.1: 54871479-54871479 on Build GRCh38
5847016 c.640G>A p.V214I Chr.1: 54871586-54871586 on Build GRCh38
4009005 c.785A>G p.Q262R Chr.1: 54871441-54871441 on Build GRCh38
4755620 c.692T>C p.I231T Chr.1: 54871534-54871534 on Build GRCh38
4009006 c.736C>T p.R246W Chr.1: 54871490-54871490 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771211483 Chr.1: 54871427-54871427 on Build GRCh38
rs750059090 Chr.1: 54871358-54871358 on Build GRCh38
rs189200040 Chr.1: 54871361-54871361 on Build GRCh38
rs145971053 Chr.1: 54871515-54871515 on Build GRCh38
rs114852984 Chr.1: 54871678-54871678 on Build GRCh38
rs772464402 Chr.1: 54871548-54871548 on Build GRCh38
rs758190462 Chr.1: 54871407-54871407 on Build GRCh38
rs2303535 Chr.1: 54871707-54871707 on Build GRCh38
rs913546908 Chr.1: 54871285-54871285 on Build GRCh38
rs375033508 Chr.1: 54871340-54871340 on Build GRCh38
rs1020891543 Chr.1: 54871297-54871297 on Build GRCh38
rs886042137 Chr.1: 54871415-54871415 on Build GRCh38
rs201789591 Chr.1: 54871331-54871331 on Build GRCh38
rs1019107299 Chr.1: 54871530-54871530 on Build GRCh38
rs138667252 Chr.1: 54871495-54871495 on Build GRCh38
rs764700447 Chr.1: 54871496-54871496 on Build GRCh38
rs778868306 Chr.1: 54871391-54871391 on Build GRCh38
rs137897500 Chr.1: 54871301-54871301 on Build GRCh38
rs754465094 Chr.1: 54871386-54871386 on Build GRCh38
rs150166147 Chr.1: 54871490-54871490 on Build GRCh38
rs143779194 Chr.1: 54871587-54871587 on Build GRCh38
rs750647923 Chr.1: 54871628-54871628 on Build GRCh38
rs545948766 Chr.1: 54871680-54871680 on Build GRCh38
rs762174947 Chr.1: 54871484-54871484 on Build GRCh38
rs200217334 Chr.1: 54871499-54871499 on Build GRCh38
rs2303534 Chr.1: 54871642-54871642 on Build GRCh38
rs902454280 Chr.1: 54871680-54871680 on Build GRCh38
rs528459787 Chr.1: 54871706-54871706 on Build GRCh38
rs752614299 Chr.1: 54871400-54871400 on Build GRCh38
rs138043637 Chr.1: 54871500-54871500 on Build GRCh38
rs142421002 Chr.1: 54871428-54871428 on Build GRCh38
rs886046421 Chr.1: 54871514-54871514 on Build GRCh38
rs764133417 Chr.1: 54871504-54871504 on Build GRCh38
rs776917991 Chr.1: 54871311-54871311 on Build GRCh38
rs770279004 Chr.1: 54871322-54871322 on Build GRCh38
rs762400676 Chr.1: 54871598-54871598 on Build GRCh38
rs987353276 Chr.1: 54871368-54871368 on Build GRCh38
rs766019921 Chr.1: 54871367-54871367 on Build GRCh38
rs919273988 Chr.1: 54871699-54871699 on Build GRCh38
rs780333161 Chr.1: 54871637-54871637 on Build GRCh38
rs764486755 Chr.1: 54871353-54871353 on Build GRCh38
rs759168678 Chr.1: 54871561-54871561 on Build GRCh38
rs186533929 Chr.1: 54871444-54871444 on Build GRCh38
rs200735479 Chr.1: 54871389-54871389 on Build GRCh38
rs147896894 Chr.1: 54871445-54871445 on Build GRCh38
rs755078643 Chr.1: 54871644-54871644 on Build GRCh38
rs608458 Chr.1: 54871277-54871277 on Build GRCh38
rs756361953 Chr.1: 54871630-54871630 on Build GRCh38
rs995887297 Chr.1: 54871715-54871715 on Build GRCh38
rs764798976 Chr.1: 54871566-54871566 on Build GRCh38
rs768999622 Chr.1: 54871475-54871475 on Build GRCh38
rs779358484 Chr.1: 54871647-54871647 on Build GRCh38
rs776036482 Chr.1: 54871454-54871454 on Build GRCh38
rs201021800 Chr.1: 54871411-54871411 on Build GRCh38
rs748700105 Chr.1: 54871648-54871648 on Build GRCh38
rs763423346 Chr.1: 54871461-54871461 on Build GRCh38
rs180903448 Chr.1: 54871599-54871599 on Build GRCh38
rs147656722 Chr.1: 54871489-54871489 on Build GRCh38
rs376885844 Chr.1: 54871538-54871538 on Build GRCh38
rs572047085 Chr.1: 54871505-54871505 on Build GRCh38
rs759541207 Chr.1: 54871320-54871320 on Build GRCh38
rs201280799 Chr.1: 54871547-54871547 on Build GRCh38
rs191246223 Chr.1: 54871508-54871508 on Build GRCh38
rs771121325 Chr.1: 54871302-54871302 on Build GRCh38
rs370233954 Chr.1: 54871643-54871643 on Build GRCh38
rs147213053 Chr.1: 54871611-54871611 on Build GRCh38
rs375614612 Chr.1: 54871662-54871662 on Build GRCh38
rs889196742 Chr.1: 54871514-54871514 on Build GRCh38
rs375934272 Chr.1: 54871377-54871377 on Build GRCh38
rs759137429 Chr.1: 54871541-54871541 on Build GRCh38
rs150688144 Chr.1: 54871610-54871610 on Build GRCh38
rs139813073 Chr.1: 54871537-54871537 on Build GRCh38
rs763339091 Chr.1: 54871350-54871350 on Build GRCh38
rs926459170 Chr.1: 54871345-54871345 on Build GRCh38
rs1010206690 Chr.1: 54871586-54871586 on Build GRCh38
rs756200011 Chr.1: 54871522-54871522 on Build GRCh38
rs181555625 Chr.1: 54871379-54871379 on Build GRCh38
rs772411201 Chr.1: 54871653-54871653 on Build GRCh38
rs776322733 Chr.1: 54871557-54871557 on Build GRCh38
rs774628804 Chr.1: 54871476-54871476 on Build GRCh38
rs556751117 Chr.1: 54871284-54871284 on Build GRCh38
rs762566563 Chr.1: 54871591-54871591 on Build GRCh38

View Full Product Details