Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 55007937-55008466 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BSND
Entrez Gene ID
7809
Gene Name
barttin CLCNK type accessory beta subunit
Gene Aliases
BART, DFNB73
UniGene
Hs.151291
Gene Chromosome Location
Chr.1: 54998944-55008792 on Build GRCh38
Transcripts
NM_057176.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5969488 c.587C>T p.S196F Chr.1: 55008252-55008252 on Build GRCh38
7296290 c.781A>G p.I261V Chr.1: 55008446-55008446 on Build GRCh38
3865851 c.609G>A p.M203I Chr.1: 55008274-55008274 on Build GRCh38
426500 c.785C>T p.A262V Chr.1: 55008450-55008450 on Build GRCh38
1734910 c.701C>T p.P234L Chr.1: 55008366-55008366 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773375286 Chr.1: 55008215-55008215 on Build GRCh38
rs887851650 Chr.1: 55008085-55008085 on Build GRCh38
rs886046423 Chr.1: 55008269-55008269 on Build GRCh38
rs568944655 Chr.1: 55008119-55008119 on Build GRCh38
rs778789684 Chr.1: 55008299-55008299 on Build GRCh38
rs926142735 Chr.1: 55008402-55008402 on Build GRCh38
rs765819444 Chr.1: 55008259-55008259 on Build GRCh38
rs368804400 Chr.1: 55008374-55008374 on Build GRCh38
rs764627159 Chr.1: 55008165-55008165 on Build GRCh38
rs141880409 Chr.1: 55008312-55008312 on Build GRCh38
rs763366653 Chr.1: 55008264-55008264 on Build GRCh38
rs770850590 Chr.1: 55008240-55008240 on Build GRCh38
rs778736351 Chr.1: 55008178-55008178 on Build GRCh38
rs202112062 Chr.1: 55008340-55008340 on Build GRCh38
rs781440424 Chr.1: 55008394-55008394 on Build GRCh38
rs776770864 Chr.1: 55008245-55008245 on Build GRCh38
rs977790666 Chr.1: 55008175-55008175 on Build GRCh38
rs769271244 Chr.1: 55008413-55008413 on Build GRCh38
rs376582654 Chr.1: 55008431-55008431 on Build GRCh38
rs1014323811 Chr.1: 55008192-55008192 on Build GRCh38
rs1027496341 Chr.1: 55008307-55008307 on Build GRCh38
rs758133850 Chr.1: 55008301-55008301 on Build GRCh38
rs961327092 Chr.1: 55008113-55008113 on Build GRCh38
rs777491010 Chr.1: 55008386-55008386 on Build GRCh38
rs769932971 Chr.1: 55008403-55008403 on Build GRCh38
rs767298542 Chr.1: 55008265-55008265 on Build GRCh38
rs775854681 Chr.1: 55008148-55008148 on Build GRCh38
rs75478926 Chr.1: 55008129-55008129 on Build GRCh38
rs897566847 Chr.1: 55008275-55008275 on Build GRCh38
rs147394986 Chr.1: 55008380-55008380 on Build GRCh38
rs765871427 Chr.1: 55008281-55008281 on Build GRCh38
rs372172746 Chr.1: 55008396-55008396 on Build GRCh38
rs760301805 Chr.1: 55008368-55008368 on Build GRCh38
rs563130435 Chr.1: 55008366-55008366 on Build GRCh38
rs538456748 Chr.1: 55008102-55008102 on Build GRCh38
rs570883274 Chr.1: 55008068-55008068 on Build GRCh38
rs752564097 Chr.1: 55008378-55008378 on Build GRCh38
rs747914501 Chr.1: 55008190-55008190 on Build GRCh38
rs965444829 Chr.1: 55008363-55008363 on Build GRCh38
rs564677385 Chr.1: 55007985-55007985 on Build GRCh38
rs753374999 Chr.1: 55008285-55008285 on Build GRCh38
rs767943885 Chr.1: 55008146-55008146 on Build GRCh38
rs547525254 Chr.1: 55008035-55008035 on Build GRCh38
rs746800403 Chr.1: 55008391-55008391 on Build GRCh38
rs953160292 Chr.1: 55008316-55008316 on Build GRCh38
rs552319963 Chr.1: 55008106-55008106 on Build GRCh38
rs796666396 Chr.1: 55008432-55008432 on Build GRCh38
rs754819005 Chr.1: 55008409-55008409 on Build GRCh38
rs749985123 Chr.1: 55008236-55008236 on Build GRCh38
rs900741588 Chr.1: 55008304-55008304 on Build GRCh38
rs148609746 Chr.1: 55008428-55008428 on Build GRCh38
rs144028158 Chr.1: 55008003-55008003 on Build GRCh38
rs886046424 Chr.1: 55008361-55008361 on Build GRCh38
rs554899983 Chr.1: 55008160-55008160 on Build GRCh38
rs376253031 Chr.1: 55008355-55008355 on Build GRCh38
rs776088488 Chr.1: 55008350-55008350 on Build GRCh38
rs749981870 Chr.1: 55008277-55008277 on Build GRCh38
rs149472387 Chr.1: 55008392-55008392 on Build GRCh38
rs904666101 Chr.1: 55008147-55008147 on Build GRCh38
rs996325835 Chr.1: 55008019-55008019 on Build GRCh38
rs780708480 Chr.1: 55008408-55008408 on Build GRCh38
rs200071888 Chr.1: 55008300-55008300 on Build GRCh38
rs776289912 Chr.1: 55008439-55008439 on Build GRCh38
rs200886926 Chr.1: 55008262-55008262 on Build GRCh38
rs772313634 Chr.1: 55008205-55008205 on Build GRCh38
rs986062867 Chr.1: 55008357-55008357 on Build GRCh38
rs1018862150 Chr.1: 55008361-55008361 on Build GRCh38
rs368273749 Chr.1: 55008235-55008235 on Build GRCh38
rs760023611 Chr.1: 55008252-55008252 on Build GRCh38
rs138955530 Chr.1: 55008375-55008375 on Build GRCh38
rs915034319 Chr.1: 55008164-55008164 on Build GRCh38
rs111572097 Chr.1: 55008040-55008041 on Build GRCh38
rs766033314 Chr.1: 55008372-55008372 on Build GRCh38
rs755650865 Chr.1: 55008279-55008279 on Build GRCh38
rs561771976 Chr.1: 55007992-55007992 on Build GRCh38
rs754955966 Chr.1: 55008296-55008296 on Build GRCh38
rs745963643 Chr.1: 55008395-55008395 on Build GRCh38
rs1034836498 Chr.1: 55008145-55008145 on Build GRCh38
rs199696535 Chr.1: 55008435-55008435 on Build GRCh38
rs986010163 Chr.1: 55007983-55007983 on Build GRCh38
rs770459093 Chr.1: 55008342-55008342 on Build GRCh38
rs377334497 Chr.1: 55008211-55008211 on Build GRCh38
rs373384432 Chr.1: 55008107-55008107 on Build GRCh38
rs747263212 Chr.1: 55008315-55008315 on Build GRCh38
rs776628376 Chr.1: 55008339-55008339 on Build GRCh38
rs887870715 Chr.1: 55008353-55008353 on Build GRCh38
rs776107884 Chr.1: 55008338-55008338 on Build GRCh38
rs148085906 Chr.1: 55008367-55008367 on Build GRCh38
rs953276316 Chr.1: 55007981-55007981 on Build GRCh38

View Full Product Details