Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 47152625-47153127 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf61
Entrez Gene ID
285051
Gene Name
chromosome 2 open reading frame 61
Gene Aliases
-
UniGene
Hs.531575
Gene Chromosome Location
Chr.2: 47086991-47155378 on Build GRCh38
Transcripts
NM_001163561.1 NM_173649.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1752575 c.104C>G p.S35C Chr.2: 47152994-47152994 on Build GRCh38
3581868 c.90C>T p.A30A Chr.2: 47153008-47153008 on Build GRCh38
6467273 c.130A>G p.I44V Chr.2: 47152968-47152968 on Build GRCh38
3581867 c.90C>T p.A30A Chr.2: 47153008-47153008 on Build GRCh38
6467272 c.130A>G p.I44V Chr.2: 47152968-47152968 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745446170 Chr.2: 47152922-47152922 on Build GRCh38
rs768608020 Chr.2: 47152949-47152949 on Build GRCh38
rs369277550 Chr.2: 47152987-47152987 on Build GRCh38
rs115781850 Chr.2: 47152936-47152936 on Build GRCh38
rs755294433 Chr.2: 47152951-47152951 on Build GRCh38
rs759775865 Chr.2: 47153002-47153002 on Build GRCh38
rs750158173 Chr.2: 47153031-47153031 on Build GRCh38
rs988505388 Chr.2: 47152689-47152689 on Build GRCh38
rs899641221 Chr.2: 47152773-47152773 on Build GRCh38
rs780035502 Chr.2: 47153038-47153038 on Build GRCh38
rs560466819 Chr.2: 47152900-47152900 on Build GRCh38
rs755807146 Chr.2: 47153037-47153037 on Build GRCh38
rs780843512 Chr.2: 47153029-47153029 on Build GRCh38
rs769777749 Chr.2: 47152931-47152931 on Build GRCh38
rs773430353 Chr.2: 47152970-47152970 on Build GRCh38
rs533168254 Chr.2: 47152861-47152861 on Build GRCh38
rs55917773 Chr.2: 47152786-47152786 on Build GRCh38
rs572374815 Chr.2: 47152660-47152660 on Build GRCh38
rs747974764 Chr.2: 47153061-47153061 on Build GRCh38
rs780909648 Chr.2: 47152917-47152917 on Build GRCh38
rs778925573 Chr.2: 47153054-47153054 on Build GRCh38
rs979113986 Chr.2: 47153089-47153089 on Build GRCh38
rs815810 Chr.2: 47152748-47152748 on Build GRCh38
rs112521810 Chr.2: 47153034-47153034 on Build GRCh38
rs756793816 Chr.2: 47153025-47153025 on Build GRCh38
rs1042958093 Chr.2: 47152780-47152780 on Build GRCh38
rs996135911 Chr.2: 47152832-47152832 on Build GRCh38
rs1015513124 Chr.2: 47152827-47152827 on Build GRCh38
rs751459495 Chr.2: 47152980-47152982 on Build GRCh38
rs1043569424 Chr.2: 47152765-47152765 on Build GRCh38
rs815809 Chr.2: 47153064-47153064 on Build GRCh38
rs1016488075 Chr.2: 47152948-47152948 on Build GRCh38
rs913814313 Chr.2: 47152747-47152747 on Build GRCh38
rs181752950 Chr.2: 47152764-47152764 on Build GRCh38
rs996956148 Chr.2: 47152775-47152775 on Build GRCh38
rs960396340 Chr.2: 47152878-47152878 on Build GRCh38
rs764946158 Chr.2: 47152653-47152654 on Build GRCh38
rs962933916 Chr.2: 47152829-47152829 on Build GRCh38
rs377141569 Chr.2: 47152965-47152965 on Build GRCh38
rs572890103 Chr.2: 47152996-47152996 on Build GRCh38
rs545196321 Chr.2: 47152885-47152885 on Build GRCh38
rs749127246 Chr.2: 47153048-47153048 on Build GRCh38
rs753086145 Chr.2: 47153008-47153008 on Build GRCh38
rs17036300 Chr.2: 47153006-47153006 on Build GRCh38
rs567658237 Chr.2: 47152998-47152998 on Build GRCh38
rs970617306 Chr.2: 47152853-47152853 on Build GRCh38
rs749287998 Chr.2: 47152940-47152940 on Build GRCh38
rs756670155 Chr.2: 47152910-47152910 on Build GRCh38
rs1057347544 Chr.2: 47152837-47152837 on Build GRCh38
rs760886753 Chr.2: 47152979-47152979 on Build GRCh38
rs923022208 Chr.2: 47152650-47152650 on Build GRCh38
rs774117936 Chr.2: 47152955-47152955 on Build GRCh38
rs980569503 Chr.2: 47152858-47152858 on Build GRCh38
rs1013615168 Chr.2: 47152869-47152869 on Build GRCh38
rs373367018 Chr.2: 47153050-47153050 on Build GRCh38
rs896596560 Chr.2: 47152851-47152851 on Build GRCh38
rs1023144202 Chr.2: 47152848-47152848 on Build GRCh38
rs539843999 Chr.2: 47152715-47152715 on Build GRCh38
rs937164231 Chr.2: 47152799-47152799 on Build GRCh38
rs912895580 Chr.2: 47152724-47152724 on Build GRCh38
rs1026311386 Chr.2: 47152875-47152875 on Build GRCh38
rs560403618 Chr.2: 47153065-47153065 on Build GRCh38
rs761976769 Chr.2: 47152963-47152963 on Build GRCh38
rs144588020 Chr.2: 47152961-47152961 on Build GRCh38
rs1004168003 Chr.2: 47152825-47152825 on Build GRCh38
rs891151054 Chr.2: 47152804-47152804 on Build GRCh38
rs544292125 Chr.2: 47152784-47152784 on Build GRCh38
rs571619138 Chr.2: 47153076-47153076 on Build GRCh38

View Full Product Details