Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 55173961-55174501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLHC1
Entrez Gene ID
130162
Gene Name
clathrin heavy chain linker domain containing 1
Gene Aliases
C2orf63
UniGene
Hs.347014
Gene Chromosome Location
Chr.2: 55172547-55232563 on Build GRCh38
Transcripts
NM_001135598.1 NM_152385.2 XM_005264136.2 XM_006711937.3 XM_006711938.3 XM_006711939.3 XM_011532519.2 XM_011532520.2 XM_011532521.2 XM_011532522.2 XM_011532523.2 XM_011532524.1 XM_011532525.2 XM_011532527.2 XM_011532528.1 XM_011532529.2 XM_011532530.1 XM_011532534.1 XM_011532535.2 XM_017003333.1 XM_017003334.1 XM_017003335.1 XM_017003336.1 XM_017003337.1 XM_017003338.1 XM_017003339.1 XM_017003340.1 XM_017003341.1 XM_017003342.1 XM_017003343.1 XM_017003344.1 XM_017003345.1 XM_017003346.1 XM_017003347.1 XM_017003349.1 XM_017003350.1

SNP Detail

SNP ID Chromosome Location
rs991456332 Chr.2: 55174051-55174051 on Build GRCh38
rs929096101 Chr.2: 55174223-55174223 on Build GRCh38
rs547952674 Chr.2: 55174435-55174435 on Build GRCh38
rs1022670330 Chr.2: 55174334-55174334 on Build GRCh38
rs933162883 Chr.2: 55174131-55174131 on Build GRCh38
rs958657196 Chr.2: 55174041-55174041 on Build GRCh38
rs139866186 Chr.2: 55174232-55174232 on Build GRCh38
rs755020799 Chr.2: 55174081-55174082 on Build GRCh38
rs371406722 Chr.2: 55174097-55174098 on Build GRCh38
rs998920956 Chr.2: 55174386-55174386 on Build GRCh38
rs567927268 Chr.2: 55174448-55174448 on Build GRCh38
rs1035919755 Chr.2: 55174029-55174029 on Build GRCh38
rs771313170 Chr.2: 55174300-55174300 on Build GRCh38
rs575158067 Chr.2: 55174206-55174206 on Build GRCh38
rs1021603021 Chr.2: 55174375-55174375 on Build GRCh38
rs559143676 Chr.2: 55174403-55174403 on Build GRCh38
rs760521940 Chr.2: 55174345-55174346 on Build GRCh38
rs150677161 Chr.2: 55174222-55174222 on Build GRCh38
rs910474237 Chr.2: 55174152-55174152 on Build GRCh38
rs116054944 Chr.2: 55174068-55174068 on Build GRCh38
rs772033028 Chr.2: 55174271-55174271 on Build GRCh38
rs954530076 Chr.2: 55174419-55174419 on Build GRCh38
rs924497558 Chr.2: 55174122-55174122 on Build GRCh38
rs146463473 Chr.2: 55174013-55174013 on Build GRCh38
rs779077490 Chr.2: 55174013-55174014 on Build GRCh38
rs184534646 Chr.2: 55174267-55174267 on Build GRCh38
rs763095682 Chr.2: 55174272-55174272 on Build GRCh38
rs1012867081 Chr.2: 55174322-55174322 on Build GRCh38
rs572591982 Chr.2: 55174173-55174173 on Build GRCh38
rs973048349 Chr.2: 55174462-55174462 on Build GRCh38
rs769889666 Chr.2: 55174257-55174257 on Build GRCh38
rs555205886 Chr.2: 55174197-55174197 on Build GRCh38
rs544143643 Chr.2: 55174210-55174210 on Build GRCh38
rs866686730 Chr.2: 55174454-55174454 on Build GRCh38
rs977639146 Chr.2: 55174080-55174080 on Build GRCh38
rs969194290 Chr.2: 55174382-55174382 on Build GRCh38
rs541600095 Chr.2: 55174196-55174196 on Build GRCh38
rs974702850 Chr.2: 55174452-55174452 on Build GRCh38
rs539103838 Chr.2: 55174075-55174075 on Build GRCh38
rs201196741 Chr.2: 55174031-55174031 on Build GRCh38
rs6713058 Chr.2: 55174426-55174426 on Build GRCh38
rs894763438 Chr.2: 55174298-55174298 on Build GRCh38
rs933816513 Chr.2: 55174204-55174204 on Build GRCh38
rs1021655286 Chr.2: 55174052-55174052 on Build GRCh38
rs192608998 Chr.2: 55174159-55174159 on Build GRCh38
rs762118236 Chr.2: 55174014-55174014 on Build GRCh38
rs78364975 Chr.2: 55174014-55174014 on Build GRCh38
rs542161524 Chr.2: 55174410-55174410 on Build GRCh38
rs987363528 Chr.2: 55174139-55174139 on Build GRCh38
rs573150187 Chr.2: 55174276-55174276 on Build GRCh38
rs923348636 Chr.2: 55174202-55174202 on Build GRCh38

View Full Product Details