Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 55177498-55178011 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLHC1
Entrez Gene ID
130162
Gene Name
clathrin heavy chain linker domain containing 1
Gene Aliases
C2orf63
UniGene
Hs.347014
Gene Chromosome Location
Chr.2: 55172547-55232563 on Build GRCh38
Transcripts
NM_001135598.1 NM_152385.2 XM_005264136.2 XM_006711937.3 XM_006711938.3 XM_006711939.3 XM_011532519.2 XM_011532520.2 XM_011532521.2 XM_011532522.2 XM_011532523.2 XM_011532524.1 XM_011532525.2 XM_011532527.2 XM_011532528.1 XM_011532529.2 XM_011532530.1 XM_011532534.1 XM_011532535.2 XM_017003333.1 XM_017003334.1 XM_017003335.1 XM_017003336.1 XM_017003337.1 XM_017003338.1 XM_017003339.1 XM_017003340.1 XM_017003341.1 XM_017003342.1 XM_017003343.1 XM_017003344.1 XM_017003345.1 XM_017003346.1 XM_017003347.1 XM_017003349.1 XM_017003350.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6397603 c.1412G>T p.C471F Chr.2: 55177754-55177754 on Build GRCh38
6394688 c.1407G>T p.M469I Chr.2: 55177759-55177759 on Build GRCh38
3053018 c.1563A>G p.I521M Chr.2: 55177603-55177603 on Build GRCh38
6387835 c.1398G>T p.Q466H Chr.2: 55177768-55177768 on Build GRCh38
1198710 c.1493T>G p.L498R Chr.2: 55177673-55177673 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200853883 Chr.2: 55177958-55177959 on Build GRCh38
rs751030292 Chr.2: 55177796-55177796 on Build GRCh38
rs397753374 Chr.2: 55177960-55177961 on Build GRCh38
rs760712433 Chr.2: 55177827-55177827 on Build GRCh38
rs997999341 Chr.2: 55177808-55177808 on Build GRCh38
rs544550121 Chr.2: 55177976-55177976 on Build GRCh38
rs780985757 Chr.2: 55177722-55177722 on Build GRCh38
rs763541968 Chr.2: 55177794-55177794 on Build GRCh38
rs985549012 Chr.2: 55177679-55177679 on Build GRCh38
rs746120253 Chr.2: 55177622-55177622 on Build GRCh38
rs762632306 Chr.2: 55177790-55177790 on Build GRCh38
rs1049626600 Chr.2: 55177840-55177840 on Build GRCh38
rs746947773 Chr.2: 55177610-55177610 on Build GRCh38
rs749438373 Chr.2: 55177640-55177640 on Build GRCh38
rs949542774 Chr.2: 55177680-55177680 on Build GRCh38
rs755400307 Chr.2: 55177569-55177608 on Build GRCh38
rs772653130 Chr.2: 55177767-55177767 on Build GRCh38
rs768659419 Chr.2: 55177757-55177757 on Build GRCh38
rs572892152 Chr.2: 55177756-55177756 on Build GRCh38
rs200921597 Chr.2: 55177803-55177803 on Build GRCh38
rs945333940 Chr.2: 55177792-55177792 on Build GRCh38
rs982270387 Chr.2: 55177697-55177697 on Build GRCh38
rs773720803 Chr.2: 55177773-55177773 on Build GRCh38
rs763472634 Chr.2: 55177555-55177555 on Build GRCh38
rs568759143 Chr.2: 55177730-55177730 on Build GRCh38
rs752200561 Chr.2: 55177595-55177595 on Build GRCh38
rs760979549 Chr.2: 55177929-55177929 on Build GRCh38
rs889700577 Chr.2: 55177869-55177869 on Build GRCh38
rs749504729 Chr.2: 55177811-55177811 on Build GRCh38
rs533665450 Chr.2: 55177813-55177813 on Build GRCh38
rs764443223 Chr.2: 55177559-55177559 on Build GRCh38
rs896229122 Chr.2: 55177909-55177909 on Build GRCh38
rs755951708 Chr.2: 55177806-55177806 on Build GRCh38
rs777636223 Chr.2: 55177607-55177607 on Build GRCh38
rs1017044217 Chr.2: 55177618-55177618 on Build GRCh38
rs746077380 Chr.2: 55177881-55177881 on Build GRCh38
rs762142643 Chr.2: 55177569-55177569 on Build GRCh38
rs757899897 Chr.2: 55177685-55177685 on Build GRCh38
rs897291254 Chr.2: 55177925-55177925 on Build GRCh38
rs778590859 Chr.2: 55177591-55177591 on Build GRCh38
rs181837553 Chr.2: 55177908-55177908 on Build GRCh38
rs764912516 Chr.2: 55177684-55177684 on Build GRCh38
rs766832824 Chr.2: 55177778-55177778 on Build GRCh38
rs778874153 Chr.2: 55177815-55177815 on Build GRCh38
rs767806848 Chr.2: 55177670-55177670 on Build GRCh38
rs763633523 Chr.2: 55177689-55177689 on Build GRCh38
rs768769581 Chr.2: 55177651-55177651 on Build GRCh38
rs748196506 Chr.2: 55177819-55177819 on Build GRCh38
rs993825300 Chr.2: 55177949-55177949 on Build GRCh38
rs1023990929 Chr.2: 55177950-55177950 on Build GRCh38
rs896651545 Chr.2: 55177573-55177573 on Build GRCh38
rs761184708 Chr.2: 55177774-55177774 on Build GRCh38
rs542937675 Chr.2: 55177912-55177912 on Build GRCh38
rs750347831 Chr.2: 55177805-55177805 on Build GRCh38
rs770677081 Chr.2: 55177611-55177611 on Build GRCh38
rs1028229539 Chr.2: 55177571-55177571 on Build GRCh38
rs6545477 Chr.2: 55177789-55177789 on Build GRCh38
rs953999406 Chr.2: 55177677-55177677 on Build GRCh38
rs753588211 Chr.2: 55177580-55177580 on Build GRCh38
rs200555380 Chr.2: 55177725-55177725 on Build GRCh38
rs758358191 Chr.2: 55177596-55177596 on Build GRCh38
rs961306239 Chr.2: 55177938-55177938 on Build GRCh38
rs146577030 Chr.2: 55177931-55177931 on Build GRCh38
rs201825769 Chr.2: 55177629-55177629 on Build GRCh38
rs982237766 Chr.2: 55177981-55177981 on Build GRCh38
rs75442422 Chr.2: 55177901-55177901 on Build GRCh38
rs13032294 Chr.2: 55177747-55177747 on Build GRCh38
rs372061125 Chr.2: 55177783-55177783 on Build GRCh38
rs752103802 Chr.2: 55177567-55177567 on Build GRCh38
rs754641278 Chr.2: 55177586-55177586 on Build GRCh38
rs954121313 Chr.2: 55177593-55177593 on Build GRCh38
rs937641242 Chr.2: 55177866-55177866 on Build GRCh38
rs556536524 Chr.2: 55177930-55177930 on Build GRCh38
rs34877029 Chr.2: 55177952-55177953 on Build GRCh38
rs912610834 Chr.2: 55177693-55177693 on Build GRCh38
rs1019741914 Chr.2: 55177968-55177968 on Build GRCh38
rs779795629 Chr.2: 55177807-55177807 on Build GRCh38
rs767902628 Chr.2: 55177576-55177576 on Build GRCh38
rs566741388 Chr.2: 55177631-55177631 on Build GRCh38
rs781289849 Chr.2: 55177797-55177798 on Build GRCh38
rs141158674 Chr.2: 55177616-55177616 on Build GRCh38
rs774452457 Chr.2: 55177762-55177762 on Build GRCh38
rs748207070 Chr.2: 55177826-55177826 on Build GRCh38
rs757178355 Chr.2: 55177719-55177719 on Build GRCh38
rs14026 Chr.2: 55177658-55177658 on Build GRCh38
rs1001925626 Chr.2: 55177536-55177536 on Build GRCh38
rs201808625 Chr.2: 55177952-55177952 on Build GRCh38
rs762352104 Chr.2: 55177661-55177661 on Build GRCh38
rs200822172 Chr.2: 55177703-55177703 on Build GRCh38
rs748627006 Chr.2: 55177765-55177765 on Build GRCh38
rs1033357354 Chr.2: 55177549-55177549 on Build GRCh38
rs759278522 Chr.2: 55177671-55177671 on Build GRCh38

View Full Product Details