Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 106078023-106078529 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf40
Entrez Gene ID
84417
Gene Name
chromosome 2 open reading frame 40
Gene Aliases
ECRG4
UniGene
Hs.43125
Gene Chromosome Location
Chr.2: 106063290-106078155 on Build GRCh38
Transcripts
NM_032411.2 XM_006712799.3 XM_011511993.2 XM_017005106.1

SNP Detail

SNP ID Chromosome Location
rs925565310 Chr.2: 106078306-106078306 on Build GRCh38
rs532980118 Chr.2: 106078330-106078330 on Build GRCh38
rs907094668 Chr.2: 106078103-106078103 on Build GRCh38
rs971528644 Chr.2: 106078475-106078477 on Build GRCh38
rs1901462 Chr.2: 106078198-106078198 on Build GRCh38
rs984189218 Chr.2: 106078499-106078499 on Build GRCh38
rs1036942695 Chr.2: 106078420-106078420 on Build GRCh38
rs140346398 Chr.2: 106078484-106078484 on Build GRCh38
rs916629666 Chr.2: 106078329-106078329 on Build GRCh38
rs1028595646 Chr.2: 106078441-106078441 on Build GRCh38
rs886423111 Chr.2: 106078391-106078391 on Build GRCh38
rs765822965 Chr.2: 106078208-106078208 on Build GRCh38
rs1055213005 Chr.2: 106078050-106078050 on Build GRCh38
rs574392130 Chr.2: 106078225-106078225 on Build GRCh38
rs978294258 Chr.2: 106078305-106078305 on Build GRCh38
rs966345370 Chr.2: 106078505-106078505 on Build GRCh38
rs962574425 Chr.2: 106078162-106078162 on Build GRCh38
rs539433638 Chr.2: 106078051-106078051 on Build GRCh38
rs1013552601 Chr.2: 106078088-106078088 on Build GRCh38
rs750008210 Chr.2: 106078143-106078143 on Build GRCh38
rs889515808 Chr.2: 106078478-106078478 on Build GRCh38
rs544938713 Chr.2: 106078253-106078253 on Build GRCh38
rs563032627 Chr.2: 106078294-106078294 on Build GRCh38
rs966376374 Chr.2: 106078273-106078273 on Build GRCh38
rs1004121177 Chr.2: 106078137-106078137 on Build GRCh38
rs970448393 Chr.2: 106078200-106078200 on Build GRCh38
rs898416968 Chr.2: 106078422-106078422 on Build GRCh38
rs144345623 Chr.2: 106078201-106078201 on Build GRCh38
rs926326210 Chr.2: 106078487-106078487 on Build GRCh38
rs115903262 Chr.2: 106078347-106078347 on Build GRCh38
rs926290699 Chr.2: 106078202-106078202 on Build GRCh38
rs184659670 Chr.2: 106078111-106078111 on Build GRCh38
rs1019345325 Chr.2: 106078493-106078493 on Build GRCh38
rs954152989 Chr.2: 106078076-106078076 on Build GRCh38
rs1015545309 Chr.2: 106078154-106078154 on Build GRCh38
rs936844087 Chr.2: 106078323-106078323 on Build GRCh38
rs1007918250 Chr.2: 106078489-106078489 on Build GRCh38
rs76643480 Chr.2: 106078336-106078336 on Build GRCh38
rs545819264 Chr.2: 106078289-106078291 on Build GRCh38
rs573347779 Chr.2: 106078089-106078089 on Build GRCh38
rs146574620 Chr.2: 106078413-106078413 on Build GRCh38
rs1055707831 Chr.2: 106078327-106078327 on Build GRCh38
rs995783592 Chr.2: 106078193-106078193 on Build GRCh38
rs909780999 Chr.2: 106078264-106078265 on Build GRCh38
rs755791594 Chr.2: 106078157-106078157 on Build GRCh38
rs527343105 Chr.2: 106078440-106078440 on Build GRCh38
rs76980952 Chr.2: 106078087-106078087 on Build GRCh38
rs1046433893 Chr.2: 106078101-106078101 on Build GRCh38
rs1024467760 Chr.2: 106078470-106078470 on Build GRCh38
rs188531357 Chr.2: 106078475-106078475 on Build GRCh38
rs912241629 Chr.2: 106078226-106078226 on Build GRCh38
rs1000568952 Chr.2: 106078392-106078392 on Build GRCh38
rs200427067 Chr.2: 106078421-106078421 on Build GRCh38
rs540845056 Chr.2: 106078184-106078184 on Build GRCh38
rs1049775212 Chr.2: 106078386-106078386 on Build GRCh38
rs1028266067 Chr.2: 106078069-106078073 on Build GRCh38
rs1048310683 Chr.2: 106078261-106078261 on Build GRCh38

View Full Product Details