Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 108449545-108450009 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCC2
Entrez Gene ID
9648
Gene Name
GRIP and coiled-coil domain containing 2
Gene Aliases
GCC185, RANBP2L4, REN53
UniGene
Hs.436505
Gene Chromosome Location
Chr.2: 108448561-108509398 on Build GRCh38
Transcripts
NM_181453.3 NR_028063.1 XM_006712870.3 XM_006712871.1 XM_006712872.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6873878 c.13G>A p.V5I Chr.2: 108449639-108449639 on Build GRCh38
1398665 c.26T>C p.V9A Chr.2: 108449652-108449652 on Build GRCh38
5745937 c.62A>G p.K21R Chr.2: 108449688-108449688 on Build GRCh38
6249778 c.13G>T p.V5F Chr.2: 108449639-108449639 on Build GRCh38
7109188 c.52G>C p.G18R Chr.2: 108449678-108449678 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757986667 Chr.2: 108449666-108449666 on Build GRCh38
rs200193162 Chr.2: 108449734-108449734 on Build GRCh38
rs753789053 Chr.2: 108449638-108449638 on Build GRCh38
rs1054108833 Chr.2: 108449824-108449824 on Build GRCh38
rs188633754 Chr.2: 108449650-108449650 on Build GRCh38
rs763461312 Chr.2: 108449609-108449609 on Build GRCh38
rs923617013 Chr.2: 108449765-108449765 on Build GRCh38
rs1000638413 Chr.2: 108449933-108449933 on Build GRCh38
rs769929324 Chr.2: 108449874-108449875 on Build GRCh38
rs759400767 Chr.2: 108449714-108449714 on Build GRCh38
rs181041821 Chr.2: 108449706-108449706 on Build GRCh38
rs748992918 Chr.2: 108449587-108449587 on Build GRCh38
rs972060201 Chr.2: 108449960-108449960 on Build GRCh38
rs909314548 Chr.2: 108449670-108449670 on Build GRCh38
rs994837839 Chr.2: 108449603-108449603 on Build GRCh38
rs777363501 Chr.2: 108449671-108449671 on Build GRCh38
rs879122104 Chr.2: 108449779-108449780 on Build GRCh38
rs530811677 Chr.2: 108449767-108449767 on Build GRCh38
rs747781767 Chr.2: 108449595-108449595 on Build GRCh38
rs917477019 Chr.2: 108449778-108449778 on Build GRCh38
rs752439198 Chr.2: 108449716-108449716 on Build GRCh38
rs772529861 Chr.2: 108449597-108449597 on Build GRCh38
rs772148444 Chr.2: 108449705-108449705 on Build GRCh38
rs751212670 Chr.2: 108449726-108449726 on Build GRCh38
rs980287933 Chr.2: 108449963-108449963 on Build GRCh38
rs200794254 Chr.2: 108449764-108449765 on Build GRCh38
rs763102966 Chr.2: 108449778-108449778 on Build GRCh38
rs565658518 Chr.2: 108449766-108449766 on Build GRCh38
rs374147621 Chr.2: 108449708-108449708 on Build GRCh38
rs1030740316 Chr.2: 108449937-108449937 on Build GRCh38
rs746549960 Chr.2: 108449950-108449952 on Build GRCh38
rs930736283 Chr.2: 108449827-108449827 on Build GRCh38
rs139499066 Chr.2: 108449596-108449596 on Build GRCh38
rs2460946 Chr.2: 108449685-108449685 on Build GRCh38
rs138524973 Chr.2: 108449633-108449633 on Build GRCh38
rs146387944 Chr.2: 108449654-108449654 on Build GRCh38
rs2378113 Chr.2: 108449968-108449968 on Build GRCh38
rs181630876 Chr.2: 108449589-108449589 on Build GRCh38
rs1020101348 Chr.2: 108449568-108449568 on Build GRCh38
rs989100475 Chr.2: 108449770-108449770 on Build GRCh38
rs1028840996 Chr.2: 108449897-108449897 on Build GRCh38
rs575289171 Chr.2: 108449599-108449599 on Build GRCh38
rs770454650 Chr.2: 108449680-108449680 on Build GRCh38
rs780484058 Chr.2: 108449683-108449683 on Build GRCh38
rs200954666 Chr.2: 108449600-108449600 on Build GRCh38
rs777340644 Chr.2: 108449722-108449722 on Build GRCh38
rs370453746 Chr.2: 108449678-108449678 on Build GRCh38
rs1027613747 Chr.2: 108449614-108449614 on Build GRCh38
rs868629803 Chr.2: 108449831-108449831 on Build GRCh38
rs753835513 Chr.2: 108449712-108449712 on Build GRCh38
rs552194254 Chr.2: 108449791-108449791 on Build GRCh38
rs745810912 Chr.2: 108449599-108449600 on Build GRCh38
rs76672099 Chr.2: 108449855-108449855 on Build GRCh38
rs901748202 Chr.2: 108449882-108449882 on Build GRCh38
rs965809112 Chr.2: 108449869-108449869 on Build GRCh38
rs78477242 Chr.2: 108449810-108449810 on Build GRCh38
rs1028907587 Chr.2: 108449895-108449895 on Build GRCh38
rs975946085 Chr.2: 108449594-108449594 on Build GRCh38
rs972135043 Chr.2: 108449975-108449975 on Build GRCh38
rs377365761 Chr.2: 108449779-108449779 on Build GRCh38
rs2460944 Chr.2: 108449888-108449888 on Build GRCh38
rs765029505 Chr.2: 108449715-108449715 on Build GRCh38
rs184220438 Chr.2: 108449629-108449629 on Build GRCh38
rs796482720 Chr.2: 108449778-108449779 on Build GRCh38
rs766465172 Chr.2: 108449709-108449709 on Build GRCh38
rs1020552525 Chr.2: 108449860-108449860 on Build GRCh38
rs909187414 Chr.2: 108449967-108449967 on Build GRCh38
rs368775555 Chr.2: 108449618-108449618 on Build GRCh38
rs569012150 Chr.2: 108449866-108449866 on Build GRCh38
rs546897738 Chr.2: 108449817-108449817 on Build GRCh38
rs956428055 Chr.2: 108449944-108449944 on Build GRCh38
rs985227337 Chr.2: 108449956-108449956 on Build GRCh38
rs979143330 Chr.2: 108449764-108449764 on Build GRCh38
rs953279450 Chr.2: 108449918-108449918 on Build GRCh38
rs556775909 Chr.2: 108449861-108449861 on Build GRCh38
rs2460945 Chr.2: 108449832-108449832 on Build GRCh38
rs750423096 Chr.2: 108449634-108449634 on Build GRCh38
rs771875009 Chr.2: 108449685-108449686 on Build GRCh38
rs760715372 Chr.2: 108449707-108449707 on Build GRCh38
rs752351322 Chr.2: 108449662-108449662 on Build GRCh38
rs577571534 Chr.2: 108449636-108449636 on Build GRCh38
rs141863315 Chr.2: 108449875-108449876 on Build GRCh38
rs905950178 Chr.2: 108449602-108449602 on Build GRCh38
rs923564579 Chr.2: 108449578-108449578 on Build GRCh38
rs71732032 Chr.2: 108449876-108449877 on Build GRCh38
rs754734319 Chr.2: 108449588-108449588 on Build GRCh38
rs375154747 Chr.2: 108449697-108449697 on Build GRCh38
rs950282055 Chr.2: 108449785-108449785 on Build GRCh38
rs967846570 Chr.2: 108449927-108449927 on Build GRCh38
rs772586984 Chr.2: 108449970-108449970 on Build GRCh38
rs905436181 Chr.2: 108449806-108449806 on Build GRCh38
rs534352399 Chr.2: 108449796-108449796 on Build GRCh38
rs750268938 Chr.2: 108449778-108449779 on Build GRCh38
rs749808863 Chr.2: 108449684-108449684 on Build GRCh38
rs111776505 Chr.2: 108449798-108449798 on Build GRCh38
rs758098906 Chr.2: 108449717-108449717 on Build GRCh38

View Full Product Details