Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 108468987-108469493 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCC2
Entrez Gene ID
9648
Gene Name
GRIP and coiled-coil domain containing 2
Gene Aliases
GCC185, RANBP2L4, REN53
UniGene
Hs.436505
Gene Chromosome Location
Chr.2: 108448561-108509398 on Build GRCh38
Transcripts
NM_181453.3 NR_028063.1 XM_006712870.3 XM_006712871.1 XM_006712872.3 XM_011512213.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6669882 c.240_242delTCT p.L83delL Chr.2: 108469003-108469005 on Build GRCh38
6152833 c.311A>T p.Q104L Chr.2: 108469074-108469074 on Build GRCh38
5623203 c.247C>A p.L83M Chr.2: 108469010-108469010 on Build GRCh38
4657678 c.319G>A p.E107K Chr.2: 108469082-108469082 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs568418478 Chr.2: 108469249-108469249 on Build GRCh38
rs894344412 Chr.2: 108469040-108469040 on Build GRCh38
rs778787954 Chr.2: 108469018-108469018 on Build GRCh38
rs919910894 Chr.2: 108469071-108469071 on Build GRCh38
rs71980528 Chr.2: 108469199-108469202 on Build GRCh38
rs929582369 Chr.2: 108469369-108469369 on Build GRCh38
rs944181332 Chr.2: 108469123-108469123 on Build GRCh38
rs985868595 Chr.2: 108469437-108469437 on Build GRCh38
rs772082347 Chr.2: 108469053-108469053 on Build GRCh38
rs912603489 Chr.2: 108469453-108469453 on Build GRCh38
rs748107883 Chr.2: 108469045-108469045 on Build GRCh38
rs748569537 Chr.2: 108469213-108469213 on Build GRCh38
rs1033399117 Chr.2: 108469250-108469250 on Build GRCh38
rs1053236169 Chr.2: 108469469-108469469 on Build GRCh38
rs764672136 Chr.2: 108469198-108469199 on Build GRCh38
rs991958766 Chr.2: 108469302-108469302 on Build GRCh38
rs759044283 Chr.2: 108469112-108469112 on Build GRCh38
rs919518702 Chr.2: 108469367-108469367 on Build GRCh38
rs894208439 Chr.2: 108469459-108469459 on Build GRCh38
rs371427920 Chr.2: 108469119-108469119 on Build GRCh38
rs1021572147 Chr.2: 108469100-108469100 on Build GRCh38
rs746838759 Chr.2: 108469062-108469062 on Build GRCh38
rs903495476 Chr.2: 108469155-108469155 on Build GRCh38
rs1002289242 Chr.2: 108469206-108469206 on Build GRCh38
rs189553987 Chr.2: 108469428-108469428 on Build GRCh38
rs766652667 Chr.2: 108469136-108469136 on Build GRCh38
rs557045453 Chr.2: 108469395-108469395 on Build GRCh38
rs13424452 Chr.2: 108469131-108469131 on Build GRCh38
rs773465866 Chr.2: 108469375-108469375 on Build GRCh38
rs773689803 Chr.2: 108469129-108469129 on Build GRCh38
rs973938794 Chr.2: 108469407-108469407 on Build GRCh38
rs79693524 Chr.2: 108469202-108469205 on Build GRCh38
rs372582443 Chr.2: 108469420-108469420 on Build GRCh38
rs575622034 Chr.2: 108469415-108469415 on Build GRCh38
rs535957278 Chr.2: 108469259-108469259 on Build GRCh38
rs772547354 Chr.2: 108469324-108469324 on Build GRCh38
rs533096957 Chr.2: 108469104-108469106 on Build GRCh38
rs961219614 Chr.2: 108469334-108469334 on Build GRCh38
rs546790618 Chr.2: 108469149-108469149 on Build GRCh38
rs367799733 Chr.2: 108469326-108469326 on Build GRCh38
rs947208096 Chr.2: 108469463-108469463 on Build GRCh38
rs199880778 Chr.2: 108469202-108469202 on Build GRCh38
rs944112064 Chr.2: 108469460-108469460 on Build GRCh38
rs973502036 Chr.2: 108469288-108469288 on Build GRCh38
rs769360192 Chr.2: 108469114-108469114 on Build GRCh38
rs534605698 Chr.2: 108469127-108469127 on Build GRCh38
rs963530560 Chr.2: 108469279-108469279 on Build GRCh38
rs939935158 Chr.2: 108469010-108469010 on Build GRCh38
rs777741289 Chr.2: 108469058-108469058 on Build GRCh38
rs755149519 Chr.2: 108469016-108469016 on Build GRCh38
rs200556970 Chr.2: 108469106-108469106 on Build GRCh38
rs938350582 Chr.2: 108469418-108469418 on Build GRCh38
rs954713235 Chr.2: 108469159-108469159 on Build GRCh38
rs770609525 Chr.2: 108469101-108469101 on Build GRCh38
rs1055569214 Chr.2: 108469443-108469443 on Build GRCh38
rs200468428 Chr.2: 108469035-108469035 on Build GRCh38
rs932621103 Chr.2: 108469429-108469429 on Build GRCh38
rs768670565 Chr.2: 108469124-108469124 on Build GRCh38

View Full Product Details