Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 108894791-108895299 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDAR
Entrez Gene ID
10913
Gene Name
ectodysplasin A receptor
Gene Aliases
DL, ECTD10A, ECTD10B, ED1R, ED3, ED5, EDA-A1R, EDA1R, EDA3, HRM1
UniGene
Hs.171971
Gene Chromosome Location
Chr.2: 108894471-108989372 on Build GRCh38
Transcripts
NM_022336.3 XM_006712204.1 XM_011510502.2 XM_011510503.2 XM_017004623.1

Gene Symbol
RANBP2
Entrez Gene ID
5903
Gene Name
RAN binding protein 2
Gene Aliases
ADANE, ANE1, IIAE3, NUP358, TRP1, TRP2
UniGene
Hs.199561
Gene Chromosome Location
Chr.2: 108719446-109842301 on Build GRCh38
Transcripts
NM_022336.3 XM_006712204.1 XM_011510502.2 XM_011510503.2 XM_017004623.1

SNP Detail

SNP ID Chromosome Location
rs1042451601 Chr.2: 108894925-108894925 on Build GRCh38
rs540842456 Chr.2: 108895022-108895022 on Build GRCh38
rs954213103 Chr.2: 108894953-108894953 on Build GRCh38
rs1051307286 Chr.2: 108895263-108895263 on Build GRCh38
rs915148623 Chr.2: 108895225-108895225 on Build GRCh38
rs552720761 Chr.2: 108895041-108895041 on Build GRCh38
rs893891277 Chr.2: 108894899-108894899 on Build GRCh38
rs946814895 Chr.2: 108895146-108895146 on Build GRCh38
rs886054737 Chr.2: 108895146-108895146 on Build GRCh38
rs75934517 Chr.2: 108894991-108894991 on Build GRCh38
rs756408508 Chr.2: 108895109-108895109 on Build GRCh38
rs953263473 Chr.2: 108894895-108894895 on Build GRCh38
rs890040265 Chr.2: 108895273-108895273 on Build GRCh38
rs996998421 Chr.2: 108895145-108895146 on Build GRCh38
rs187935909 Chr.2: 108895123-108895123 on Build GRCh38
rs998466541 Chr.2: 108894938-108894938 on Build GRCh38
rs953118893 Chr.2: 108895148-108895148 on Build GRCh38
rs1018740359 Chr.2: 108895156-108895156 on Build GRCh38
rs973037587 Chr.2: 108894996-108894996 on Build GRCh38
rs200762071 Chr.2: 108895007-108895007 on Build GRCh38
rs1007471031 Chr.2: 108894960-108894960 on Build GRCh38
rs569939303 Chr.2: 108894867-108894867 on Build GRCh38
rs1028928428 Chr.2: 108895148-108895149 on Build GRCh38
rs1010899371 Chr.2: 108894923-108894923 on Build GRCh38
rs545744696 Chr.2: 108895210-108895210 on Build GRCh38
rs921691592 Chr.2: 108894892-108894893 on Build GRCh38
rs1042398708 Chr.2: 108895197-108895197 on Build GRCh38
rs946552537 Chr.2: 108894994-108894994 on Build GRCh38
rs1004052299 Chr.2: 108895028-108895028 on Build GRCh38
rs987828286 Chr.2: 108895200-108895200 on Build GRCh38
rs200168149 Chr.2: 108895250-108895251 on Build GRCh38
rs368551221 Chr.2: 108895100-108895100 on Build GRCh38
rs902748572 Chr.2: 108894926-108894926 on Build GRCh38
rs1017125023 Chr.2: 108895127-108895127 on Build GRCh38
rs35846821 Chr.2: 108894916-108894917 on Build GRCh38
rs558429529 Chr.2: 108894998-108894998 on Build GRCh38
rs750606176 Chr.2: 108895095-108895095 on Build GRCh38
rs915200979 Chr.2: 108894984-108894984 on Build GRCh38
rs574502076 Chr.2: 108895071-108895071 on Build GRCh38
rs866004644 Chr.2: 108895081-108895081 on Build GRCh38
rs1017319187 Chr.2: 108894970-108894970 on Build GRCh38
rs902486310 Chr.2: 108895209-108895209 on Build GRCh38
rs926448622 Chr.2: 108895262-108895262 on Build GRCh38
rs1055177501 Chr.2: 108894870-108894870 on Build GRCh38
rs987227738 Chr.2: 108895151-108895151 on Build GRCh38
rs981973048 Chr.2: 108895056-108895056 on Build GRCh38
rs145115545 Chr.2: 108895012-108895015 on Build GRCh38
rs189855301 Chr.2: 108895133-108895133 on Build GRCh38
rs963490576 Chr.2: 108894985-108894985 on Build GRCh38
rs565595816 Chr.2: 108895229-108895229 on Build GRCh38
rs115501541 Chr.2: 108894829-108894829 on Build GRCh38
rs946638269 Chr.2: 108895232-108895232 on Build GRCh38
rs150055772 Chr.2: 108894942-108894942 on Build GRCh38
rs541552562 Chr.2: 108895076-108895076 on Build GRCh38
rs147877575 Chr.2: 108895069-108895070 on Build GRCh38
rs182138755 Chr.2: 108895239-108895239 on Build GRCh38

View Full Product Details