Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 108910698-108911190 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDAR
Entrez Gene ID
10913
Gene Name
ectodysplasin A receptor
Gene Aliases
DL, ECTD10A, ECTD10B, ED1R, ED3, ED5, EDA-A1R, EDA1R, EDA3, HRM1
UniGene
Hs.171971
Gene Chromosome Location
Chr.2: 108894471-108989372 on Build GRCh38
Transcripts
NM_022336.3 XM_006712204.1 XM_011510502.2 XM_011510503.2 XM_017004623.1

Gene Symbol
RANBP2
Entrez Gene ID
5903
Gene Name
RAN binding protein 2
Gene Aliases
ADANE, ANE1, IIAE3, NUP358, TRP1, TRP2
UniGene
Hs.199561
Gene Chromosome Location
Chr.2: 108719446-109842301 on Build GRCh38
Transcripts
NM_022336.3 XM_006712204.1 XM_011510502.2 XM_011510503.2 XM_017004623.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3565150 c.662G>A p.G221D Chr.2: 108910940-108910940 on Build GRCh38
3990510 c.771G>A p.P257P Chr.2: 108910831-108910831 on Build GRCh38
5157345 c.653delC p.P218fs*48 Chr.2: 108910949-108910949 on Build GRCh38
6891645 c.638C>T p.T213I Chr.2: 108910964-108910964 on Build GRCh38
4930669 c.671A>T p.H224L Chr.2: 108910835-108910835 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376646267 Chr.2: 108910888-108910888 on Build GRCh38
rs751689993 Chr.2: 108910798-108910798 on Build GRCh38
rs762163525 Chr.2: 108911038-108911038 on Build GRCh38
rs527248830 Chr.2: 108910808-108910808 on Build GRCh38
rs760041106 Chr.2: 108910854-108910854 on Build GRCh38
rs377483763 Chr.2: 108910931-108910931 on Build GRCh38
rs749715266 Chr.2: 108911024-108911024 on Build GRCh38
rs765598659 Chr.2: 108910779-108910779 on Build GRCh38
rs770292889 Chr.2: 108911001-108911001 on Build GRCh38
rs948939371 Chr.2: 108910751-108910751 on Build GRCh38
rs773222347 Chr.2: 108911078-108911078 on Build GRCh38
rs368706624 Chr.2: 108910853-108910853 on Build GRCh38
rs957850287 Chr.2: 108911134-108911134 on Build GRCh38
rs747761003 Chr.2: 108910898-108910898 on Build GRCh38
rs200521015 Chr.2: 108910818-108910818 on Build GRCh38
rs562628749 Chr.2: 108910747-108910747 on Build GRCh38
rs994080945 Chr.2: 108910725-108910725 on Build GRCh38
rs917390912 Chr.2: 108910750-108910750 on Build GRCh38
rs781294991 Chr.2: 108910805-108910805 on Build GRCh38
rs779752511 Chr.2: 108910896-108910896 on Build GRCh38
rs260630 Chr.2: 108910917-108910917 on Build GRCh38
rs967379483 Chr.2: 108911059-108911059 on Build GRCh38
rs1015722033 Chr.2: 108911130-108911130 on Build GRCh38
rs200208516 Chr.2: 108910941-108910941 on Build GRCh38
rs766326456 Chr.2: 108910956-108910956 on Build GRCh38
rs952675426 Chr.2: 108910717-108910717 on Build GRCh38
rs997108648 Chr.2: 108910886-108910886 on Build GRCh38
rs201950451 Chr.2: 108910996-108910996 on Build GRCh38
rs767662584 Chr.2: 108910890-108910890 on Build GRCh38
rs771512216 Chr.2: 108910844-108910844 on Build GRCh38
rs191975348 Chr.2: 108910942-108910942 on Build GRCh38
rs908558583 Chr.2: 108910736-108910736 on Build GRCh38
rs372067031 Chr.2: 108910813-108910813 on Build GRCh38
rs765958662 Chr.2: 108911144-108911144 on Build GRCh38
rs148157322 Chr.2: 108911132-108911132 on Build GRCh38
rs185922326 Chr.2: 108910927-108910927 on Build GRCh38
rs769079016 Chr.2: 108910928-108910928 on Build GRCh38
rs756970592 Chr.2: 108911099-108911100 on Build GRCh38
rs774830746 Chr.2: 108911028-108911028 on Build GRCh38
rs779845734 Chr.2: 108910821-108910821 on Build GRCh38
rs746044070 Chr.2: 108910831-108910831 on Build GRCh38
rs373163180 Chr.2: 108910930-108910930 on Build GRCh38
rs755113054 Chr.2: 108910792-108910794 on Build GRCh38
rs765570387 Chr.2: 108910859-108910859 on Build GRCh38
rs904980286 Chr.2: 108910773-108910773 on Build GRCh38
rs533098592 Chr.2: 108911167-108911167 on Build GRCh38
rs768323596 Chr.2: 108910822-108910822 on Build GRCh38
rs759899621 Chr.2: 108910764-108910764 on Build GRCh38
rs369135198 Chr.2: 108910893-108910893 on Build GRCh38
rs746403053 Chr.2: 108910912-108910912 on Build GRCh38
rs562536513 Chr.2: 108911141-108911141 on Build GRCh38
rs114808659 Chr.2: 108910887-108910887 on Build GRCh38
rs143058412 Chr.2: 108911021-108911021 on Build GRCh38
rs996791353 Chr.2: 108911007-108911007 on Build GRCh38
rs753658252 Chr.2: 108910975-108910975 on Build GRCh38
rs748926409 Chr.2: 108910740-108910740 on Build GRCh38
rs757324481 Chr.2: 108910799-108910799 on Build GRCh38
rs368489348 Chr.2: 108911093-108911093 on Build GRCh38
rs181034518 Chr.2: 108910904-108910904 on Build GRCh38
rs906240797 Chr.2: 108910719-108910719 on Build GRCh38
rs760595921 Chr.2: 108910946-108910946 on Build GRCh38
rs540596441 Chr.2: 108910990-108910990 on Build GRCh38
rs901116756 Chr.2: 108911002-108911002 on Build GRCh38
rs936383353 Chr.2: 108910959-108910959 on Build GRCh38
rs371233610 Chr.2: 108910992-108910992 on Build GRCh38
rs145268306 Chr.2: 108910780-108910780 on Build GRCh38
rs797044436 Chr.2: 108910784-108910787 on Build GRCh38
rs749292747 Chr.2: 108910891-108910892 on Build GRCh38
rs763155082 Chr.2: 108910875-108910875 on Build GRCh38
rs777037287 Chr.2: 108910762-108910762 on Build GRCh38
rs1049566207 Chr.2: 108911064-108911064 on Build GRCh38
rs188529241 Chr.2: 108910752-108910752 on Build GRCh38
rs760820677 Chr.2: 108911084-108911084 on Build GRCh38
rs3749108 Chr.2: 108910783-108910783 on Build GRCh38
rs775893324 Chr.2: 108910866-108910866 on Build GRCh38
rs1014698148 Chr.2: 108911161-108911161 on Build GRCh38
rs148212997 Chr.2: 108910995-108910995 on Build GRCh38
rs368251062 Chr.2: 108910797-108910797 on Build GRCh38
rs372462035 Chr.2: 108911120-108911120 on Build GRCh38
rs372183192 Chr.2: 108910903-108910903 on Build GRCh38
rs764367022 Chr.2: 108910884-108910884 on Build GRCh38
rs34862638 Chr.2: 108910910-108910911 on Build GRCh38
rs139371163 Chr.2: 108910905-108910905 on Build GRCh38
rs150286846 Chr.2: 108910999-108910999 on Build GRCh38
rs781419713 Chr.2: 108911055-108911055 on Build GRCh38
rs769162793 Chr.2: 108911026-108911026 on Build GRCh38
rs573163925 Chr.2: 108910983-108910983 on Build GRCh38
rs200137108 Chr.2: 108910829-108910829 on Build GRCh38
rs759449643 Chr.2: 108911102-108911102 on Build GRCh38
rs754642923 Chr.2: 108910897-108910897 on Build GRCh38
rs777519649 Chr.2: 108910888-108910900 on Build GRCh38
rs772418418 Chr.2: 108911048-108911048 on Build GRCh38
rs540285589 Chr.2: 108910894-108910894 on Build GRCh38
rs775781155 Chr.2: 108910921-108910921 on Build GRCh38
rs544391061 Chr.2: 108910723-108910723 on Build GRCh38
rs758652158 Chr.2: 108910843-108910843 on Build GRCh38
rs376622302 Chr.2: 108910832-108910832 on Build GRCh38
rs993490630 Chr.2: 108910743-108910743 on Build GRCh38
rs768070647 Chr.2: 108910741-108910741 on Build GRCh38
rs865973121 Chr.2: 108911031-108911031 on Build GRCh38
rs766864982 Chr.2: 108910760-108910760 on Build GRCh38

View Full Product Details