Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 110639636-110640134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BUB1
Entrez Gene ID
699
Gene Name
BUB1 mitotic checkpoint serine/threonine kinase
Gene Aliases
BUB1A, BUB1L, hBUB1
UniGene
Hs.469649
Gene Chromosome Location
Chr.2: 110637698-110678107 on Build GRCh38
Transcripts
NM_001278616.1 NM_001278617.1 NM_004336.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1398761 c.2959G>A p.D987N Chr.2: 110639845-110639845 on Build GRCh38
4666420 c.2958C>T p.I986I Chr.2: 110639846-110639846 on Build GRCh38
4582568 c.2982A>G p.T994T Chr.2: 110639822-110639822 on Build GRCh38
4991577 c.3043C>T p.P1015S Chr.2: 110639761-110639761 on Build GRCh38
4991576 c.3044C>T p.P1015L Chr.2: 110639760-110639760 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1044261853 Chr.2: 110639965-110639965 on Build GRCh38
rs751736912 Chr.2: 110639933-110639933 on Build GRCh38
rs904584894 Chr.2: 110639994-110639994 on Build GRCh38
rs768338575 Chr.2: 110639901-110639901 on Build GRCh38
rs148368189 Chr.2: 110639846-110639846 on Build GRCh38
rs149445802 Chr.2: 110639822-110639822 on Build GRCh38
rs755314258 Chr.2: 110639861-110639861 on Build GRCh38
rs761826243 Chr.2: 110639911-110639911 on Build GRCh38
rs765310857 Chr.2: 110639912-110639912 on Build GRCh38
rs775026607 Chr.2: 110639882-110639882 on Build GRCh38
rs1009005268 Chr.2: 110640029-110640029 on Build GRCh38
rs972495810 Chr.2: 110639669-110639669 on Build GRCh38
rs907916980 Chr.2: 110639880-110639880 on Build GRCh38
rs1031418099 Chr.2: 110639988-110639988 on Build GRCh38
rs748650973 Chr.2: 110639863-110639863 on Build GRCh38
rs55952031 Chr.2: 110639768-110639768 on Build GRCh38
rs112515450 Chr.2: 110639661-110639661 on Build GRCh38
rs773918857 Chr.2: 110639751-110639751 on Build GRCh38
rs891542211 Chr.2: 110640012-110640012 on Build GRCh38
rs750585350 Chr.2: 110639819-110639819 on Build GRCh38
rs1010767925 Chr.2: 110639936-110639936 on Build GRCh38
rs757336873 Chr.2: 110639799-110639799 on Build GRCh38
rs765400716 Chr.2: 110639801-110639801 on Build GRCh38
rs776631812 Chr.2: 110639905-110639905 on Build GRCh38
rs377544977 Chr.2: 110639887-110639887 on Build GRCh38
rs760418033 Chr.2: 110639789-110639789 on Build GRCh38
rs942921022 Chr.2: 110639827-110639827 on Build GRCh38
rs904369242 Chr.2: 110639974-110639974 on Build GRCh38
rs777029694 Chr.2: 110639714-110639714 on Build GRCh38
rs764042909 Chr.2: 110639791-110639791 on Build GRCh38
rs771141605 Chr.2: 110639920-110639923 on Build GRCh38
rs770143203 Chr.2: 110639776-110639778 on Build GRCh38
rs747377863 Chr.2: 110639691-110639691 on Build GRCh38
rs769170073 Chr.2: 110639700-110639700 on Build GRCh38
rs369276113 Chr.2: 110639961-110639961 on Build GRCh38
rs750495460 Chr.2: 110639915-110639915 on Build GRCh38
rs990622281 Chr.2: 110639660-110639661 on Build GRCh38
rs770345159 Chr.2: 110639865-110639865 on Build GRCh38
rs753838787 Chr.2: 110639795-110639795 on Build GRCh38
rs528634792 Chr.2: 110639924-110639924 on Build GRCh38
rs139042805 Chr.2: 110639720-110639720 on Build GRCh38
rs548390657 Chr.2: 110639989-110639989 on Build GRCh38
rs762968559 Chr.2: 110639923-110639923 on Build GRCh38
rs191275100 Chr.2: 110639873-110639873 on Build GRCh38
rs775324806 Chr.2: 110639773-110639773 on Build GRCh38
rs74693163 Chr.2: 110640057-110640057 on Build GRCh38
rs747353009 Chr.2: 110639851-110639851 on Build GRCh38
rs999953024 Chr.2: 110639975-110639975 on Build GRCh38
rs753109866 Chr.2: 110639951-110639951 on Build GRCh38
rs186775527 Chr.2: 110639870-110639870 on Build GRCh38
rs755296735 Chr.2: 110639942-110639942 on Build GRCh38
rs766567725 Chr.2: 110639929-110639929 on Build GRCh38
rs369159867 Chr.2: 110639769-110639769 on Build GRCh38
rs150708782 Chr.2: 110639662-110639662 on Build GRCh38
rs773628416 Chr.2: 110639815-110639816 on Build GRCh38
rs948625888 Chr.2: 110639964-110639964 on Build GRCh38
rs763228139 Chr.2: 110639856-110639856 on Build GRCh38
rs763092002 Chr.2: 110640062-110640062 on Build GRCh38
rs781524924 Chr.2: 110639945-110639945 on Build GRCh38

View Full Product Details