Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 112181550-112182059 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBLN7
Entrez Gene ID
129804
Gene Name
fibulin 7
Gene Aliases
TM14
UniGene
Hs.437696
Gene Chromosome Location
Chr.2: 112138385-112254888 on Build GRCh38
Transcripts
NM_001128165.1 NM_153214.2 XM_006712260.3 XM_011510585.2 XM_011510587.2 XM_017003317.1 XM_017003318.1 XM_017003319.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6499709 c.579G>A p.A193A Chr.2: 112181785-112181785 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs4848280 Chr.2: 112181626-112181626 on Build GRCh38
rs756399788 Chr.2: 112181796-112181796 on Build GRCh38
rs374275003 Chr.2: 112181810-112181810 on Build GRCh38
rs895081310 Chr.2: 112182024-112182024 on Build GRCh38
rs994753465 Chr.2: 112181612-112181612 on Build GRCh38
rs544719522 Chr.2: 112181667-112181667 on Build GRCh38
rs757590942 Chr.2: 112181945-112181945 on Build GRCh38
rs780750281 Chr.2: 112181858-112181858 on Build GRCh38
rs943451263 Chr.2: 112181746-112181746 on Build GRCh38
rs934922250 Chr.2: 112181689-112181689 on Build GRCh38
rs899018643 Chr.2: 112181595-112181595 on Build GRCh38
rs920640347 Chr.2: 112181947-112181947 on Build GRCh38
rs775640753 Chr.2: 112181909-112181909 on Build GRCh38
rs550343439 Chr.2: 112181833-112181833 on Build GRCh38
rs547539097 Chr.2: 112181770-112181770 on Build GRCh38
rs1039034838 Chr.2: 112181942-112181942 on Build GRCh38
rs182683993 Chr.2: 112181999-112181999 on Build GRCh38
rs754142667 Chr.2: 112181932-112181932 on Build GRCh38
rs887025417 Chr.2: 112181884-112181884 on Build GRCh38
rs767418109 Chr.2: 112181905-112181905 on Build GRCh38
rs564885174 Chr.2: 112181680-112181680 on Build GRCh38
rs1039559535 Chr.2: 112181762-112181762 on Build GRCh38
rs1026211541 Chr.2: 112181622-112181622 on Build GRCh38
rs770730720 Chr.2: 112181900-112181900 on Build GRCh38
rs755656525 Chr.2: 112181972-112181972 on Build GRCh38
rs978972242 Chr.2: 112181676-112181676 on Build GRCh38
rs764362413 Chr.2: 112181925-112181925 on Build GRCh38
rs915385767 Chr.2: 112181988-112181989 on Build GRCh38
rs1047601901 Chr.2: 112181784-112181784 on Build GRCh38
rs867890573 Chr.2: 112181893-112181893 on Build GRCh38
rs916068278 Chr.2: 112181665-112181665 on Build GRCh38
rs548509268 Chr.2: 112181903-112181903 on Build GRCh38
rs539326562 Chr.2: 112181902-112181902 on Build GRCh38
rs772876968 Chr.2: 112181888-112181888 on Build GRCh38
rs886346448 Chr.2: 112181791-112181791 on Build GRCh38
rs74804124 Chr.2: 112181632-112181632 on Build GRCh38
rs749045398 Chr.2: 112181892-112181892 on Build GRCh38
rs756955236 Chr.2: 112181878-112181878 on Build GRCh38
rs1045237192 Chr.2: 112182019-112182019 on Build GRCh38
rs959107674 Chr.2: 112181644-112181644 on Build GRCh38
rs754197494 Chr.2: 112181827-112181827 on Build GRCh38
rs920904411 Chr.2: 112181737-112181737 on Build GRCh38
rs924817249 Chr.2: 112181891-112181891 on Build GRCh38
rs371149020 Chr.2: 112181701-112181701 on Build GRCh38
rs924311496 Chr.2: 112181953-112181953 on Build GRCh38
rs747604486 Chr.2: 112181787-112181787 on Build GRCh38
rs995038482 Chr.2: 112181771-112181771 on Build GRCh38
rs777307357 Chr.2: 112182012-112182012 on Build GRCh38
rs749971374 Chr.2: 112181967-112181967 on Build GRCh38
rs1034666493 Chr.2: 112181809-112181809 on Build GRCh38
rs911935095 Chr.2: 112181731-112181731 on Build GRCh38
rs769383900 Chr.2: 112181886-112181886 on Build GRCh38
rs141297870 Chr.2: 112182034-112182034 on Build GRCh38
rs4849050 Chr.2: 112181971-112181971 on Build GRCh38
rs530268620 Chr.2: 112181829-112181829 on Build GRCh38
rs780439125 Chr.2: 112181963-112181963 on Build GRCh38
rs200955793 Chr.2: 112181834-112181834 on Build GRCh38
rs962412763 Chr.2: 112181679-112181679 on Build GRCh38
rs760850849 Chr.2: 112181914-112181914 on Build GRCh38
rs750821856 Chr.2: 112181951-112181951 on Build GRCh38
rs1052615164 Chr.2: 112181807-112181807 on Build GRCh38
rs904482402 Chr.2: 112181699-112181699 on Build GRCh38
rs558777055 Chr.2: 112181949-112181949 on Build GRCh38
rs987708648 Chr.2: 112181904-112181904 on Build GRCh38
rs1004537286 Chr.2: 112181885-112181885 on Build GRCh38
rs969261446 Chr.2: 112181673-112181673 on Build GRCh38
rs779246427 Chr.2: 112181832-112181832 on Build GRCh38
rs990999717 Chr.2: 112181648-112181648 on Build GRCh38
rs1003499380 Chr.2: 112181797-112181797 on Build GRCh38
rs570292101 Chr.2: 112181861-112181861 on Build GRCh38
rs915413813 Chr.2: 112181808-112181808 on Build GRCh38
rs905214446 Chr.2: 112181588-112181588 on Build GRCh38
rs766973417 Chr.2: 112181883-112181883 on Build GRCh38
rs1034715585 Chr.2: 112181641-112181641 on Build GRCh38
rs758874302 Chr.2: 112181956-112181956 on Build GRCh38

View Full Product Details