Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 112182701-112183201 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBLN7
Entrez Gene ID
129804
Gene Name
fibulin 7
Gene Aliases
TM14
UniGene
Hs.437696
Gene Chromosome Location
Chr.2: 112138385-112254888 on Build GRCh38
Transcripts
NM_001128165.1 NM_153214.2 XM_006712260.3 XM_011510585.2 XM_011510587.2 XM_017003317.1 XM_017003318.1 XM_017003319.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4084206 c.684T>C p.C228C Chr.2: 112182804-112182804 on Build GRCh38
4084205 c.672C>T p.D224D Chr.2: 112182792-112182792 on Build GRCh38
6661603 c.744G>A p.P248P Chr.2: 112182864-112182864 on Build GRCh38
1398827 c.792C>T p.D264D Chr.2: 112182912-112182912 on Build GRCh38
5544988 c.709C>T p.P237S Chr.2: 112182829-112182829 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777328529 Chr.2: 112182948-112182950 on Build GRCh38
rs199894123 Chr.2: 112182875-112182875 on Build GRCh38
rs762092531 Chr.2: 112182801-112182801 on Build GRCh38
rs766836093 Chr.2: 112182820-112182820 on Build GRCh38
rs1005179197 Chr.2: 112183085-112183085 on Build GRCh38
rs540886073 Chr.2: 112182999-112182999 on Build GRCh38
rs925155895 Chr.2: 112183170-112183170 on Build GRCh38
rs1053320839 Chr.2: 112183151-112183151 on Build GRCh38
rs767206123 Chr.2: 112182889-112182889 on Build GRCh38
rs770544265 Chr.2: 112182744-112182744 on Build GRCh38
rs780033468 Chr.2: 112182846-112182846 on Build GRCh38
rs534725543 Chr.2: 112182813-112182813 on Build GRCh38
rs779992593 Chr.2: 112182912-112182912 on Build GRCh38
rs1015804868 Chr.2: 112182958-112182958 on Build GRCh38
rs771987669 Chr.2: 112182781-112182781 on Build GRCh38
rs75864194 Chr.2: 112183082-112183082 on Build GRCh38
rs572352470 Chr.2: 112182983-112182983 on Build GRCh38
rs545728449 Chr.2: 112182770-112182773 on Build GRCh38
rs538804942 Chr.2: 112182885-112182885 on Build GRCh38
rs771793225 Chr.2: 112182843-112182843 on Build GRCh38
rs769733891 Chr.2: 112182954-112182954 on Build GRCh38
rs76320146 Chr.2: 112183152-112183152 on Build GRCh38
rs773393060 Chr.2: 112182971-112182971 on Build GRCh38
rs377292553 Chr.2: 112182798-112182798 on Build GRCh38
rs760028458 Chr.2: 112182881-112182881 on Build GRCh38
rs147423265 Chr.2: 112182792-112182792 on Build GRCh38
rs912460727 Chr.2: 112183100-112183100 on Build GRCh38
rs201347772 Chr.2: 112182852-112182852 on Build GRCh38
rs373897520 Chr.2: 112182863-112182863 on Build GRCh38
rs745627243 Chr.2: 112182834-112182834 on Build GRCh38
rs1049702180 Chr.2: 112183040-112183040 on Build GRCh38
rs925164250 Chr.2: 112182920-112182920 on Build GRCh38
rs765880707 Chr.2: 112183146-112183146 on Build GRCh38
rs750741970 Chr.2: 112182814-112182814 on Build GRCh38
rs763280507 Chr.2: 112182874-112182874 on Build GRCh38
rs781301058 Chr.2: 112182938-112182938 on Build GRCh38
rs765986862 Chr.2: 112183156-112183159 on Build GRCh38
rs1023698911 Chr.2: 112183147-112183147 on Build GRCh38
rs568501949 Chr.2: 112182833-112182833 on Build GRCh38
rs565930395 Chr.2: 112183051-112183051 on Build GRCh38
rs774188892 Chr.2: 112182760-112182760 on Build GRCh38
rs760134150 Chr.2: 112182838-112182838 on Build GRCh38
rs935281403 Chr.2: 112182967-112182967 on Build GRCh38
rs996318513 Chr.2: 112183022-112183022 on Build GRCh38
rs899598810 Chr.2: 112183013-112183013 on Build GRCh38
rs372879160 Chr.2: 112182789-112182789 on Build GRCh38
rs139679712 Chr.2: 112182876-112182876 on Build GRCh38
rs765383477 Chr.2: 112182812-112182812 on Build GRCh38
rs535390766 Chr.2: 112182950-112182950 on Build GRCh38
rs912250152 Chr.2: 112182992-112182992 on Build GRCh38
rs992363846 Chr.2: 112182759-112182759 on Build GRCh38
rs753408985 Chr.2: 112182831-112182831 on Build GRCh38
rs778588026 Chr.2: 112182898-112182898 on Build GRCh38
rs569662326 Chr.2: 112182899-112182899 on Build GRCh38
rs375997636 Chr.2: 112182961-112182961 on Build GRCh38
rs756863178 Chr.2: 112182832-112182832 on Build GRCh38
rs369109624 Chr.2: 112182778-112182778 on Build GRCh38
rs1011281216 Chr.2: 112182860-112182860 on Build GRCh38
rs3811643 Chr.2: 112183001-112183001 on Build GRCh38
rs1014915603 Chr.2: 112183095-112183095 on Build GRCh38
rs960726462 Chr.2: 112183109-112183109 on Build GRCh38
rs557506367 Chr.2: 112183153-112183153 on Build GRCh38
rs758171350 Chr.2: 112182908-112182908 on Build GRCh38
rs954046533 Chr.2: 112183072-112183072 on Build GRCh38
rs773341440 Chr.2: 112182873-112182873 on Build GRCh38
rs147173660 Chr.2: 112182913-112182913 on Build GRCh38
rs768657853 Chr.2: 112182933-112182933 on Build GRCh38
rs769255084 Chr.2: 112182942-112182943 on Build GRCh38
rs753327780 Chr.2: 112182888-112182888 on Build GRCh38
rs776911504 Chr.2: 112182799-112182799 on Build GRCh38
rs557130843 Chr.2: 112182853-112182853 on Build GRCh38
rs79906489 Chr.2: 112182825-112182825 on Build GRCh38
rs183971080 Chr.2: 112182729-112182729 on Build GRCh38
rs777249747 Chr.2: 112182827-112182827 on Build GRCh38
rs890866636 Chr.2: 112182730-112182730 on Build GRCh38
rs369700908 Chr.2: 112182847-112182847 on Build GRCh38
rs763254491 Chr.2: 112182818-112182818 on Build GRCh38

View Full Product Details