Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 112187028-112187520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBLN7
Entrez Gene ID
129804
Gene Name
fibulin 7
Gene Aliases
TM14
UniGene
Hs.437696
Gene Chromosome Location
Chr.2: 112138385-112254888 on Build GRCh38
Transcripts
NM_001128165.1 NM_153214.2 XM_006712260.3 XM_011510585.2 XM_011510587.2 XM_017003317.1 XM_017003318.1 XM_017003319.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1398830 c.1171C>T p.R391W Chr.2: 112187357-112187357 on Build GRCh38
4683771 c.1024_1025delCT p.L344fs*94 Chr.2: 112187210-112187211 on Build GRCh38
6316162 c.976G>T p.D326Y Chr.2: 112187162-112187162 on Build GRCh38
6204940 c.958C>T p.R320W Chr.2: 112187144-112187144 on Build GRCh38
7101420 c.1030C>G p.L344V Chr.2: 112187216-112187216 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770492147 Chr.2: 112187195-112187195 on Build GRCh38
rs752373171 Chr.2: 112187132-112187132 on Build GRCh38
rs370949329 Chr.2: 112187427-112187427 on Build GRCh38
rs140178022 Chr.2: 112187431-112187431 on Build GRCh38
rs768238890 Chr.2: 112187381-112187381 on Build GRCh38
rs143978448 Chr.2: 112187335-112187335 on Build GRCh38
rs1038928820 Chr.2: 112187066-112187066 on Build GRCh38
rs765162406 Chr.2: 112187242-112187242 on Build GRCh38
rs536232013 Chr.2: 112187205-112187205 on Build GRCh38
rs572200014 Chr.2: 112187234-112187234 on Build GRCh38
rs148652111 Chr.2: 112187312-112187312 on Build GRCh38
rs756450842 Chr.2: 112187422-112187422 on Build GRCh38
rs746489625 Chr.2: 112187441-112187441 on Build GRCh38
rs145108622 Chr.2: 112187178-112187178 on Build GRCh38
rs911965347 Chr.2: 112187262-112187262 on Build GRCh38
rs199709086 Chr.2: 112187300-112187300 on Build GRCh38
rs781690102 Chr.2: 112187269-112187269 on Build GRCh38
rs770547057 Chr.2: 112187111-112187111 on Build GRCh38
rs753743458 Chr.2: 112187144-112187144 on Build GRCh38
rs886822407 Chr.2: 112187077-112187077 on Build GRCh38
rs766235578 Chr.2: 112187406-112187406 on Build GRCh38
rs780732262 Chr.2: 112187445-112187445 on Build GRCh38
rs762450106 Chr.2: 112187092-112187092 on Build GRCh38
rs755216343 Chr.2: 112187253-112187253 on Build GRCh38
rs964817003 Chr.2: 112187470-112187470 on Build GRCh38
rs781615983 Chr.2: 112187495-112187495 on Build GRCh38
rs761732994 Chr.2: 112187237-112187237 on Build GRCh38
rs367632309 Chr.2: 112187496-112187496 on Build GRCh38
rs769506660 Chr.2: 112187460-112187460 on Build GRCh38
rs760281370 Chr.2: 112187235-112187235 on Build GRCh38
rs186694313 Chr.2: 112187127-112187127 on Build GRCh38
rs749054176 Chr.2: 112187465-112187465 on Build GRCh38
rs763176195 Chr.2: 112187269-112187270 on Build GRCh38
rs773935441 Chr.2: 112187212-112187212 on Build GRCh38
rs747787624 Chr.2: 112187448-112187448 on Build GRCh38
rs550163758 Chr.2: 112187068-112187068 on Build GRCh38
rs371987292 Chr.2: 112187190-112187190 on Build GRCh38
rs772939951 Chr.2: 112187398-112187398 on Build GRCh38
rs150736251 Chr.2: 112187328-112187328 on Build GRCh38
rs140094535 Chr.2: 112187464-112187464 on Build GRCh38
rs200379992 Chr.2: 112187150-112187150 on Build GRCh38
rs201355169 Chr.2: 112187329-112187329 on Build GRCh38
rs913866668 Chr.2: 112187180-112187181 on Build GRCh38
rs200992956 Chr.2: 112187330-112187330 on Build GRCh38
rs759128288 Chr.2: 112187218-112187218 on Build GRCh38
rs766532212 Chr.2: 112187270-112187271 on Build GRCh38
rs199980872 Chr.2: 112187358-112187358 on Build GRCh38
rs774433929 Chr.2: 112187407-112187407 on Build GRCh38
rs773788291 Chr.2: 112187228-112187228 on Build GRCh38
rs536952219 Chr.2: 112187067-112187067 on Build GRCh38
rs778949008 Chr.2: 112187148-112187148 on Build GRCh38
rs140504449 Chr.2: 112187236-112187236 on Build GRCh38
rs774067722 Chr.2: 112187126-112187126 on Build GRCh38
rs748531043 Chr.2: 112187272-112187272 on Build GRCh38
rs527375702 Chr.2: 112187419-112187419 on Build GRCh38
rs746659537 Chr.2: 112187290-112187290 on Build GRCh38
rs143675954 Chr.2: 112187276-112187276 on Build GRCh38
rs865870547 Chr.2: 112187114-112187114 on Build GRCh38
rs991636546 Chr.2: 112187188-112187188 on Build GRCh38
rs766472285 Chr.2: 112187327-112187327 on Build GRCh38
rs767207144 Chr.2: 112187128-112187128 on Build GRCh38
rs746503301 Chr.2: 112187380-112187380 on Build GRCh38
rs762847963 Chr.2: 112187320-112187320 on Build GRCh38
rs756575507 Chr.2: 112187348-112187348 on Build GRCh38
rs747840798 Chr.2: 112187391-112187391 on Build GRCh38
rs369753216 Chr.2: 112187279-112187279 on Build GRCh38
rs1026006745 Chr.2: 112187113-112187113 on Build GRCh38
rs750455955 Chr.2: 112187244-112187244 on Build GRCh38
rs763894916 Chr.2: 112187139-112187139 on Build GRCh38
rs368513842 Chr.2: 112187423-112187423 on Build GRCh38
rs565012617 Chr.2: 112187386-112187386 on Build GRCh38
rs757234528 Chr.2: 112187145-112187145 on Build GRCh38
rs777226796 Chr.2: 112187086-112187086 on Build GRCh38
rs762947528 Chr.2: 112187399-112187399 on Build GRCh38
rs760537934 Chr.2: 112187138-112187138 on Build GRCh38
rs759714093 Chr.2: 112187417-112187417 on Build GRCh38
rs768361747 Chr.2: 112187292-112187292 on Build GRCh38
rs755086860 Chr.2: 112187340-112187341 on Build GRCh38
rs201195702 Chr.2: 112187177-112187177 on Build GRCh38
rs147018646 Chr.2: 112187275-112187275 on Build GRCh38
rs558661682 Chr.2: 112187185-112187185 on Build GRCh38
rs868863342 Chr.2: 112187389-112187389 on Build GRCh38
rs778318785 Chr.2: 112187349-112187349 on Build GRCh38
rs779612135 Chr.2: 112187377-112187377 on Build GRCh38
rs200640724 Chr.2: 112187280-112187280 on Build GRCh38
rs370318617 Chr.2: 112187311-112187311 on Build GRCh38
rs377093141 Chr.2: 112187245-112187245 on Build GRCh38
rs1023113582 Chr.2: 112187384-112187384 on Build GRCh38
rs145709063 Chr.2: 112187160-112187161 on Build GRCh38
rs758493175 Chr.2: 112187154-112187154 on Build GRCh38
rs202243216 Chr.2: 112187426-112187426 on Build GRCh38
rs751785078 Chr.2: 112187252-112187252 on Build GRCh38
rs751613701 Chr.2: 112187339-112187340 on Build GRCh38
rs769554941 Chr.2: 112187396-112187396 on Build GRCh38
rs374169244 Chr.2: 112187357-112187357 on Build GRCh38
rs201976044 Chr.2: 112187420-112187420 on Build GRCh38
rs115924283 Chr.2: 112187053-112187053 on Build GRCh38
rs1003977294 Chr.2: 112187149-112187149 on Build GRCh38
rs781725463 Chr.2: 112187187-112187187 on Build GRCh38
rs757716376 Chr.2: 112187430-112187430 on Build GRCh38
rs754453864 Chr.2: 112187444-112187444 on Build GRCh38
rs530197977 Chr.2: 112187463-112187463 on Build GRCh38
rs776892875 Chr.2: 112187301-112187301 on Build GRCh38
rs138160056 Chr.2: 112187238-112187238 on Build GRCh38

View Full Product Details