Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 112231497-112232044 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBLN7
Entrez Gene ID
129804
Gene Name
fibulin 7
Gene Aliases
TM14
UniGene
Hs.437696
Gene Chromosome Location
Chr.2: 112138385-112254888 on Build GRCh38
Transcripts
XM_017003317.1 NM_032494.2 XM_011511998.1 XM_017005107.1

Gene Symbol
ZC3H8
Entrez Gene ID
84524
Gene Name
zinc finger CCCH-type containing 8
Gene Aliases
Fliz1, ZC3HDC8
UniGene
Hs.418416
Gene Chromosome Location
Chr.2: 112215035-112255087 on Build GRCh38
Transcripts
XM_017003317.1 NM_032494.2 XM_011511998.1 XM_017005107.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4922442 c.748A>C p.K250Q Chr.2: 112231933-112231933 on Build GRCh38
7448809 c.839C>A p.A280D Chr.2: 112231842-112231842 on Build GRCh38
1005498 c.798T>G p.F266L Chr.2: 112231883-112231883 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs748316181 Chr.2: 112231879-112231879 on Build GRCh38
rs770061992 Chr.2: 112231884-112231884 on Build GRCh38
rs778735347 Chr.2: 112231811-112231811 on Build GRCh38
rs778565181 Chr.2: 112231819-112231819 on Build GRCh38
rs778072351 Chr.2: 112231673-112231673 on Build GRCh38
rs552275446 Chr.2: 112231709-112231709 on Build GRCh38
rs376119163 Chr.2: 112231804-112231804 on Build GRCh38
rs781135871 Chr.2: 112231952-112231952 on Build GRCh38
rs1015287934 Chr.2: 112231631-112231631 on Build GRCh38
rs773511770 Chr.2: 112231895-112231895 on Build GRCh38
rs768598046 Chr.2: 112231864-112231864 on Build GRCh38
rs1028169560 Chr.2: 112231524-112231524 on Build GRCh38
rs543025884 Chr.2: 112231719-112231719 on Build GRCh38
rs901280612 Chr.2: 112231982-112231982 on Build GRCh38
rs760134954 Chr.2: 112231955-112231955 on Build GRCh38
rs917010363 Chr.2: 112231671-112231671 on Build GRCh38
rs763494832 Chr.2: 112231976-112231976 on Build GRCh38
rs576887974 Chr.2: 112231809-112231809 on Build GRCh38
rs746507987 Chr.2: 112231831-112231834 on Build GRCh38
rs867354910 Chr.2: 112231890-112231890 on Build GRCh38
rs981844954 Chr.2: 112231979-112231979 on Build GRCh38
rs780162705 Chr.2: 112231843-112231843 on Build GRCh38
rs745660635 Chr.2: 112231821-112231821 on Build GRCh38
rs1054186506 Chr.2: 112231767-112231767 on Build GRCh38
rs957510369 Chr.2: 112231760-112231760 on Build GRCh38
rs114195994 Chr.2: 112231711-112231711 on Build GRCh38
rs992550305 Chr.2: 112231657-112231657 on Build GRCh38
rs572190745 Chr.2: 112231628-112231628 on Build GRCh38
rs563261710 Chr.2: 112231807-112231807 on Build GRCh38
rs774812862 Chr.2: 112231929-112231929 on Build GRCh38
rs997150139 Chr.2: 112231636-112231636 on Build GRCh38
rs762341678 Chr.2: 112231984-112231985 on Build GRCh38
rs766836197 Chr.2: 112231912-112231912 on Build GRCh38
rs867588649 Chr.2: 112232006-112232006 on Build GRCh38
rs945375235 Chr.2: 112231823-112231823 on Build GRCh38
rs776830085 Chr.2: 112231868-112231868 on Build GRCh38
rs779614609 Chr.2: 112231804-112231805 on Build GRCh38
rs996612691 Chr.2: 112232011-112232011 on Build GRCh38
rs777271418 Chr.2: 112231854-112231855 on Build GRCh38
rs980953431 Chr.2: 112231695-112231695 on Build GRCh38
rs984175817 Chr.2: 112231613-112231613 on Build GRCh38
rs753452640 Chr.2: 112231987-112231987 on Build GRCh38
rs936966588 Chr.2: 112231755-112231755 on Build GRCh38
rs768341644 Chr.2: 112231838-112231838 on Build GRCh38
rs1007410786 Chr.2: 112231538-112231538 on Build GRCh38
rs763387471 Chr.2: 112231789-112231789 on Build GRCh38
rs534641054 Chr.2: 112231644-112231644 on Build GRCh38
rs554439786 Chr.2: 112231700-112231700 on Build GRCh38
rs773646250 Chr.2: 112231754-112231757 on Build GRCh38
rs915960186 Chr.2: 112231945-112231945 on Build GRCh38
rs756989772 Chr.2: 112231814-112231814 on Build GRCh38
rs1017341259 Chr.2: 112231602-112231602 on Build GRCh38
rs763332483 Chr.2: 112231907-112231907 on Build GRCh38
rs747256273 Chr.2: 112231677-112231677 on Build GRCh38
rs754918705 Chr.2: 112231740-112231741 on Build GRCh38
rs758351113 Chr.2: 112231833-112231833 on Build GRCh38
rs747045212 Chr.2: 112231860-112231860 on Build GRCh38
rs887644255 Chr.2: 112231535-112231535 on Build GRCh38
rs952679652 Chr.2: 112231601-112231601 on Build GRCh38
rs968015927 Chr.2: 112231691-112231691 on Build GRCh38
rs749776661 Chr.2: 112231961-112231961 on Build GRCh38

View Full Product Details