Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 112756286-112756795 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CKAP2L
Entrez Gene ID
150468
Gene Name
cytoskeleton associated protein 2 like
Gene Aliases
-
UniGene
Hs.434250
Gene Chromosome Location
Chr.2: 112736349-112764677 on Build GRCh38
Transcripts
NM_001304361.1 NM_152515.4 NR_130712.1 XM_011510666.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3039882 c.642T>C p.Y214Y Chr.2: 112756729-112756729 on Build GRCh38
6629527 c.1075C>T p.Q359* Chr.2: 112756296-112756296 on Build GRCh38
3039880 c.973G>A p.E325K Chr.2: 112756398-112756398 on Build GRCh38
6153667 c.586G>C p.E196Q Chr.2: 112756785-112756785 on Build GRCh38
6153668 c.624G>T p.K208N Chr.2: 112756747-112756747 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs186817501 Chr.2: 112756515-112756515 on Build GRCh38
rs966834130 Chr.2: 112756650-112756650 on Build GRCh38
rs768009129 Chr.2: 112756618-112756618 on Build GRCh38
rs764680963 Chr.2: 112756767-112756767 on Build GRCh38
rs757551001 Chr.2: 112756411-112756411 on Build GRCh38
rs34015720 Chr.2: 112756415-112756415 on Build GRCh38
rs776622742 Chr.2: 112756667-112756667 on Build GRCh38
rs771957695 Chr.2: 112756540-112756540 on Build GRCh38
rs753907689 Chr.2: 112756467-112756467 on Build GRCh38
rs756244469 Chr.2: 112756388-112756388 on Build GRCh38
rs772047220 Chr.2: 112756455-112756455 on Build GRCh38
rs772584190 Chr.2: 112756340-112756340 on Build GRCh38
rs750628668 Chr.2: 112756481-112756481 on Build GRCh38
rs763203294 Chr.2: 112756604-112756604 on Build GRCh38
rs531044518 Chr.2: 112756330-112756330 on Build GRCh38
rs570758592 Chr.2: 112756391-112756391 on Build GRCh38
rs779233001 Chr.2: 112756688-112756688 on Build GRCh38
rs369686797 Chr.2: 112756770-112756770 on Build GRCh38
rs773878907 Chr.2: 112756451-112756451 on Build GRCh38
rs762736339 Chr.2: 112756357-112756357 on Build GRCh38
rs200765044 Chr.2: 112756416-112756416 on Build GRCh38
rs373739143 Chr.2: 112756575-112756575 on Build GRCh38
rs766057775 Chr.2: 112756359-112756359 on Build GRCh38
rs17042341 Chr.2: 112756582-112756582 on Build GRCh38
rs778322279 Chr.2: 112756626-112756626 on Build GRCh38
rs746341672 Chr.2: 112756339-112756339 on Build GRCh38
rs766519512 Chr.2: 112756713-112756713 on Build GRCh38
rs747594129 Chr.2: 112756346-112756346 on Build GRCh38
rs375043973 Chr.2: 112756366-112756366 on Build GRCh38
rs371196442 Chr.2: 112756550-112756550 on Build GRCh38
rs781603729 Chr.2: 112756317-112756318 on Build GRCh38
rs1049855572 Chr.2: 112756738-112756738 on Build GRCh38
rs954478174 Chr.2: 112756458-112756458 on Build GRCh38
rs1025758784 Chr.2: 112756328-112756328 on Build GRCh38
rs189588016 Chr.2: 112756406-112756406 on Build GRCh38
rs778614679 Chr.2: 112756535-112756535 on Build GRCh38
rs765436681 Chr.2: 112756473-112756473 on Build GRCh38
rs773216009 Chr.2: 112756704-112756704 on Build GRCh38
rs753213373 Chr.2: 112756736-112756736 on Build GRCh38
rs764989782 Chr.2: 112756347-112756347 on Build GRCh38
rs17042342 Chr.2: 112756603-112756603 on Build GRCh38
rs745569587 Chr.2: 112756536-112756536 on Build GRCh38
rs769308437 Chr.2: 112756350-112756350 on Build GRCh38
rs756266631 Chr.2: 112756319-112756319 on Build GRCh38
rs143618159 Chr.2: 112756463-112756463 on Build GRCh38
rs747573709 Chr.2: 112756429-112756429 on Build GRCh38
rs777307131 Chr.2: 112756439-112756439 on Build GRCh38
rs766709636 Chr.2: 112756609-112756609 on Build GRCh38
rs138942105 Chr.2: 112756548-112756548 on Build GRCh38
rs748101494 Chr.2: 112756668-112756668 on Build GRCh38
rs376487793 Chr.2: 112756739-112756739 on Build GRCh38
rs746736829 Chr.2: 112756639-112756639 on Build GRCh38
rs759317813 Chr.2: 112756454-112756454 on Build GRCh38
rs752799817 Chr.2: 112756729-112756730 on Build GRCh38
rs1005653117 Chr.2: 112756753-112756753 on Build GRCh38
rs769888267 Chr.2: 112756692-112756692 on Build GRCh38
rs753973906 Chr.2: 112756399-112756399 on Build GRCh38
rs551009951 Chr.2: 112756331-112756331 on Build GRCh38
rs765291620 Chr.2: 112756594-112756594 on Build GRCh38
rs776441081 Chr.2: 112756551-112756551 on Build GRCh38
rs758072975 Chr.2: 112756635-112756635 on Build GRCh38
rs146911893 Chr.2: 112756539-112756539 on Build GRCh38
rs760732259 Chr.2: 112756457-112756457 on Build GRCh38
rs545818608 Chr.2: 112756615-112756615 on Build GRCh38
rs779886328 Chr.2: 112756638-112756638 on Build GRCh38
rs769290334 Chr.2: 112756431-112756431 on Build GRCh38
rs370718483 Chr.2: 112756341-112756341 on Build GRCh38
rs374752968 Chr.2: 112756524-112756524 on Build GRCh38
rs775560983 Chr.2: 112756456-112756456 on Build GRCh38
rs750775927 Chr.2: 112756759-112756759 on Build GRCh38
rs761833875 Chr.2: 112756471-112756471 on Build GRCh38
rs267598836 Chr.2: 112756599-112756599 on Build GRCh38
rs536752121 Chr.2: 112756521-112756521 on Build GRCh38
rs759880871 Chr.2: 112756729-112756729 on Build GRCh38
rs371786477 Chr.2: 112756708-112756708 on Build GRCh38
rs745490560 Chr.2: 112756634-112756634 on Build GRCh38
rs34358218 Chr.2: 112756449-112756449 on Build GRCh38
rs751012449 Chr.2: 112756474-112756474 on Build GRCh38
rs756783256 Chr.2: 112756620-112756620 on Build GRCh38
rs199885814 Chr.2: 112756488-112756488 on Build GRCh38
rs1001299911 Chr.2: 112756593-112756593 on Build GRCh38
rs371752157 Chr.2: 112756724-112756724 on Build GRCh38
rs199612253 Chr.2: 112756520-112756520 on Build GRCh38
rs772838057 Chr.2: 112756355-112756355 on Build GRCh38
rs926888334 Chr.2: 112756342-112756342 on Build GRCh38
rs554363912 Chr.2: 112756499-112756499 on Build GRCh38
rs964664665 Chr.2: 112756402-112756402 on Build GRCh38
rs727502804 Chr.2: 112756620-112756620 on Build GRCh38
rs367945388 Chr.2: 112756661-112756661 on Build GRCh38
rs756625382 Chr.2: 112756761-112756761 on Build GRCh38
rs758748773 Chr.2: 112756486-112756486 on Build GRCh38
rs774146065 Chr.2: 112756364-112756364 on Build GRCh38
rs775471274 Chr.2: 112756543-112756543 on Build GRCh38
rs377476756 Chr.2: 112756338-112756338 on Build GRCh38
rs748769677 Chr.2: 112756441-112756441 on Build GRCh38
rs778079829 Chr.2: 112756329-112756329 on Build GRCh38
rs1041097435 Chr.2: 112756435-112756435 on Build GRCh38
rs767835982 Chr.2: 112756734-112756734 on Build GRCh38

View Full Product Details