Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 114442193-114442739 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPP10
Entrez Gene ID
57628
Gene Name
dipeptidyl peptidase like 10
Gene Aliases
DPL2, DPPY, DPRP-3, DPRP3
UniGene
Hs.580539
Gene Chromosome Location
Chr.2: 114442358-115845752 on Build GRCh38
Transcripts
NM_001321907.1 NM_001321910.1 NM_001321913.1 NM_020868.4

SNP Detail

SNP ID Chromosome Location
rs1023027485 Chr.2: 114442365-114442365 on Build GRCh38
rs546308754 Chr.2: 114442258-114442258 on Build GRCh38
rs771813624 Chr.2: 114442403-114442403 on Build GRCh38
rs988919753 Chr.2: 114442422-114442422 on Build GRCh38
rs966322127 Chr.2: 114442670-114442670 on Build GRCh38
rs544578444 Chr.2: 114442630-114442630 on Build GRCh38
rs900708324 Chr.2: 114442587-114442587 on Build GRCh38
rs1010416495 Chr.2: 114442647-114442647 on Build GRCh38
rs151095638 Chr.2: 114442669-114442669 on Build GRCh38
rs907956851 Chr.2: 114442560-114442560 on Build GRCh38
rs772997455 Chr.2: 114442286-114442287 on Build GRCh38
rs566848720 Chr.2: 114442596-114442596 on Build GRCh38
rs1048199146 Chr.2: 114442563-114442563 on Build GRCh38
rs966425061 Chr.2: 114442373-114442373 on Build GRCh38
rs909258065 Chr.2: 114442568-114442568 on Build GRCh38
rs891532271 Chr.2: 114442645-114442645 on Build GRCh38
rs942125489 Chr.2: 114442582-114442582 on Build GRCh38
rs984908316 Chr.2: 114442553-114442553 on Build GRCh38
rs143161868 Chr.2: 114442284-114442285 on Build GRCh38
rs746850694 Chr.2: 114442577-114442577 on Build GRCh38
rs958030968 Chr.2: 114442387-114442387 on Build GRCh38
rs929355986 Chr.2: 114442561-114442561 on Build GRCh38
rs907991142 Chr.2: 114442708-114442708 on Build GRCh38
rs1029239823 Chr.2: 114442691-114442691 on Build GRCh38
rs970621845 Chr.2: 114442539-114442539 on Build GRCh38
rs553520446 Chr.2: 114442576-114442576 on Build GRCh38
rs982050485 Chr.2: 114442542-114442542 on Build GRCh38
rs952049203 Chr.2: 114442692-114442692 on Build GRCh38
rs189904100 Chr.2: 114442617-114442617 on Build GRCh38
rs1037260856 Chr.2: 114442583-114442583 on Build GRCh38
rs776705828 Chr.2: 114442707-114442707 on Build GRCh38
rs1048260491 Chr.2: 114442277-114442277 on Build GRCh38
rs575851383 Chr.2: 114442618-114442618 on Build GRCh38
rs982019279 Chr.2: 114442693-114442693 on Build GRCh38
rs564346413 Chr.2: 114442225-114442225 on Build GRCh38
rs939860330 Chr.2: 114442573-114442573 on Build GRCh38
rs1025524207 Chr.2: 114442697-114442697 on Build GRCh38
rs549741674 Chr.2: 114442251-114442251 on Build GRCh38
rs532814683 Chr.2: 114442242-114442242 on Build GRCh38
rs748641469 Chr.2: 114442713-114442713 on Build GRCh38
rs974788940 Chr.2: 114442474-114442474 on Build GRCh38
rs1039180564 Chr.2: 114442292-114442292 on Build GRCh38
rs1004042511 Chr.2: 114442284-114442284 on Build GRCh38
rs930722907 Chr.2: 114442504-114442504 on Build GRCh38
rs895748189 Chr.2: 114442585-114442585 on Build GRCh38
rs144719983 Chr.2: 114442324-114442324 on Build GRCh38
rs921889127 Chr.2: 114442500-114442500 on Build GRCh38
rs1013952249 Chr.2: 114442347-114442347 on Build GRCh38
rs905633275 Chr.2: 114442366-114442366 on Build GRCh38
rs1019222158 Chr.2: 114442427-114442427 on Build GRCh38
rs888247830 Chr.2: 114442282-114442282 on Build GRCh38
rs928625942 Chr.2: 114442552-114442552 on Build GRCh38
rs551562446 Chr.2: 114442477-114442477 on Build GRCh38
rs999866279 Chr.2: 114442369-114442369 on Build GRCh38
rs577830041 Chr.2: 114442705-114442705 on Build GRCh38
rs143048199 Chr.2: 114442322-114442322 on Build GRCh38
rs186372952 Chr.2: 114442508-114442508 on Build GRCh38
rs142904148 Chr.2: 114442603-114442603 on Build GRCh38
rs984786237 Chr.2: 114442700-114442700 on Build GRCh38
rs910570472 Chr.2: 114442627-114442641 on Build GRCh38
rs78271629 Chr.2: 114442377-114442377 on Build GRCh38
rs202063163 Chr.2: 114442249-114442250 on Build GRCh38
rs1032058695 Chr.2: 114442380-114442380 on Build GRCh38
rs997736712 Chr.2: 114442323-114442323 on Build GRCh38
rs916430433 Chr.2: 114442535-114442535 on Build GRCh38
rs975366344 Chr.2: 114442681-114442681 on Build GRCh38
rs764907741 Chr.2: 114442284-114442287 on Build GRCh38
rs571471098 Chr.2: 114442501-114442501 on Build GRCh38
rs747017269 Chr.2: 114442643-114442643 on Build GRCh38
rs148542349 Chr.2: 114442450-114442450 on Build GRCh38
rs957782510 Chr.2: 114442640-114442640 on Build GRCh38
rs537638282 Chr.2: 114442467-114442467 on Build GRCh38

View Full Product Details