Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 115499430-115499930 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPP10
Entrez Gene ID
57628
Gene Name
dipeptidyl peptidase like 10
Gene Aliases
DPL2, DPPY, DPRP-3, DPRP3
UniGene
Hs.580539
Gene Chromosome Location
Chr.2: 114442358-115845752 on Build GRCh38
Transcripts
NM_001004360.3 NM_001178034.1 NM_001178036.1 NM_001178037.1 NM_001321905.1 NM_001321906.1 NM_001321907.1 NM_001321908.1 NM_001321909.1 NM_001321910.1 NM_001321911.1 NM_001321912.1 NM_001321913.1 NM_001321914.1 NM_020868.4 XM_017004565.1 XM_017004566.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
280590 c.342T>G p.T114T Chr.2: 115499601-115499601 on Build GRCh38
3565571 c.297G>A p.L99L Chr.2: 115499556-115499556 on Build GRCh38
5895568 c.314G>A p.G105E Chr.2: 115499540-115499540 on Build GRCh38
5826136 c.320T>A p.V107D Chr.2: 115499546-115499546 on Build GRCh38
6534560 c.342A>G p.T114T Chr.2: 115499580-115499580 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761060820 Chr.2: 115499559-115499559 on Build GRCh38
rs965167003 Chr.2: 115499841-115499841 on Build GRCh38
rs746345806 Chr.2: 115499538-115499538 on Build GRCh38
rs141809253 Chr.2: 115499533-115499533 on Build GRCh38
rs887205166 Chr.2: 115499780-115499780 on Build GRCh38
rs191798266 Chr.2: 115499886-115499886 on Build GRCh38
rs1013617412 Chr.2: 115499845-115499845 on Build GRCh38
rs931412223 Chr.2: 115499769-115499769 on Build GRCh38
rs773176791 Chr.2: 115499566-115499566 on Build GRCh38
rs1046885880 Chr.2: 115499681-115499681 on Build GRCh38
rs745704058 Chr.2: 115499498-115499500 on Build GRCh38
rs543593030 Chr.2: 115499894-115499894 on Build GRCh38
rs764930663 Chr.2: 115499550-115499550 on Build GRCh38
rs761151832 Chr.2: 115499903-115499903 on Build GRCh38
rs973768035 Chr.2: 115499905-115499905 on Build GRCh38
rs756259621 Chr.2: 115499515-115499515 on Build GRCh38
rs369811818 Chr.2: 115499703-115499703 on Build GRCh38
rs757675936 Chr.2: 115499634-115499634 on Build GRCh38
rs557623590 Chr.2: 115499464-115499464 on Build GRCh38
rs1031397953 Chr.2: 115499870-115499870 on Build GRCh38
rs1047050765 Chr.2: 115499510-115499510 on Build GRCh38
rs772718178 Chr.2: 115499695-115499695 on Build GRCh38
rs746424543 Chr.2: 115499655-115499655 on Build GRCh38
rs758196072 Chr.2: 115499648-115499648 on Build GRCh38
rs756358446 Chr.2: 115499597-115499597 on Build GRCh38
rs374186916 Chr.2: 115499577-115499577 on Build GRCh38
rs144249280 Chr.2: 115499530-115499530 on Build GRCh38
rs202131701 Chr.2: 115499541-115499541 on Build GRCh38
rs77285207 Chr.2: 115499806-115499806 on Build GRCh38
rs1047615759 Chr.2: 115499773-115499773 on Build GRCh38
rs1021790095 Chr.2: 115499853-115499853 on Build GRCh38
rs943286536 Chr.2: 115499733-115499733 on Build GRCh38
rs762772381 Chr.2: 115499570-115499570 on Build GRCh38
rs146013809 Chr.2: 115499624-115499624 on Build GRCh38
rs1057062642 Chr.2: 115499804-115499804 on Build GRCh38
rs767675553 Chr.2: 115499587-115499587 on Build GRCh38
rs920458556 Chr.2: 115499755-115499755 on Build GRCh38
rs895317746 Chr.2: 115499668-115499668 on Build GRCh38
rs62154959 Chr.2: 115499861-115499861 on Build GRCh38
rs964650709 Chr.2: 115499893-115499893 on Build GRCh38
rs139947044 Chr.2: 115499581-115499581 on Build GRCh38
rs761571881 Chr.2: 115499545-115499545 on Build GRCh38
rs911046656 Chr.2: 115499501-115499501 on Build GRCh38
rs750203671 Chr.2: 115499532-115499532 on Build GRCh38
rs978461469 Chr.2: 115499599-115499599 on Build GRCh38
rs146805294 Chr.2: 115499721-115499721 on Build GRCh38
rs141887141 Chr.2: 115499799-115499799 on Build GRCh38
rs780411777 Chr.2: 115499670-115499670 on Build GRCh38
rs949559181 Chr.2: 115499678-115499678 on Build GRCh38
rs369443192 Chr.2: 115499498-115499498 on Build GRCh38
rs965239376 Chr.2: 115499516-115499516 on Build GRCh38
rs755282001 Chr.2: 115499722-115499722 on Build GRCh38
rs766383071 Chr.2: 115499474-115499474 on Build GRCh38
rs954391560 Chr.2: 115499706-115499706 on Build GRCh38
rs990993697 Chr.2: 115499904-115499904 on Build GRCh38
rs977611512 Chr.2: 115499676-115499676 on Build GRCh38
rs1013046831 Chr.2: 115499605-115499605 on Build GRCh38
rs143608967 Chr.2: 115499665-115499665 on Build GRCh38
rs751774063 Chr.2: 115499691-115499691 on Build GRCh38
rs375478365 Chr.2: 115499865-115499865 on Build GRCh38
rs559609621 Chr.2: 115499738-115499738 on Build GRCh38
rs998601616 Chr.2: 115499868-115499868 on Build GRCh38
rs751696392 Chr.2: 115499480-115499480 on Build GRCh38
rs931397690 Chr.2: 115499458-115499458 on Build GRCh38
rs1008531018 Chr.2: 115499885-115499885 on Build GRCh38
rs1055330679 Chr.2: 115499656-115499656 on Build GRCh38
rs754869602 Chr.2: 115499586-115499586 on Build GRCh38
rs755031056 Chr.2: 115499482-115499482 on Build GRCh38
rs781119878 Chr.2: 115499484-115499484 on Build GRCh38
rs778160232 Chr.2: 115499521-115499521 on Build GRCh38
rs1026969236 Chr.2: 115499727-115499727 on Build GRCh38
rs373910244 Chr.2: 115499596-115499596 on Build GRCh38
rs954465176 Chr.2: 115499877-115499877 on Build GRCh38
rs537382794 Chr.2: 115499455-115499455 on Build GRCh38
rs144222031 Chr.2: 115499800-115499800 on Build GRCh38
rs1020424876 Chr.2: 115499856-115499856 on Build GRCh38
rs777904268 Chr.2: 115499604-115499604 on Build GRCh38
rs925512727 Chr.2: 115499614-115499614 on Build GRCh38

View Full Product Details