Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 121339049-121339563 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLASP1
Entrez Gene ID
23332
Gene Name
cytoplasmic linker associated protein 1
Gene Aliases
MAST1
UniGene
Hs.469840
Gene Chromosome Location
Chr.2: 121337776-121649476 on Build GRCh38
Transcripts
NM_001142273.1 NM_001142274.1 NM_001207051.1 NM_015282.2 XM_006712377.1 XM_006712381.1 XM_006712382.1 XM_006712383.1 XM_006712385.1 XM_011510848.1 XM_011510849.1 XM_017003657.1 XM_017003665.1 XM_017003667.1 XM_017003669.1 XM_017003670.1 XM_017003672.1 XM_017003673.1 XM_017003674.1 XM_017003675.1 XM_017003676.1 XM_017003677.1 XM_017003678.1 XM_017003679.1 XM_017003681.1 XM_017003682.1 XM_017003683.1 XM_017003684.1 XM_017003685.1 XM_017003686.1 XM_017003687.1 XM_017003688.1 XM_017003689.1

SNP Detail

SNP ID Chromosome Location
rs1032353890 Chr.2: 121339106-121339106 on Build GRCh38
rs950035198 Chr.2: 121339211-121339211 on Build GRCh38
rs902519358 Chr.2: 121339297-121339297 on Build GRCh38
rs951047453 Chr.2: 121339435-121339435 on Build GRCh38
rs72965491 Chr.2: 121339089-121339089 on Build GRCh38
rs893978953 Chr.2: 121339107-121339107 on Build GRCh38
rs547645631 Chr.2: 121339522-121339523 on Build GRCh38
rs747062521 Chr.2: 121339430-121339431 on Build GRCh38
rs540674850 Chr.2: 121339170-121339171 on Build GRCh38
rs1053815928 Chr.2: 121339326-121339326 on Build GRCh38
rs764961023 Chr.2: 121339133-121339134 on Build GRCh38
rs1018466702 Chr.2: 121339381-121339381 on Build GRCh38
rs922174062 Chr.2: 121339269-121339269 on Build GRCh38
rs7421055 Chr.2: 121339128-121339128 on Build GRCh38
rs1046417821 Chr.2: 121339085-121339085 on Build GRCh38
rs1011621700 Chr.2: 121339134-121339135 on Build GRCh38
rs1001490210 Chr.2: 121339115-121339118 on Build GRCh38
rs867643082 Chr.2: 121339169-121339170 on Build GRCh38
rs991349479 Chr.2: 121339170-121339170 on Build GRCh38
rs111713630 Chr.2: 121339098-121339099 on Build GRCh38
rs999598666 Chr.2: 121339105-121339105 on Build GRCh38
rs965683551 Chr.2: 121339127-121339127 on Build GRCh38
rs937172095 Chr.2: 121339168-121339168 on Build GRCh38
rs951087132 Chr.2: 121339137-121339137 on Build GRCh38
rs935294179 Chr.2: 121339320-121339320 on Build GRCh38
rs975055640 Chr.2: 121339264-121339265 on Build GRCh38
rs138231136 Chr.2: 121339477-121339477 on Build GRCh38
rs145666707 Chr.2: 121339414-121339414 on Build GRCh38
rs552366730 Chr.2: 121339112-121339112 on Build GRCh38
rs966169235 Chr.2: 121339205-121339205 on Build GRCh38
rs570616718 Chr.2: 121339115-121339115 on Build GRCh38
rs766929124 Chr.2: 121339459-121339459 on Build GRCh38
rs369053885 Chr.2: 121339271-121339271 on Build GRCh38
rs1045838678 Chr.2: 121339390-121339390 on Build GRCh38
rs928489311 Chr.2: 121339391-121339391 on Build GRCh38
rs537753562 Chr.2: 121339304-121339304 on Build GRCh38
rs1006913983 Chr.2: 121339354-121339354 on Build GRCh38
rs1020000569 Chr.2: 121339135-121339136 on Build GRCh38
rs879133868 Chr.2: 121339132-121339137 on Build GRCh38
rs367614507 Chr.2: 121339171-121339171 on Build GRCh38
rs556088812 Chr.2: 121339382-121339382 on Build GRCh38
rs893861555 Chr.2: 121339331-121339331 on Build GRCh38
rs774158562 Chr.2: 121339452-121339452 on Build GRCh38
rs527623780 Chr.2: 121339357-121339357 on Build GRCh38
rs983875786 Chr.2: 121339144-121339144 on Build GRCh38
rs1025344373 Chr.2: 121339135-121339135 on Build GRCh38
rs909610245 Chr.2: 121339156-121339156 on Build GRCh38
rs13395135 Chr.2: 121339073-121339073 on Build GRCh38
rs902839271 Chr.2: 121339473-121339473 on Build GRCh38
rs892843362 Chr.2: 121339132-121339132 on Build GRCh38
rs907969705 Chr.2: 121339375-121339375 on Build GRCh38
rs13395216 Chr.2: 121339118-121339118 on Build GRCh38
rs893010894 Chr.2: 121339427-121339427 on Build GRCh38
rs958467540 Chr.2: 121339514-121339514 on Build GRCh38
rs1026279141 Chr.2: 121339424-121339424 on Build GRCh38
rs3839033 Chr.2: 121339132-121339133 on Build GRCh38
rs1042018261 Chr.2: 121339290-121339290 on Build GRCh38
rs991234949 Chr.2: 121339535-121339535 on Build GRCh38

View Full Product Details