Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 46168469-46168909 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPATC1L
Entrez Gene ID
84221
Gene Name
spermatogenesis and centriole associated 1-like
Gene Aliases
C21orf56
UniGene
Hs.381214
Gene Chromosome Location
Chr.21: 46161148-46185159 on Build GRCh38
Transcripts
NM_001142854.1 NM_032261.4 XM_005261188.4 XM_017028480.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1616185 c.331C>A p.Q111K Chr.21: 46168521-46168521 on Build GRCh38
4747167 c.327delC p.S110fs*28 Chr.21: 46168525-46168525 on Build GRCh38
5584637 c.345G>A p.K115K Chr.21: 46168507-46168507 on Build GRCh38
6873357 c.251C>T p.S84F Chr.21: 46168601-46168601 on Build GRCh38
1592794 c.249A>G p.T83T Chr.21: 46168603-46168603 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1004514339 Chr.21: 46168784-46168784 on Build GRCh38
rs986928642 Chr.21: 46168675-46168675 on Build GRCh38
rs577984682 Chr.21: 46168776-46168776 on Build GRCh38
rs11909543 Chr.21: 46168732-46168732 on Build GRCh38
rs919258004 Chr.21: 46168648-46168648 on Build GRCh38
rs569011729 Chr.21: 46168696-46168696 on Build GRCh38
rs74486947 Chr.21: 46168513-46168513 on Build GRCh38
rs749935979 Chr.21: 46168530-46168531 on Build GRCh38
rs1019177102 Chr.21: 46168533-46168533 on Build GRCh38
rs902121113 Chr.21: 46168731-46168731 on Build GRCh38
rs955161288 Chr.21: 46168669-46168669 on Build GRCh38
rs564783696 Chr.21: 46168875-46168875 on Build GRCh38
rs369176014 Chr.21: 46168626-46168626 on Build GRCh38
rs540248819 Chr.21: 46168622-46168622 on Build GRCh38
rs750914245 Chr.21: 46168541-46168541 on Build GRCh38
rs930703269 Chr.21: 46168500-46168500 on Build GRCh38
rs528241329 Chr.21: 46168567-46168567 on Build GRCh38
rs975072941 Chr.21: 46168659-46168659 on Build GRCh38
rs769712144 Chr.21: 46168656-46168656 on Build GRCh38
rs1010608695 Chr.21: 46168553-46168553 on Build GRCh38
rs755647414 Chr.21: 46168566-46168566 on Build GRCh38
rs139572736 Chr.21: 46168775-46168775 on Build GRCh38
rs780571636 Chr.21: 46168557-46168557 on Build GRCh38
rs776404445 Chr.21: 46168492-46168492 on Build GRCh38
rs146604399 Chr.21: 46168866-46168866 on Build GRCh38
rs61140994 Chr.21: 46168829-46168829 on Build GRCh38
rs984890280 Chr.21: 46168887-46168887 on Build GRCh38
rs749667531 Chr.21: 46168561-46168561 on Build GRCh38
rs755434231 Chr.21: 46168545-46168545 on Build GRCh38
rs764284214 Chr.21: 46168528-46168528 on Build GRCh38
rs1021971391 Chr.21: 46168817-46168817 on Build GRCh38
rs58861127 Chr.21: 46168821-46168821 on Build GRCh38
rs887022544 Chr.21: 46168587-46168587 on Build GRCh38
rs767593483 Chr.21: 46168540-46168540 on Build GRCh38
rs768986619 Chr.21: 46168635-46168635 on Build GRCh38
rs884134 Chr.21: 46168514-46168514 on Build GRCh38
rs1038273248 Chr.21: 46168809-46168809 on Build GRCh38
rs1044443805 Chr.21: 46168771-46168771 on Build GRCh38
rs962373317 Chr.21: 46168629-46168629 on Build GRCh38
rs770955212 Chr.21: 46168490-46168490 on Build GRCh38
rs972005663 Chr.21: 46168867-46168867 on Build GRCh38
rs765041623 Chr.21: 46168505-46168505 on Build GRCh38
rs759481276 Chr.21: 46168494-46168494 on Build GRCh38
rs561457465 Chr.21: 46168555-46168555 on Build GRCh38
rs1004141259 Chr.21: 46168621-46168621 on Build GRCh38
rs531977796 Chr.21: 46168647-46168647 on Build GRCh38
rs1049294112 Chr.21: 46168501-46168501 on Build GRCh38
rs934970090 Chr.21: 46168518-46168518 on Build GRCh38
rs533422076 Chr.21: 46168889-46168889 on Build GRCh38
rs575885966 Chr.21: 46168822-46168822 on Build GRCh38
rs898448624 Chr.21: 46168812-46168812 on Build GRCh38
rs1034893922 Chr.21: 46168575-46168575 on Build GRCh38
rs77889636 Chr.21: 46168551-46168551 on Build GRCh38
rs780707321 Chr.21: 46168627-46168627 on Build GRCh38
rs560270320 Chr.21: 46168719-46168719 on Build GRCh38
rs777273076 Chr.21: 46168568-46168568 on Build GRCh38
rs757252512 Chr.21: 46168539-46168539 on Build GRCh38
rs751705498 Chr.21: 46168532-46168532 on Build GRCh38
rs550872680 Chr.21: 46168663-46168663 on Build GRCh38
rs540345533 Chr.21: 46168888-46168888 on Build GRCh38
rs146820112 Chr.21: 46168709-46168709 on Build GRCh38
rs955088375 Chr.21: 46168826-46168826 on Build GRCh38
rs545720107 Chr.21: 46168504-46168504 on Build GRCh38
rs1028017544 Chr.21: 46168665-46168665 on Build GRCh38
rs907308900 Chr.21: 46168558-46168558 on Build GRCh38
rs919256522 Chr.21: 46168872-46168872 on Build GRCh38
rs538630265 Chr.21: 46168780-46168780 on Build GRCh38
rs566327442 Chr.21: 46168723-46168723 on Build GRCh38
rs762075482 Chr.21: 46168727-46168727 on Build GRCh38
rs28588245 Chr.21: 46168877-46168877 on Build GRCh38
rs548022105 Chr.21: 46168722-46168722 on Build GRCh38
rs928695503 Chr.21: 46168772-46168772 on Build GRCh38
rs200705216 Chr.21: 46168548-46168548 on Build GRCh38

View Full Product Details