Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 46182487-46182981 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPATC1L
Entrez Gene ID
84221
Gene Name
spermatogenesis and centriole associated 1-like
Gene Aliases
C21orf56
UniGene
Hs.381214
Gene Chromosome Location
Chr.21: 46161148-46185159 on Build GRCh38
Transcripts
NM_001142854.1 NM_032261.4 XM_005261188.4 XM_017028480.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6038520 c.155C>T p.A52V Chr.21: 46182662-46182662 on Build GRCh38
6410103 c.67C>T p.R23C Chr.21: 46182750-46182750 on Build GRCh38
3991995 c.12C>T p.G4G Chr.21: 46182805-46182805 on Build GRCh38
3964036 c.68G>T p.R23L Chr.21: 46182749-46182749 on Build GRCh38
5035426 c.45C>T p.N15N Chr.21: 46182772-46182772 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535746289 Chr.21: 46182643-46182643 on Build GRCh38
rs973736433 Chr.21: 46182688-46182688 on Build GRCh38
rs878961892 Chr.21: 46182582-46182582 on Build GRCh38
rs766841416 Chr.21: 46182628-46182629 on Build GRCh38
rs774467955 Chr.21: 46182618-46182618 on Build GRCh38
rs780872707 Chr.21: 46182858-46182858 on Build GRCh38
rs766201850 Chr.21: 46182893-46182903 on Build GRCh38
rs900648713 Chr.21: 46182614-46182614 on Build GRCh38
rs761333673 Chr.21: 46182634-46182634 on Build GRCh38
rs780631215 Chr.21: 46182720-46182720 on Build GRCh38
rs1004736497 Chr.21: 46182825-46182825 on Build GRCh38
rs983449018 Chr.21: 46182723-46182723 on Build GRCh38
rs745365276 Chr.21: 46182722-46182722 on Build GRCh38
rs560379686 Chr.21: 46182860-46182860 on Build GRCh38
rs779930837 Chr.21: 46182739-46182739 on Build GRCh38
rs779300435 Chr.21: 46182690-46182690 on Build GRCh38
rs1033489758 Chr.21: 46182638-46182638 on Build GRCh38
rs35921830 Chr.21: 46182831-46182832 on Build GRCh38
rs954957061 Chr.21: 46182883-46182883 on Build GRCh38
rs537523590 Chr.21: 46182622-46182622 on Build GRCh38
rs913515766 Chr.21: 46182771-46182771 on Build GRCh38
rs765535842 Chr.21: 46182823-46182823 on Build GRCh38
rs759406154 Chr.21: 46182708-46182708 on Build GRCh38
rs149414972 Chr.21: 46182859-46182860 on Build GRCh38
rs921984256 Chr.21: 46182769-46182769 on Build GRCh38
rs751196576 Chr.21: 46182692-46182692 on Build GRCh38
rs113955394 Chr.21: 46182836-46182836 on Build GRCh38
rs138357031 Chr.21: 46182910-46182910 on Build GRCh38
rs527851540 Chr.21: 46182529-46182529 on Build GRCh38
rs750444314 Chr.21: 46182687-46182687 on Build GRCh38
rs554119745 Chr.21: 46182928-46182928 on Build GRCh38
rs75252048 Chr.21: 46182518-46182518 on Build GRCh38
rs929330003 Chr.21: 46182555-46182555 on Build GRCh38
rs773224278 Chr.21: 46182789-46182789 on Build GRCh38
rs566694792 Chr.21: 46182898-46182898 on Build GRCh38
rs768906011 Chr.21: 46182617-46182617 on Build GRCh38
rs761452906 Chr.21: 46182616-46182616 on Build GRCh38
rs534033767 Chr.21: 46182801-46182801 on Build GRCh38
rs917798061 Chr.21: 46182552-46182552 on Build GRCh38
rs891783121 Chr.21: 46182620-46182620 on Build GRCh38
rs1053366967 Chr.21: 46182770-46182770 on Build GRCh38
rs1026669182 Chr.21: 46182849-46182849 on Build GRCh38
rs753098996 Chr.21: 46182824-46182824 on Build GRCh38
rs549793422 Chr.21: 46182627-46182627 on Build GRCh38
rs546782709 Chr.21: 46182844-46182844 on Build GRCh38
rs912790937 Chr.21: 46182596-46182596 on Build GRCh38
rs767481063 Chr.21: 46182749-46182749 on Build GRCh38
rs1001961658 Chr.21: 46182633-46182633 on Build GRCh38
rs760074300 Chr.21: 46182819-46182819 on Build GRCh38
rs1015470987 Chr.21: 46182822-46182822 on Build GRCh38
rs138704000 Chr.21: 46182561-46182561 on Build GRCh38
rs953717203 Chr.21: 46182854-46182854 on Build GRCh38
rs1001552831 Chr.21: 46182788-46182788 on Build GRCh38
rs543964172 Chr.21: 46182579-46182579 on Build GRCh38
rs572288533 Chr.21: 46182729-46182729 on Build GRCh38
rs754430859 Chr.21: 46182811-46182811 on Build GRCh38
rs755367806 Chr.21: 46182667-46182667 on Build GRCh38
rs912307813 Chr.21: 46182882-46182882 on Build GRCh38
rs942110944 Chr.21: 46182580-46182580 on Build GRCh38
rs979003791 Chr.21: 46182939-46182939 on Build GRCh38
rs973808746 Chr.21: 46182549-46182549 on Build GRCh38
rs950663287 Chr.21: 46182820-46182820 on Build GRCh38
rs149384097 Chr.21: 46182750-46182750 on Build GRCh38
rs532397772 Chr.21: 46182940-46182940 on Build GRCh38
rs544371646 Chr.21: 46182778-46182778 on Build GRCh38
rs1013488971 Chr.21: 46182659-46182659 on Build GRCh38
rs762711263 Chr.21: 46182613-46182613 on Build GRCh38
rs570245905 Chr.21: 46182573-46182573 on Build GRCh38
rs1057171091 Chr.21: 46182790-46182790 on Build GRCh38
rs1052037931 Chr.21: 46182624-46182624 on Build GRCh38
rs908182095 Chr.21: 46182530-46182530 on Build GRCh38
rs939709081 Chr.21: 46182535-46182535 on Build GRCh38
rs947665824 Chr.21: 46182930-46182930 on Build GRCh38
rs114558842 Chr.21: 46182787-46182787 on Build GRCh38
rs974814601 Chr.21: 46182753-46182753 on Build GRCh38
rs553756950 Chr.21: 46182719-46182719 on Build GRCh38
rs557963450 Chr.21: 46182772-46182772 on Build GRCh38
rs754303901 Chr.21: 46182661-46182661 on Build GRCh38
rs368890284 Chr.21: 46182631-46182631 on Build GRCh38
rs773844866 Chr.21: 46182630-46182630 on Build GRCh38
rs963359305 Chr.21: 46182851-46182851 on Build GRCh38
rs1013458020 Chr.21: 46182802-46182802 on Build GRCh38
rs992071175 Chr.21: 46182548-46182548 on Build GRCh38
rs762647619 Chr.21: 46182567-46182567 on Build GRCh38
rs143719440 Chr.21: 46182777-46182777 on Build GRCh38
rs55819909 Chr.21: 46182805-46182805 on Build GRCh38

View Full Product Details