Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 17143011-17143509 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CECR5
Entrez Gene ID
27440
Gene Name
cat eye syndrome chromosome region, candidate 5
Gene Aliases
-
UniGene
Hs.26890
Gene Chromosome Location
Chr.22: 17137520-17165287 on Build GRCh38
Transcripts
NM_017829.5 NM_033070.2 XM_005261236.3 XM_011546126.2 XM_011546127.1 XM_017028781.1 XM_017028782.1 XM_017028783.1 XM_017028784.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
444666 c.563G>A p.R188H Chr.22: 17143106-17143106 on Build GRCh38
4815543 c.473G>A p.R158H Chr.22: 17143106-17143106 on Build GRCh38
2891104 c.545C>T p.P182L Chr.22: 17143124-17143124 on Build GRCh38
4812055 c.459G>A p.R153R Chr.22: 17143120-17143120 on Build GRCh38
7213108 c.472C>T p.R158C Chr.22: 17143107-17143107 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs928144219 Chr.22: 17143185-17143185 on Build GRCh38
rs939100270 Chr.22: 17143206-17143206 on Build GRCh38
rs367588837 Chr.22: 17143175-17143175 on Build GRCh38
rs956659348 Chr.22: 17143408-17143408 on Build GRCh38
rs759095965 Chr.22: 17143098-17143098 on Build GRCh38
rs764162912 Chr.22: 17143143-17143143 on Build GRCh38
rs775146956 Chr.22: 17143180-17143180 on Build GRCh38
rs570484714 Chr.22: 17143354-17143354 on Build GRCh38
rs879806193 Chr.22: 17143485-17143485 on Build GRCh38
rs146462332 Chr.22: 17143173-17143173 on Build GRCh38
rs765576371 Chr.22: 17143258-17143258 on Build GRCh38
rs374912416 Chr.22: 17143107-17143107 on Build GRCh38
rs969612227 Chr.22: 17143426-17143426 on Build GRCh38
rs879584705 Chr.22: 17143124-17143124 on Build GRCh38
rs370743035 Chr.22: 17143059-17143059 on Build GRCh38
rs770234564 Chr.22: 17143094-17143094 on Build GRCh38
rs560893696 Chr.22: 17143110-17143110 on Build GRCh38
rs150388066 Chr.22: 17143483-17143483 on Build GRCh38
rs994464595 Chr.22: 17143272-17143272 on Build GRCh38
rs1048930883 Chr.22: 17143281-17143281 on Build GRCh38
rs1036554724 Chr.22: 17143234-17143234 on Build GRCh38
rs745883462 Chr.22: 17143340-17143340 on Build GRCh38
rs764969011 Chr.22: 17143113-17143113 on Build GRCh38
rs201579046 Chr.22: 17143055-17143055 on Build GRCh38
rs769218856 Chr.22: 17143106-17143106 on Build GRCh38
rs981457514 Chr.22: 17143430-17143430 on Build GRCh38
rs773818594 Chr.22: 17143032-17143032 on Build GRCh38
rs2302521 Chr.22: 17143182-17143182 on Build GRCh38
rs939552794 Chr.22: 17143446-17143446 on Build GRCh38
rs773468883 Chr.22: 17143111-17143111 on Build GRCh38
rs928143748 Chr.22: 17143436-17143438 on Build GRCh38
rs573740970 Chr.22: 17143384-17143384 on Build GRCh38
rs371838901 Chr.22: 17143169-17143169 on Build GRCh38
rs765575668 Chr.22: 17143107-17143107 on Build GRCh38
rs2302522 Chr.22: 17143325-17143325 on Build GRCh38
rs897492332 Chr.22: 17143248-17143248 on Build GRCh38
rs555428021 Chr.22: 17143375-17143375 on Build GRCh38
rs140351855 Chr.22: 17143482-17143482 on Build GRCh38
rs368034523 Chr.22: 17143112-17143112 on Build GRCh38
rs190413708 Chr.22: 17143123-17143123 on Build GRCh38
rs915475403 Chr.22: 17143423-17143423 on Build GRCh38
rs549019615 Chr.22: 17143087-17143087 on Build GRCh38
rs781035453 Chr.22: 17143085-17143085 on Build GRCh38
rs889045629 Chr.22: 17143291-17143291 on Build GRCh38
rs758257649 Chr.22: 17143132-17143132 on Build GRCh38
rs772086839 Chr.22: 17143171-17143171 on Build GRCh38
rs577817549 Chr.22: 17143460-17143460 on Build GRCh38
rs989692926 Chr.22: 17143138-17143138 on Build GRCh38
rs772264029 Chr.22: 17143471-17143472 on Build GRCh38
rs749717734 Chr.22: 17143168-17143168 on Build GRCh38
rs866838767 Chr.22: 17143126-17143126 on Build GRCh38
rs796136381 Chr.22: 17143457-17143457 on Build GRCh38
rs146667524 Chr.22: 17143474-17143474 on Build GRCh38
rs749692710 Chr.22: 17143104-17143104 on Build GRCh38
rs140927685 Chr.22: 17143459-17143459 on Build GRCh38
rs770489137 Chr.22: 17143080-17143080 on Build GRCh38
rs745480881 Chr.22: 17143154-17143154 on Build GRCh38
rs777092962 Chr.22: 17143056-17143056 on Build GRCh38
rs1036071093 Chr.22: 17143455-17143457 on Build GRCh38
rs200576728 Chr.22: 17143151-17143151 on Build GRCh38
rs756877788 Chr.22: 17143148-17143148 on Build GRCh38
rs935360383 Chr.22: 17143129-17143129 on Build GRCh38
rs375622350 Chr.22: 17143308-17143308 on Build GRCh38
rs776031098 Chr.22: 17143103-17143103 on Build GRCh38
rs753132071 Chr.22: 17143138-17143138 on Build GRCh38
rs534702416 Chr.22: 17143411-17143411 on Build GRCh38
rs780507876 Chr.22: 17143162-17143162 on Build GRCh38
rs751006046 Chr.22: 17143147-17143147 on Build GRCh38
rs888805513 Chr.22: 17143475-17143475 on Build GRCh38
rs144577734 Chr.22: 17143468-17143468 on Build GRCh38
rs766648486 Chr.22: 17143179-17143179 on Build GRCh38
rs780871971 Chr.22: 17143152-17143152 on Build GRCh38
rs755834126 Chr.22: 17143157-17143157 on Build GRCh38
rs868410482 Chr.22: 17143035-17143035 on Build GRCh38
rs750773000 Chr.22: 17143307-17143307 on Build GRCh38
rs923957528 Chr.22: 17143069-17143069 on Build GRCh38

View Full Product Details