Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 17144885-17145325 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CECR5
Entrez Gene ID
27440
Gene Name
cat eye syndrome chromosome region, candidate 5
Gene Aliases
-
UniGene
Hs.26890
Gene Chromosome Location
Chr.22: 17137520-17165287 on Build GRCh38
Transcripts
NM_017829.5 NM_033070.2 XM_005261236.3 XM_011546126.2 XM_011546127.1 XM_017028781.1 XM_017028782.1 XM_017028783.1 XM_017028784.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6262223 c.414C>A p.D138E Chr.22: 17145057-17145057 on Build GRCh38
2891107 c.523C>T p.R175W Chr.22: 17145038-17145038 on Build GRCh38
6626023 c.430C>T p.R144W Chr.22: 17145041-17145041 on Build GRCh38
3740331 c.519G>A p.E173E Chr.22: 17145042-17145042 on Build GRCh38
4914122 c.354-1G>T p.? Chr.22: 17145118-17145118 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771115671 Chr.22: 17145033-17145033 on Build GRCh38
rs1025521564 Chr.22: 17145163-17145163 on Build GRCh38
rs1016277457 Chr.22: 17145233-17145233 on Build GRCh38
rs765869148 Chr.22: 17145159-17145159 on Build GRCh38
rs746035661 Chr.22: 17145106-17145106 on Build GRCh38
rs751544336 Chr.22: 17145136-17145137 on Build GRCh38
rs199989327 Chr.22: 17145038-17145038 on Build GRCh38
rs865849519 Chr.22: 17145079-17145079 on Build GRCh38
rs749669932 Chr.22: 17145205-17145205 on Build GRCh38
rs951258080 Chr.22: 17144914-17144914 on Build GRCh38
rs763919548 Chr.22: 17144979-17144979 on Build GRCh38
rs372710015 Chr.22: 17145117-17145117 on Build GRCh38
rs779162881 Chr.22: 17145009-17145009 on Build GRCh38
rs749584297 Chr.22: 17145001-17145001 on Build GRCh38
rs779267988 Chr.22: 17145092-17145092 on Build GRCh38
rs760706545 Chr.22: 17145149-17145149 on Build GRCh38
rs1055453439 Chr.22: 17145109-17145109 on Build GRCh38
rs772570016 Chr.22: 17145024-17145024 on Build GRCh38
rs375737033 Chr.22: 17145122-17145122 on Build GRCh38
rs763098773 Chr.22: 17145030-17145031 on Build GRCh38
rs779821303 Chr.22: 17144999-17144999 on Build GRCh38
rs112248558 Chr.22: 17144930-17144930 on Build GRCh38
rs536344493 Chr.22: 17145036-17145036 on Build GRCh38
rs774184853 Chr.22: 17145118-17145118 on Build GRCh38
rs776965135 Chr.22: 17145037-17145037 on Build GRCh38
rs55754435 Chr.22: 17144939-17144939 on Build GRCh38
rs749947487 Chr.22: 17145074-17145074 on Build GRCh38
rs143121345 Chr.22: 17144909-17144909 on Build GRCh38
rs749968227 Chr.22: 17144989-17144989 on Build GRCh38
rs764315054 Chr.22: 17145096-17145098 on Build GRCh38
rs778294990 Chr.22: 17145098-17145098 on Build GRCh38
rs150711823 Chr.22: 17145093-17145093 on Build GRCh38
rs181942834 Chr.22: 17145080-17145080 on Build GRCh38
rs561586536 Chr.22: 17145197-17145197 on Build GRCh38
rs761785628 Chr.22: 17145048-17145048 on Build GRCh38
rs369297006 Chr.22: 17145105-17145105 on Build GRCh38
rs139992562 Chr.22: 17145107-17145107 on Build GRCh38
rs975260124 Chr.22: 17145291-17145291 on Build GRCh38
rs35665085 Chr.22: 17145025-17145025 on Build GRCh38
rs140387564 Chr.22: 17145089-17145089 on Build GRCh38
rs963475883 Chr.22: 17145235-17145235 on Build GRCh38
rs770998254 Chr.22: 17145101-17145101 on Build GRCh38
rs759168169 Chr.22: 17145166-17145166 on Build GRCh38
rs375791641 Chr.22: 17145028-17145028 on Build GRCh38
rs773330618 Chr.22: 17145145-17145145 on Build GRCh38
rs541910955 Chr.22: 17145132-17145132 on Build GRCh38
rs753288897 Chr.22: 17145161-17145161 on Build GRCh38
rs555709557 Chr.22: 17144935-17144935 on Build GRCh38
rs144220705 Chr.22: 17145091-17145091 on Build GRCh38
rs371956696 Chr.22: 17145041-17145041 on Build GRCh38
rs561616824 Chr.22: 17145040-17145040 on Build GRCh38
rs1014125329 Chr.22: 17145155-17145155 on Build GRCh38
rs369910266 Chr.22: 17144995-17144995 on Build GRCh38
rs751465979 Chr.22: 17144980-17144980 on Build GRCh38
rs768335992 Chr.22: 17145056-17145056 on Build GRCh38
rs532624454 Chr.22: 17145211-17145211 on Build GRCh38
rs755415589 Chr.22: 17145007-17145007 on Build GRCh38
rs771356381 Chr.22: 17145250-17145250 on Build GRCh38
rs201826876 Chr.22: 17145099-17145099 on Build GRCh38
rs748646198 Chr.22: 17145013-17145013 on Build GRCh38
rs758905911 Chr.22: 17145095-17145095 on Build GRCh38
rs757292200 Chr.22: 17144981-17144981 on Build GRCh38
rs921947600 Chr.22: 17145292-17145292 on Build GRCh38

View Full Product Details