Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 17188111-17188570 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CECR1
Entrez Gene ID
51816
Gene Name
cat eye syndrome chromosome region, candidate 1
Gene Aliases
ADA2, ADGF, IDGFL, PAN, SNEDS
UniGene
Hs.170310
Gene Chromosome Location
Chr.22: 17178790-17221854 on Build GRCh38
Transcripts
NM_001282225.1 NM_001282226.1 NM_001282227.1 NM_001282228.1 NM_001282229.1 NM_177405.2 XM_006724080.3 XM_011546133.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3842132 c.332C>T p.P111L Chr.22: 17188365-17188365 on Build GRCh38
4002079 c.1004A>G p.H335R Chr.22: 17188416-17188416 on Build GRCh38
725680 c.1072G>C p.G358R Chr.22: 17188348-17188348 on Build GRCh38
3551952 c.275C>T p.S92F Chr.22: 17188422-17188422 on Build GRCh38
725678 c.1071C>T p.A357A Chr.22: 17188349-17188349 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777906358 Chr.22: 17188329-17188329 on Build GRCh38
rs913957832 Chr.22: 17188541-17188541 on Build GRCh38
rs760228190 Chr.22: 17188443-17188443 on Build GRCh38
rs368021522 Chr.22: 17188322-17188322 on Build GRCh38
rs971794431 Chr.22: 17188275-17188275 on Build GRCh38
rs772102280 Chr.22: 17188492-17188492 on Build GRCh38
rs775440641 Chr.22: 17188342-17188342 on Build GRCh38
rs373498904 Chr.22: 17188360-17188360 on Build GRCh38
rs566686180 Chr.22: 17188510-17188510 on Build GRCh38
rs202040790 Chr.22: 17188445-17188445 on Build GRCh38
rs142508522 Chr.22: 17188398-17188398 on Build GRCh38
rs757221265 Chr.22: 17188177-17188177 on Build GRCh38
rs763640700 Chr.22: 17188296-17188296 on Build GRCh38
rs758370176 Chr.22: 17188388-17188388 on Build GRCh38
rs764068777 Chr.22: 17188472-17188472 on Build GRCh38
rs749226595 Chr.22: 17188345-17188345 on Build GRCh38
rs186426805 Chr.22: 17188213-17188213 on Build GRCh38
rs573337330 Chr.22: 17188407-17188407 on Build GRCh38
rs748240651 Chr.22: 17188323-17188323 on Build GRCh38
rs148435119 Chr.22: 17188485-17188485 on Build GRCh38
rs9619005 Chr.22: 17188516-17188516 on Build GRCh38
rs753078362 Chr.22: 17188462-17188462 on Build GRCh38
rs980932650 Chr.22: 17188200-17188200 on Build GRCh38
rs116020027 Chr.22: 17188355-17188355 on Build GRCh38
rs374130382 Chr.22: 17188447-17188447 on Build GRCh38
rs943246871 Chr.22: 17188149-17188149 on Build GRCh38
rs200622282 Chr.22: 17188321-17188321 on Build GRCh38
rs369716341 Chr.22: 17188435-17188435 on Build GRCh38
rs910212434 Chr.22: 17188506-17188506 on Build GRCh38
rs45511697 Chr.22: 17188348-17188348 on Build GRCh38
rs776009181 Chr.22: 17188319-17188320 on Build GRCh38
rs199557749 Chr.22: 17188352-17188352 on Build GRCh38
rs752798667 Chr.22: 17188387-17188387 on Build GRCh38
rs781617794 Chr.22: 17188484-17188484 on Build GRCh38
rs377213003 Chr.22: 17188413-17188413 on Build GRCh38
rs769409589 Chr.22: 17188332-17188332 on Build GRCh38
rs1030975354 Chr.22: 17188170-17188170 on Build GRCh38
rs527514631 Chr.22: 17188515-17188515 on Build GRCh38
rs191772679 Chr.22: 17188377-17188377 on Build GRCh38
rs751730137 Chr.22: 17188474-17188474 on Build GRCh38
rs766756743 Chr.22: 17188319-17188319 on Build GRCh38
rs2231495 Chr.22: 17188416-17188416 on Build GRCh38
rs7284498 Chr.22: 17188309-17188309 on Build GRCh38
rs757523498 Chr.22: 17188479-17188479 on Build GRCh38
rs770635459 Chr.22: 17188444-17188444 on Build GRCh38
rs374159743 Chr.22: 17188508-17188508 on Build GRCh38
rs368417650 Chr.22: 17188297-17188297 on Build GRCh38
rs139750129 Chr.22: 17188449-17188449 on Build GRCh38
rs115273803 Chr.22: 17188252-17188252 on Build GRCh38
rs763530892 Chr.22: 17188464-17188464 on Build GRCh38
rs532424743 Chr.22: 17188207-17188207 on Build GRCh38
rs749835835 Chr.22: 17188320-17188321 on Build GRCh38
rs778770185 Chr.22: 17188486-17188486 on Build GRCh38
rs368334057 Chr.22: 17188311-17188311 on Build GRCh38
rs747287555 Chr.22: 17188331-17188331 on Build GRCh38
rs969945323 Chr.22: 17188199-17188199 on Build GRCh38
rs748143597 Chr.22: 17188421-17188421 on Build GRCh38
rs144447953 Chr.22: 17188349-17188349 on Build GRCh38
rs770047872 Chr.22: 17188513-17188513 on Build GRCh38
rs374974565 Chr.22: 17188351-17188351 on Build GRCh38
rs767072492 Chr.22: 17188350-17188350 on Build GRCh38
rs918915237 Chr.22: 17188294-17188294 on Build GRCh38
rs760030241 Chr.22: 17188376-17188376 on Build GRCh38
rs892506452 Chr.22: 17188176-17188176 on Build GRCh38
rs923107675 Chr.22: 17188324-17188324 on Build GRCh38
rs201667612 Chr.22: 17188354-17188354 on Build GRCh38
rs2231496 Chr.22: 17188261-17188261 on Build GRCh38
rs757613201 Chr.22: 17188411-17188411 on Build GRCh38
rs761524963 Chr.22: 17188308-17188308 on Build GRCh38
rs117996853 Chr.22: 17188189-17188189 on Build GRCh38
rs566321113 Chr.22: 17188292-17188292 on Build GRCh38
rs565311423 Chr.22: 17188198-17188198 on Build GRCh38
rs747974783 Chr.22: 17188491-17188491 on Build GRCh38
rs757020211 Chr.22: 17188250-17188250 on Build GRCh38
rs576842036 Chr.22: 17188328-17188328 on Build GRCh38
rs74317375 Chr.22: 17188375-17188375 on Build GRCh38
rs998680954 Chr.22: 17188166-17188166 on Build GRCh38
rs934506220 Chr.22: 17188511-17188511 on Build GRCh38
rs568819025 Chr.22: 17188234-17188234 on Build GRCh38
rs868563664 Chr.22: 17188458-17188458 on Build GRCh38
rs771413004 Chr.22: 17188428-17188428 on Build GRCh38
rs984398217 Chr.22: 17188504-17188505 on Build GRCh38
rs183554702 Chr.22: 17188509-17188509 on Build GRCh38
rs756382115 Chr.22: 17188318-17188318 on Build GRCh38
rs567210104 Chr.22: 17188153-17188153 on Build GRCh38
rs562538260 Chr.22: 17188507-17188507 on Build GRCh38

View Full Product Details