Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 17594303-17594799 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V1E1
Entrez Gene ID
529
Gene Name
ATPase H+ transporting V1 subunit E1
Gene Aliases
ATP6E, ATP6E2, ATP6V1E, P31, Vma4
UniGene
Hs.517338
Gene Chromosome Location
Chr.22: 17592136-17628822 on Build GRCh38
Transcripts
NM_001039366.1 NM_001039367.1 NM_001696.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6608142 c.563G>A p.R188H Chr.22: 17594584-17594584 on Build GRCh38
7197034 c.536G>T p.G179V Chr.22: 17594611-17594611 on Build GRCh38
4297105 c.601G>A p.D201N Chr.22: 17594546-17594546 on Build GRCh38
6196025 c.553A>C p.N185H Chr.22: 17594594-17594594 on Build GRCh38
725656 c.570A>C p.I190I Chr.22: 17594577-17594577 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112106599 Chr.22: 17594776-17594776 on Build GRCh38
rs780063572 Chr.22: 17594589-17594589 on Build GRCh38
rs1019636765 Chr.22: 17594425-17594425 on Build GRCh38
rs974018394 Chr.22: 17594423-17594423 on Build GRCh38
rs372524876 Chr.22: 17594513-17594513 on Build GRCh38
rs1041815235 Chr.22: 17594507-17594507 on Build GRCh38
rs953499842 Chr.22: 17594713-17594713 on Build GRCh38
rs537624234 Chr.22: 17594465-17594465 on Build GRCh38
rs201567961 Chr.22: 17594560-17594560 on Build GRCh38
rs116576310 Chr.22: 17594677-17594677 on Build GRCh38
rs369890056 Chr.22: 17594592-17594592 on Build GRCh38
rs962238387 Chr.22: 17594388-17594394 on Build GRCh38
rs76360481 Chr.22: 17594514-17594514 on Build GRCh38
rs920751871 Chr.22: 17594459-17594459 on Build GRCh38
rs930519479 Chr.22: 17594689-17594689 on Build GRCh38
rs906876439 Chr.22: 17594574-17594574 on Build GRCh38
rs371758504 Chr.22: 17594517-17594517 on Build GRCh38
rs1019583203 Chr.22: 17594724-17594724 on Build GRCh38
rs757617918 Chr.22: 17594540-17594540 on Build GRCh38
rs769018395 Chr.22: 17594732-17594732 on Build GRCh38
rs781632221 Chr.22: 17594559-17594559 on Build GRCh38
rs1015725045 Chr.22: 17594337-17594337 on Build GRCh38
rs77950405 Chr.22: 17594569-17594569 on Build GRCh38
rs398081660 Chr.22: 17594629-17594629 on Build GRCh38
rs889020026 Chr.22: 17594371-17594371 on Build GRCh38
rs201956772 Chr.22: 17594621-17594621 on Build GRCh38
rs745471844 Chr.22: 17594688-17594688 on Build GRCh38
rs751944912 Chr.22: 17594527-17594527 on Build GRCh38
rs369068029 Chr.22: 17594484-17594484 on Build GRCh38
rs966076592 Chr.22: 17594749-17594749 on Build GRCh38
rs764799703 Chr.22: 17594498-17594498 on Build GRCh38
rs986391677 Chr.22: 17594723-17594723 on Build GRCh38
rs894300508 Chr.22: 17594518-17594518 on Build GRCh38
rs944678304 Chr.22: 17594488-17594488 on Build GRCh38
rs75514826 Chr.22: 17594777-17594777 on Build GRCh38
rs373830719 Chr.22: 17594647-17594647 on Build GRCh38
rs752278157 Chr.22: 17594499-17594499 on Build GRCh38
rs1036149194 Chr.22: 17594675-17594675 on Build GRCh38
rs1004246425 Chr.22: 17594614-17594614 on Build GRCh38
rs779072261 Chr.22: 17594619-17594619 on Build GRCh38
rs768892237 Chr.22: 17594596-17594596 on Build GRCh38
rs180781581 Chr.22: 17594735-17594735 on Build GRCh38
rs796472185 Chr.22: 17594621-17594622 on Build GRCh38
rs777399205 Chr.22: 17594406-17594406 on Build GRCh38
rs1012810041 Chr.22: 17594520-17594520 on Build GRCh38
rs35683311 Chr.22: 17594621-17594621 on Build GRCh38
rs35670859 Chr.22: 17594545-17594546 on Build GRCh38
rs376802684 Chr.22: 17594668-17594668 on Build GRCh38
rs1003979218 Chr.22: 17594333-17594333 on Build GRCh38
rs748724772 Chr.22: 17594441-17594441 on Build GRCh38
rs773943708 Chr.22: 17594620-17594621 on Build GRCh38
rs759548608 Chr.22: 17594497-17594497 on Build GRCh38
rs748948153 Chr.22: 17594516-17594516 on Build GRCh38
rs936114279 Chr.22: 17594778-17594779 on Build GRCh38
rs937613166 Chr.22: 17594541-17594541 on Build GRCh38
rs568978547 Chr.22: 17594407-17594407 on Build GRCh38
rs76511316 Chr.22: 17594568-17594568 on Build GRCh38
rs1008797801 Chr.22: 17594415-17594415 on Build GRCh38
rs927591528 Chr.22: 17594523-17594523 on Build GRCh38
rs548401924 Chr.22: 17594472-17594472 on Build GRCh38
rs554265244 Chr.22: 17594621-17594622 on Build GRCh38
rs770562673 Chr.22: 17594632-17594632 on Build GRCh38
rs993006633 Chr.22: 17594557-17594557 on Build GRCh38
rs746808585 Chr.22: 17594622-17594622 on Build GRCh38

View Full Product Details