Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 17726991-17727496 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL2L13
Entrez Gene ID
23786
Gene Name
BCL2 like 13
Gene Aliases
BCL-RAMBO, Bcl2-L-13, MIL1
UniGene
Hs.631672
Gene Chromosome Location
Chr.22: 17628855-17730855 on Build GRCh38
Transcripts
NM_001270726.1 NM_001270727.1 NM_001270728.1 NM_001270729.1 NM_001270730.1 NM_001270731.1 NM_001270732.1 NM_001270733.1 NM_001270734.1 NM_001270735.1 NM_015367.3 NR_073068.1 NR_073069.1 XM_011546119.1 XM_011546120.1 XM_011546121.2 XM_011546123.1 XM_017028725.1 XM_017028726.1 XM_017028727.1 XM_017028728.1 XM_017028729.1 XM_017028730.1 XM_017028731.1 XM_017028732.1 XM_017028734.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4665423 c.1181A>G p.E394G Chr.22: 17727257-17727257 on Build GRCh38
79543 c.948C>T p.P316P Chr.22: 17727024-17727024 on Build GRCh38
345183 c.1124C>G p.S375C Chr.22: 17727200-17727200 on Build GRCh38
3552023 c.1138C>T p.L380F Chr.22: 17727214-17727214 on Build GRCh38
3363534 c.1315G>A p.G439S Chr.22: 17727391-17727391 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759873601 Chr.22: 17727297-17727297 on Build GRCh38
rs374385339 Chr.22: 17727276-17727276 on Build GRCh38
rs768699658 Chr.22: 17727108-17727108 on Build GRCh38
rs749159499 Chr.22: 17727101-17727101 on Build GRCh38
rs149030911 Chr.22: 17727099-17727099 on Build GRCh38
rs769593795 Chr.22: 17727024-17727024 on Build GRCh38
rs778881285 Chr.22: 17727367-17727367 on Build GRCh38
rs968421172 Chr.22: 17727312-17727312 on Build GRCh38
rs958501107 Chr.22: 17727335-17727335 on Build GRCh38
rs368123385 Chr.22: 17727358-17727358 on Build GRCh38
rs904543255 Chr.22: 17727178-17727178 on Build GRCh38
rs931525387 Chr.22: 17727153-17727153 on Build GRCh38
rs753166854 Chr.22: 17727305-17727305 on Build GRCh38
rs144030966 Chr.22: 17727223-17727223 on Build GRCh38
rs571557906 Chr.22: 17727144-17727144 on Build GRCh38
rs1010026322 Chr.22: 17727296-17727296 on Build GRCh38
rs769440316 Chr.22: 17727098-17727098 on Build GRCh38
rs764780025 Chr.22: 17727475-17727475 on Build GRCh38
rs780738515 Chr.22: 17727216-17727218 on Build GRCh38
rs942352727 Chr.22: 17727097-17727097 on Build GRCh38
rs748118306 Chr.22: 17727252-17727252 on Build GRCh38
rs764139260 Chr.22: 17727310-17727310 on Build GRCh38
rs147289721 Chr.22: 17727275-17727275 on Build GRCh38
rs61740640 Chr.22: 17727376-17727376 on Build GRCh38
rs770347827 Chr.22: 17727410-17727410 on Build GRCh38
rs547249047 Chr.22: 17727072-17727072 on Build GRCh38
rs374669414 Chr.22: 17727321-17727321 on Build GRCh38
rs555307632 Chr.22: 17727361-17727361 on Build GRCh38
rs763440210 Chr.22: 17727429-17727429 on Build GRCh38
rs199900741 Chr.22: 17727070-17727070 on Build GRCh38
rs748065248 Chr.22: 17727375-17727375 on Build GRCh38
rs11705271 Chr.22: 17727362-17727362 on Build GRCh38
rs200014585 Chr.22: 17727434-17727434 on Build GRCh38
rs999828949 Chr.22: 17727462-17727462 on Build GRCh38
rs201972889 Chr.22: 17727196-17727196 on Build GRCh38
rs763484431 Chr.22: 17727027-17727027 on Build GRCh38
rs377097453 Chr.22: 17727468-17727468 on Build GRCh38
rs746569142 Chr.22: 17727279-17727279 on Build GRCh38
rs757551138 Chr.22: 17727215-17727215 on Build GRCh38
rs199996614 Chr.22: 17727444-17727444 on Build GRCh38
rs775967485 Chr.22: 17727415-17727415 on Build GRCh38
rs758402613 Chr.22: 17727465-17727465 on Build GRCh38
rs755424598 Chr.22: 17727131-17727131 on Build GRCh38
rs778828085 Chr.22: 17727452-17727452 on Build GRCh38
rs756329056 Chr.22: 17727446-17727446 on Build GRCh38
rs770293996 Chr.22: 17727365-17727367 on Build GRCh38
rs771669104 Chr.22: 17727264-17727264 on Build GRCh38
rs139164101 Chr.22: 17727277-17727277 on Build GRCh38
rs767345696 Chr.22: 17727048-17727048 on Build GRCh38
rs747258223 Chr.22: 17727086-17727086 on Build GRCh38
rs1034245755 Chr.22: 17727322-17727322 on Build GRCh38
rs750105179 Chr.22: 17727053-17727053 on Build GRCh38
rs765836010 Chr.22: 17727139-17727139 on Build GRCh38
rs764654153 Chr.22: 17727433-17727433 on Build GRCh38
rs776853685 Chr.22: 17727281-17727281 on Build GRCh38
rs749176101 Chr.22: 17727238-17727238 on Build GRCh38
rs761546311 Chr.22: 17727039-17727039 on Build GRCh38
rs1014350605 Chr.22: 17727454-17727454 on Build GRCh38
rs767147739 Chr.22: 17727119-17727119 on Build GRCh38
rs775247146 Chr.22: 17727025-17727025 on Build GRCh38
rs761894973 Chr.22: 17727441-17727441 on Build GRCh38
rs775778280 Chr.22: 17727474-17727474 on Build GRCh38
rs111457323 Chr.22: 17727380-17727380 on Build GRCh38
rs79531470 Chr.22: 17727443-17727443 on Build GRCh38
rs200337502 Chr.22: 17727061-17727061 on Build GRCh38
rs752597147 Chr.22: 17727456-17727456 on Build GRCh38
rs933526163 Chr.22: 17727104-17727104 on Build GRCh38
rs768340766 Chr.22: 17727245-17727245 on Build GRCh38
rs139273724 Chr.22: 17727379-17727379 on Build GRCh38
rs776393541 Chr.22: 17727021-17727021 on Build GRCh38
rs377062475 Chr.22: 17727122-17727122 on Build GRCh38
rs117992732 Chr.22: 17727411-17727411 on Build GRCh38
rs774374858 Chr.22: 17727247-17727247 on Build GRCh38
rs200217716 Chr.22: 17727071-17727071 on Build GRCh38
rs766620136 Chr.22: 17727127-17727127 on Build GRCh38
rs777866528 Chr.22: 17727182-17727182 on Build GRCh38
rs777901838 Chr.22: 17727075-17727075 on Build GRCh38
rs577953066 Chr.22: 17727426-17727426 on Build GRCh38
rs746594037 Chr.22: 17727381-17727381 on Build GRCh38
rs9306198 Chr.22: 17727154-17727154 on Build GRCh38
rs565787483 Chr.22: 17727363-17727363 on Build GRCh38
rs376011860 Chr.22: 17727069-17727069 on Build GRCh38
rs1048649467 Chr.22: 17727383-17727383 on Build GRCh38
rs144977669 Chr.22: 17727364-17727364 on Build GRCh38
rs772950086 Chr.22: 17727121-17727121 on Build GRCh38
rs559607869 Chr.22: 17727068-17727068 on Build GRCh38
rs761341838 Chr.22: 17727118-17727118 on Build GRCh38
rs772327798 Chr.22: 17727017-17727017 on Build GRCh38
rs377746710 Chr.22: 17727412-17727412 on Build GRCh38
rs773729787 Chr.22: 17727111-17727111 on Build GRCh38
rs539074202 Chr.22: 17727280-17727280 on Build GRCh38
rs757418825 Chr.22: 17727338-17727338 on Build GRCh38
rs551684928 Chr.22: 17727038-17727038 on Build GRCh38
rs149561290 Chr.22: 17727399-17727399 on Build GRCh38
rs746369849 Chr.22: 17727334-17727334 on Build GRCh38
rs772272923 Chr.22: 17727074-17727074 on Build GRCh38
rs144171230 Chr.22: 17727476-17727476 on Build GRCh38
rs765529732 Chr.22: 17727302-17727302 on Build GRCh38
rs990839577 Chr.22: 17727100-17727100 on Build GRCh38
rs369699924 Chr.22: 17727129-17727129 on Build GRCh38

View Full Product Details