Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 17737910-17738400 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BID
Entrez Gene ID
637
Gene Name
BH3 interacting domain death agonist
Gene Aliases
FP497
UniGene
Hs.517145
Gene Chromosome Location
Chr.22: 17734140-17774665 on Build GRCh38
Transcripts
NM_001196.3 NM_001244567.1 NM_001244569.1 NM_001244570.1 NM_001244572.1 NM_197966.2 NM_197967.2 XM_017028906.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7103999 c.609G>C p.K203N Chr.22: 17738122-17738122 on Build GRCh38
4792976 c.626C>T p.T209M Chr.22: 17738105-17738105 on Build GRCh38
6223671 c.623A>T p.H208L Chr.22: 17738108-17738108 on Build GRCh38
3759024 c.666T>C p.N222N Chr.22: 17738065-17738065 on Build GRCh38
7103998 c.663G>A p.V221V Chr.22: 17738068-17738068 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748906450 Chr.22: 17737985-17737985 on Build GRCh38
rs925556370 Chr.22: 17737945-17737945 on Build GRCh38
rs538403130 Chr.22: 17737941-17737941 on Build GRCh38
rs778272070 Chr.22: 17738151-17738151 on Build GRCh38
rs369914189 Chr.22: 17738101-17738101 on Build GRCh38
rs754672801 Chr.22: 17737986-17737986 on Build GRCh38
rs773039137 Chr.22: 17738119-17738119 on Build GRCh38
rs747484734 Chr.22: 17738097-17738097 on Build GRCh38
rs752495851 Chr.22: 17738158-17738160 on Build GRCh38
rs760590436 Chr.22: 17738120-17738120 on Build GRCh38
rs199554819 Chr.22: 17738008-17738008 on Build GRCh38
rs112498689 Chr.22: 17738231-17738231 on Build GRCh38
rs191428241 Chr.22: 17738305-17738305 on Build GRCh38
rs746410035 Chr.22: 17738219-17738219 on Build GRCh38
rs749451392 Chr.22: 17738226-17738226 on Build GRCh38
rs547322681 Chr.22: 17738105-17738105 on Build GRCh38
rs778228540 Chr.22: 17738093-17738093 on Build GRCh38
rs949140935 Chr.22: 17738294-17738294 on Build GRCh38
rs374666749 Chr.22: 17738016-17738016 on Build GRCh38
rs200732958 Chr.22: 17738131-17738131 on Build GRCh38
rs763268339 Chr.22: 17738114-17738114 on Build GRCh38
rs557252956 Chr.22: 17738322-17738324 on Build GRCh38
rs771733957 Chr.22: 17738115-17738115 on Build GRCh38
rs146712574 Chr.22: 17738038-17738038 on Build GRCh38
rs185077891 Chr.22: 17738091-17738091 on Build GRCh38
rs895511972 Chr.22: 17738189-17738189 on Build GRCh38
rs752039343 Chr.22: 17738169-17738169 on Build GRCh38
rs376912423 Chr.22: 17738082-17738082 on Build GRCh38
rs541378614 Chr.22: 17737979-17737979 on Build GRCh38
rs747537974 Chr.22: 17738029-17738029 on Build GRCh38
rs535614913 Chr.22: 17738348-17738348 on Build GRCh38
rs757219056 Chr.22: 17737971-17737971 on Build GRCh38
rs1001435797 Chr.22: 17737942-17737942 on Build GRCh38
rs753402580 Chr.22: 17738077-17738077 on Build GRCh38
rs61752236 Chr.22: 17738037-17738037 on Build GRCh38
rs1055698807 Chr.22: 17738188-17738188 on Build GRCh38
rs762626576 Chr.22: 17738161-17738163 on Build GRCh38
rs764000332 Chr.22: 17738279-17738283 on Build GRCh38
rs374084609 Chr.22: 17738218-17738218 on Build GRCh38
rs142609840 Chr.22: 17738225-17738225 on Build GRCh38
rs750323234 Chr.22: 17738052-17738052 on Build GRCh38
rs766369895 Chr.22: 17738124-17738124 on Build GRCh38
rs150956841 Chr.22: 17738146-17738146 on Build GRCh38
rs189096838 Chr.22: 17738256-17738256 on Build GRCh38
rs17853595 Chr.22: 17738107-17738107 on Build GRCh38
rs11538 Chr.22: 17738065-17738065 on Build GRCh38
rs1019413784 Chr.22: 17738104-17738104 on Build GRCh38
rs778362797 Chr.22: 17738085-17738085 on Build GRCh38
rs140738257 Chr.22: 17738149-17738149 on Build GRCh38
rs753778329 Chr.22: 17738127-17738127 on Build GRCh38
rs150084106 Chr.22: 17738150-17738150 on Build GRCh38
rs200264082 Chr.22: 17737982-17737982 on Build GRCh38
rs538615870 Chr.22: 17738046-17738046 on Build GRCh38
rs760394304 Chr.22: 17738245-17738245 on Build GRCh38
rs779322937 Chr.22: 17738003-17738003 on Build GRCh38
rs200477407 Chr.22: 17737980-17737980 on Build GRCh38
rs996868952 Chr.22: 17738325-17738325 on Build GRCh38
rs371538004 Chr.22: 17738224-17738224 on Build GRCh38
rs765851664 Chr.22: 17738074-17738074 on Build GRCh38
rs139212717 Chr.22: 17738045-17738045 on Build GRCh38
rs534705188 Chr.22: 17738321-17738321 on Build GRCh38
rs767697157 Chr.22: 17738014-17738015 on Build GRCh38
rs368555557 Chr.22: 17737966-17737966 on Build GRCh38
rs959646977 Chr.22: 17738195-17738195 on Build GRCh38
rs750389519 Chr.22: 17737976-17737976 on Build GRCh38
rs771091033 Chr.22: 17738036-17738036 on Build GRCh38
rs539560010 Chr.22: 17738185-17738185 on Build GRCh38
rs752404269 Chr.22: 17738088-17738088 on Build GRCh38
rs569163642 Chr.22: 17738347-17738347 on Build GRCh38
rs917666988 Chr.22: 17737984-17737984 on Build GRCh38
rs773426650 Chr.22: 17738055-17738055 on Build GRCh38
rs767295453 Chr.22: 17737975-17737975 on Build GRCh38
rs570510927 Chr.22: 17738246-17738246 on Build GRCh38
rs745881133 Chr.22: 17738102-17738102 on Build GRCh38
rs145415034 Chr.22: 17738187-17738187 on Build GRCh38
rs759487323 Chr.22: 17738112-17738123 on Build GRCh38
rs550005605 Chr.22: 17738018-17738018 on Build GRCh38
rs776498658 Chr.22: 17738264-17738264 on Build GRCh38
rs765288306 Chr.22: 17738124-17738125 on Build GRCh38
rs967098884 Chr.22: 17738160-17738160 on Build GRCh38
rs772986045 Chr.22: 17738239-17738239 on Build GRCh38
rs367829996 Chr.22: 17738050-17738050 on Build GRCh38
rs925525205 Chr.22: 17738194-17738194 on Build GRCh38

View Full Product Details