Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19179787-19180283 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLTCL1
Entrez Gene ID
8218
Gene Name
clathrin heavy chain like 1
Gene Aliases
CHC22, CLH22, CLTCL, CLTD
UniGene
Hs.368266
Gene Chromosome Location
Chr.22: 19179473-19291716 on Build GRCh38
Transcripts
NM_001835.3 NM_007098.3 XM_011530401.2 XM_017028953.1 XM_017028954.1 XM_017028955.1 XM_017028956.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4672959 c.4904-3_4904-2insT p.? Chr.22: 19180240-19180241 on Build GRCh38
4672960 c.4904-3_4904-2insT p.? Chr.22: 19180240-19180241 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs782245948 Chr.22: 19180175-19180175 on Build GRCh38
rs547970915 Chr.22: 19179933-19179933 on Build GRCh38
rs931917252 Chr.22: 19180110-19180110 on Build GRCh38
rs996093519 Chr.22: 19179907-19179907 on Build GRCh38
rs928871495 Chr.22: 19180015-19180015 on Build GRCh38
rs530915474 Chr.22: 19179884-19179884 on Build GRCh38
rs533581212 Chr.22: 19179882-19179882 on Build GRCh38
rs527240248 Chr.22: 19180040-19180040 on Build GRCh38
rs569749506 Chr.22: 19180217-19180217 on Build GRCh38
rs572424136 Chr.22: 19180249-19180249 on Build GRCh38
rs116580497 Chr.22: 19180048-19180048 on Build GRCh38
rs782131544 Chr.22: 19180213-19180213 on Build GRCh38
rs712958 Chr.22: 19179896-19179896 on Build GRCh38
rs541747624 Chr.22: 19179867-19179867 on Build GRCh38
rs782299325 Chr.22: 19180195-19180195 on Build GRCh38
rs999172184 Chr.22: 19179972-19179972 on Build GRCh38
rs890105377 Chr.22: 19179969-19179969 on Build GRCh38
rs782501774 Chr.22: 19179863-19179863 on Build GRCh38
rs928607920 Chr.22: 19179829-19179829 on Build GRCh38
rs372445075 Chr.22: 19180200-19180200 on Build GRCh38
rs377159014 Chr.22: 19180248-19180248 on Build GRCh38
rs200374285 Chr.22: 19180228-19180228 on Build GRCh38
rs568057925 Chr.22: 19179980-19179980 on Build GRCh38
rs1052129308 Chr.22: 19180126-19180126 on Build GRCh38
rs999602029 Chr.22: 19180154-19180154 on Build GRCh38
rs781864368 Chr.22: 19180212-19180212 on Build GRCh38
rs555970939 Chr.22: 19180137-19180137 on Build GRCh38
rs1048287927 Chr.22: 19179846-19179846 on Build GRCh38
rs782429860 Chr.22: 19180251-19180251 on Build GRCh38
rs782379270 Chr.22: 19180182-19180182 on Build GRCh38
rs957746672 Chr.22: 19179977-19179977 on Build GRCh38
rs990311222 Chr.22: 19179989-19179989 on Build GRCh38
rs5746675 Chr.22: 19179908-19179908 on Build GRCh38
rs980748124 Chr.22: 19180011-19180011 on Build GRCh38
rs782092200 Chr.22: 19180196-19180196 on Build GRCh38
rs563708523 Chr.22: 19179906-19179906 on Build GRCh38
rs970306794 Chr.22: 19179995-19179995 on Build GRCh38
rs893594861 Chr.22: 19180177-19180177 on Build GRCh38
rs782152433 Chr.22: 19180194-19180194 on Build GRCh38
rs942087276 Chr.22: 19180057-19180057 on Build GRCh38
rs781838996 Chr.22: 19180231-19180231 on Build GRCh38
rs886499880 Chr.22: 19179862-19179862 on Build GRCh38
rs377564347 Chr.22: 19180045-19180045 on Build GRCh38
rs539335242 Chr.22: 19180108-19180108 on Build GRCh38
rs938808794 Chr.22: 19179843-19179843 on Build GRCh38
rs1021744983 Chr.22: 19179991-19179991 on Build GRCh38
rs1037580513 Chr.22: 19180072-19180072 on Build GRCh38
rs1031187930 Chr.22: 19179975-19179975 on Build GRCh38
rs1044413940 Chr.22: 19179817-19179817 on Build GRCh38
rs782185297 Chr.22: 19180169-19180169 on Build GRCh38
rs782194359 Chr.22: 19180095-19180095 on Build GRCh38
rs782012995 Chr.22: 19180193-19180193 on Build GRCh38
rs738904 Chr.22: 19179872-19179872 on Build GRCh38

View Full Product Details