Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19479809-19480311 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC45
Entrez Gene ID
8318
Gene Name
cell division cycle 45
Gene Aliases
CDC45L, CDC45L2, PORC-PI-1
UniGene
Hs.474217
Gene Chromosome Location
Chr.22: 19479278-19520612 on Build GRCh38
Transcripts
NM_001178010.2 NM_001178011.2 NM_003504.4 XM_005261285.2 XM_005261286.3 XM_011530415.1 XM_011530416.1 XM_011530417.2 XM_011530418.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7226657 c.88C>T p.L30L Chr.22: 19480194-19480194 on Build GRCh38
1200382 c.30C>A p.F10L Chr.22: 19479998-19479998 on Build GRCh38
5393160 c.74C>T p.S25L Chr.22: 19480180-19480180 on Build GRCh38
478728 c.52A>T p.R18W Chr.22: 19480158-19480158 on Build GRCh38
5393158 c.45G>A p.Q15Q Chr.22: 19480013-19480013 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760466371 Chr.22: 19479946-19479946 on Build GRCh38
rs753376750 Chr.22: 19480211-19480211 on Build GRCh38
rs753248916 Chr.22: 19480240-19480240 on Build GRCh38
rs772082975 Chr.22: 19480034-19480034 on Build GRCh38
rs535155496 Chr.22: 19479859-19479859 on Build GRCh38
rs767186293 Chr.22: 19480173-19480173 on Build GRCh38
rs773221544 Chr.22: 19480269-19480269 on Build GRCh38
rs373203909 Chr.22: 19480153-19480153 on Build GRCh38
rs1009015762 Chr.22: 19479891-19479891 on Build GRCh38
rs376674660 Chr.22: 19479919-19479919 on Build GRCh38
rs768613260 Chr.22: 19480260-19480260 on Build GRCh38
rs766064032 Chr.22: 19480207-19480207 on Build GRCh38
rs908286214 Chr.22: 19479968-19479968 on Build GRCh38
rs774472467 Chr.22: 19479958-19479958 on Build GRCh38
rs535099250 Chr.22: 19479962-19479962 on Build GRCh38
rs775922222 Chr.22: 19479951-19479951 on Build GRCh38
rs373683620 Chr.22: 19480040-19480040 on Build GRCh38
rs372934193 Chr.22: 19479941-19479941 on Build GRCh38
rs550254398 Chr.22: 19479940-19479940 on Build GRCh38
rs759977295 Chr.22: 19479881-19479881 on Build GRCh38
rs776344629 Chr.22: 19480020-19480021 on Build GRCh38
rs773487667 Chr.22: 19480037-19480037 on Build GRCh38
rs1009560085 Chr.22: 19479923-19479923 on Build GRCh38
rs371100740 Chr.22: 19479985-19479985 on Build GRCh38
rs746071726 Chr.22: 19480259-19480259 on Build GRCh38
rs768661708 Chr.22: 19479954-19479954 on Build GRCh38
rs894503874 Chr.22: 19480091-19480091 on Build GRCh38
rs1037976328 Chr.22: 19479990-19479990 on Build GRCh38
rs369627026 Chr.22: 19479950-19479950 on Build GRCh38
rs549095792 Chr.22: 19480236-19480236 on Build GRCh38
rs754730066 Chr.22: 19479927-19479927 on Build GRCh38
rs13447183 Chr.22: 19479959-19479959 on Build GRCh38
rs375603502 Chr.22: 19480137-19480137 on Build GRCh38
rs769525949 Chr.22: 19480042-19480042 on Build GRCh38
rs781532842 Chr.22: 19480257-19480257 on Build GRCh38
rs967570274 Chr.22: 19479875-19479875 on Build GRCh38
rs767133327 Chr.22: 19480108-19480108 on Build GRCh38
rs923394428 Chr.22: 19479901-19479901 on Build GRCh38
rs755232389 Chr.22: 19480124-19480124 on Build GRCh38
rs772167949 Chr.22: 19480267-19480267 on Build GRCh38
rs1034012211 Chr.22: 19480208-19480208 on Build GRCh38
rs889327940 Chr.22: 19480077-19480077 on Build GRCh38
rs564311551 Chr.22: 19480027-19480027 on Build GRCh38
rs779407514 Chr.22: 19480128-19480128 on Build GRCh38
rs776646954 Chr.22: 19480152-19480152 on Build GRCh38
rs910956198 Chr.22: 19480202-19480202 on Build GRCh38
rs559726930 Chr.22: 19480286-19480286 on Build GRCh38
rs758924609 Chr.22: 19480241-19480241 on Build GRCh38
rs745505016 Chr.22: 19480002-19480002 on Build GRCh38
rs13447182 Chr.22: 19479939-19479939 on Build GRCh38
rs563826047 Chr.22: 19480163-19480163 on Build GRCh38
rs1022236977 Chr.22: 19479928-19479928 on Build GRCh38
rs746235990 Chr.22: 19479944-19479944 on Build GRCh38
rs770856992 Chr.22: 19480255-19480255 on Build GRCh38
rs548010676 Chr.22: 19479960-19479960 on Build GRCh38
rs777304758 Chr.22: 19479938-19479938 on Build GRCh38
rs377264229 Chr.22: 19479946-19479946 on Build GRCh38
rs756757376 Chr.22: 19479992-19479992 on Build GRCh38
rs755773508 Chr.22: 19480112-19480112 on Build GRCh38
rs375507586 Chr.22: 19480266-19480266 on Build GRCh38
rs199895996 Chr.22: 19480146-19480146 on Build GRCh38
rs530578674 Chr.22: 19479916-19479916 on Build GRCh38
rs751141981 Chr.22: 19479991-19479991 on Build GRCh38
rs539707484 Chr.22: 19479935-19479935 on Build GRCh38
rs944849953 Chr.22: 19480088-19480088 on Build GRCh38
rs781002768 Chr.22: 19479995-19479995 on Build GRCh38
rs370872536 Chr.22: 19480249-19480249 on Build GRCh38
rs777563505 Chr.22: 19480164-19480164 on Build GRCh38
rs769229053 Chr.22: 19480025-19480025 on Build GRCh38
rs772178559 Chr.22: 19480131-19480131 on Build GRCh38
rs199753286 Chr.22: 19479979-19479979 on Build GRCh38
rs749976287 Chr.22: 19480177-19480177 on Build GRCh38
rs1051107766 Chr.22: 19480073-19480073 on Build GRCh38
rs903777460 Chr.22: 19480162-19480162 on Build GRCh38
rs751481362 Chr.22: 19480058-19480058 on Build GRCh38
rs376431394 Chr.22: 19480246-19480246 on Build GRCh38
rs547595264 Chr.22: 19479883-19479883 on Build GRCh38
rs147730653 Chr.22: 19479987-19479987 on Build GRCh38
rs967970494 Chr.22: 19479931-19479931 on Build GRCh38
rs749801430 Chr.22: 19479952-19479952 on Build GRCh38
rs990438973 Chr.22: 19479852-19479852 on Build GRCh38
rs930874171 Chr.22: 19480030-19480030 on Build GRCh38
rs747115945 Chr.22: 19480254-19480254 on Build GRCh38
rs756505067 Chr.22: 19480008-19480008 on Build GRCh38
rs5748232 Chr.22: 19480129-19480129 on Build GRCh38
rs898416016 Chr.22: 19479956-19479956 on Build GRCh38
rs757912076 Chr.22: 19479936-19479936 on Build GRCh38
rs762612258 Chr.22: 19480057-19480057 on Build GRCh38
rs570684206 Chr.22: 19479917-19479917 on Build GRCh38
rs748231994 Chr.22: 19480033-19480033 on Build GRCh38
rs200419044 Chr.22: 19479966-19479966 on Build GRCh38
rs958934901 Chr.22: 19479848-19479848 on Build GRCh38
rs755431657 Chr.22: 19480225-19480225 on Build GRCh38
rs141041145 Chr.22: 19480009-19480009 on Build GRCh38
rs753657397 Chr.22: 19479925-19479925 on Build GRCh38
rs114125269 Chr.22: 19479837-19479837 on Build GRCh38
rs779528878 Chr.22: 19480120-19480120 on Build GRCh38
rs1046942470 Chr.22: 19480275-19480275 on Build GRCh38
rs4141527 Chr.22: 19480194-19480194 on Build GRCh38
rs577826035 Chr.22: 19480069-19480069 on Build GRCh38
rs566798249 Chr.22: 19480250-19480250 on Build GRCh38
rs770638350 Chr.22: 19479956-19479957 on Build GRCh38
rs369549893 Chr.22: 19480050-19480050 on Build GRCh38
rs747011825 Chr.22: 19480142-19480142 on Build GRCh38
rs995432452 Chr.22: 19479870-19479870 on Build GRCh38
rs750143896 Chr.22: 19480111-19480111 on Build GRCh38
rs773050738 Chr.22: 19479932-19479932 on Build GRCh38
rs759247064 Chr.22: 19480034-19480034 on Build GRCh38
rs929910263 Chr.22: 19480273-19480273 on Build GRCh38
rs767368308 Chr.22: 19480063-19480063 on Build GRCh38

View Full Product Details