Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19494407-19494913 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC45
Entrez Gene ID
8318
Gene Name
cell division cycle 45
Gene Aliases
CDC45L, CDC45L2, PORC-PI-1
UniGene
Hs.474217
Gene Chromosome Location
Chr.22: 19479278-19520612 on Build GRCh38
Transcripts
NM_001178010.2 NM_001178011.2 NM_003504.4 XM_005261285.2 XM_005261286.3 XM_011530415.1 XM_011530416.1 XM_011530417.2 XM_011530418.2 XM_017028966.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4594852 c.548C>G p.S183C Chr.22: 19494512-19494512 on Build GRCh38
6004798 c.601G>A p.G201S Chr.22: 19494565-19494565 on Build GRCh38
5587466 c.552G>A p.G184G Chr.22: 19494516-19494516 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs923448835 Chr.22: 19494615-19494615 on Build GRCh38
rs754023399 Chr.22: 19494582-19494582 on Build GRCh38
rs528856178 Chr.22: 19494540-19494540 on Build GRCh38
rs769467118 Chr.22: 19494491-19494491 on Build GRCh38
rs896636923 Chr.22: 19494827-19494827 on Build GRCh38
rs558030330 Chr.22: 19494433-19494433 on Build GRCh38
rs549677338 Chr.22: 19494458-19494458 on Build GRCh38
rs151216373 Chr.22: 19494512-19494512 on Build GRCh38
rs976566988 Chr.22: 19494772-19494772 on Build GRCh38
rs776116706 Chr.22: 19494790-19494790 on Build GRCh38
rs767213571 Chr.22: 19494560-19494560 on Build GRCh38
rs768766940 Chr.22: 19494439-19494439 on Build GRCh38
rs957257216 Chr.22: 19494662-19494662 on Build GRCh38
rs760373373 Chr.22: 19494564-19494564 on Build GRCh38
rs1001116214 Chr.22: 19494607-19494607 on Build GRCh38
rs555866595 Chr.22: 19494756-19494756 on Build GRCh38
rs1024861061 Chr.22: 19494797-19494797 on Build GRCh38
rs749427341 Chr.22: 19494501-19494501 on Build GRCh38
rs773246748 Chr.22: 19494472-19494472 on Build GRCh38
rs972482024 Chr.22: 19494525-19494525 on Build GRCh38
rs759430493 Chr.22: 19494479-19494479 on Build GRCh38
rs1012181282 Chr.22: 19494784-19494784 on Build GRCh38
rs779261693 Chr.22: 19494524-19494524 on Build GRCh38
rs368191577 Chr.22: 19494654-19494654 on Build GRCh38
rs751143985 Chr.22: 19494537-19494537 on Build GRCh38
rs943657814 Chr.22: 19494601-19494601 on Build GRCh38
rs988681042 Chr.22: 19494663-19494663 on Build GRCh38
rs571756921 Chr.22: 19494839-19494839 on Build GRCh38
rs761339731 Chr.22: 19494558-19494558 on Build GRCh38
rs984758251 Chr.22: 19494775-19494775 on Build GRCh38
rs1020518186 Chr.22: 19494703-19494703 on Build GRCh38
rs966182952 Chr.22: 19494720-19494720 on Build GRCh38
rs772315522 Chr.22: 19494460-19494460 on Build GRCh38
rs775533667 Chr.22: 19494510-19494510 on Build GRCh38
rs371650955 Chr.22: 19494883-19494883 on Build GRCh38
rs746012338 Chr.22: 19494541-19494541 on Build GRCh38
rs768509916 Chr.22: 19494866-19494866 on Build GRCh38
rs933450209 Chr.22: 19494627-19494627 on Build GRCh38
rs1039252777 Chr.22: 19494608-19494608 on Build GRCh38
rs545467018 Chr.22: 19494532-19494532 on Build GRCh38
rs778945194 Chr.22: 19494445-19494445 on Build GRCh38
rs909480953 Chr.22: 19494785-19494785 on Build GRCh38
rs756569539 Chr.22: 19494565-19494565 on Build GRCh38
rs1036560512 Chr.22: 19494824-19494824 on Build GRCh38
rs1005061205 Chr.22: 19494462-19494462 on Build GRCh38
rs548610021 Chr.22: 19494584-19494584 on Build GRCh38
rs371395209 Chr.22: 19494432-19494432 on Build GRCh38
rs1042466048 Chr.22: 19494638-19494638 on Build GRCh38
rs902626904 Chr.22: 19494681-19494681 on Build GRCh38
rs984220089 Chr.22: 19494562-19494562 on Build GRCh38
rs750033004 Chr.22: 19494561-19494561 on Build GRCh38
rs575613523 Chr.22: 19494873-19494873 on Build GRCh38
rs770750861 Chr.22: 19494639-19494640 on Build GRCh38
rs910868701 Chr.22: 19494593-19494593 on Build GRCh38
rs189875945 Chr.22: 19494758-19494758 on Build GRCh38
rs766622291 Chr.22: 19494630-19494630 on Build GRCh38
rs537700556 Chr.22: 19494861-19494861 on Build GRCh38
rs1000940285 Chr.22: 19494769-19494769 on Build GRCh38
rs748178855 Chr.22: 19494450-19494450 on Build GRCh38
rs764074237 Chr.22: 19494535-19494535 on Build GRCh38
rs13447214 Chr.22: 19494774-19494774 on Build GRCh38
rs969692772 Chr.22: 19494592-19494592 on Build GRCh38
rs1024006261 Chr.22: 19494574-19494574 on Build GRCh38
rs921968379 Chr.22: 19494774-19494775 on Build GRCh38
rs1054235757 Chr.22: 19494770-19494770 on Build GRCh38

View Full Product Details