Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19495896-19496401 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC45
Entrez Gene ID
8318
Gene Name
cell division cycle 45
Gene Aliases
CDC45L, CDC45L2, PORC-PI-1
UniGene
Hs.474217
Gene Chromosome Location
Chr.22: 19479278-19520612 on Build GRCh38
Transcripts
NM_001178010.2 NM_001178011.2 NM_003504.4 XM_005261285.2 XM_005261286.3 XM_011530415.1 XM_011530416.1 XM_011530417.2 XM_011530418.2 XM_017028966.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
725604 c.576A>G p.E192E Chr.22: 19496014-19496014 on Build GRCh38
5112467 c.660C>T p.Y220Y Chr.22: 19496002-19496002 on Build GRCh38
6624450 c.661G>A p.E221K Chr.22: 19496003-19496003 on Build GRCh38
6624451 c.565G>A p.E189K Chr.22: 19496003-19496003 on Build GRCh38
271157 c.564C>T p.Y188Y Chr.22: 19496002-19496002 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs941908878 Chr.22: 19496305-19496305 on Build GRCh38
rs766934025 Chr.22: 19495988-19495988 on Build GRCh38
rs958256270 Chr.22: 19496072-19496072 on Build GRCh38
rs749108557 Chr.22: 19496038-19496038 on Build GRCh38
rs1050815728 Chr.22: 19496339-19496339 on Build GRCh38
rs933805396 Chr.22: 19496309-19496309 on Build GRCh38
rs781497533 Chr.22: 19496034-19496034 on Build GRCh38
rs1037968285 Chr.22: 19496318-19496318 on Build GRCh38
rs116978927 Chr.22: 19496359-19496359 on Build GRCh38
rs141942404 Chr.22: 19496042-19496042 on Build GRCh38
rs752577743 Chr.22: 19496104-19496104 on Build GRCh38
rs906543038 Chr.22: 19496028-19496028 on Build GRCh38
rs969511120 Chr.22: 19496131-19496131 on Build GRCh38
rs2073759 Chr.22: 19496375-19496375 on Build GRCh38
rs764493642 Chr.22: 19496109-19496110 on Build GRCh38
rs765956682 Chr.22: 19496080-19496080 on Build GRCh38
rs546984033 Chr.22: 19496118-19496118 on Build GRCh38
rs759157568 Chr.22: 19496073-19496073 on Build GRCh38
rs761467311 Chr.22: 19495983-19495983 on Build GRCh38
rs367976893 Chr.22: 19496002-19496002 on Build GRCh38
rs199886373 Chr.22: 19495950-19495950 on Build GRCh38
rs928010217 Chr.22: 19496175-19496175 on Build GRCh38
rs977193187 Chr.22: 19496177-19496177 on Build GRCh38
rs371673144 Chr.22: 19496026-19496026 on Build GRCh38
rs777689461 Chr.22: 19496020-19496020 on Build GRCh38
rs958337561 Chr.22: 19496061-19496061 on Build GRCh38
rs951473019 Chr.22: 19496231-19496231 on Build GRCh38
rs921101588 Chr.22: 19496287-19496287 on Build GRCh38
rs1054166369 Chr.22: 19496363-19496363 on Build GRCh38
rs779356050 Chr.22: 19495937-19495937 on Build GRCh38
rs200771303 Chr.22: 19495949-19495949 on Build GRCh38
rs755505484 Chr.22: 19495936-19495936 on Build GRCh38
rs1023841506 Chr.22: 19496119-19496119 on Build GRCh38
rs186747024 Chr.22: 19496290-19496290 on Build GRCh38
rs897650071 Chr.22: 19496333-19496333 on Build GRCh38
rs772778916 Chr.22: 19496135-19496135 on Build GRCh38
rs929421702 Chr.22: 19496357-19496357 on Build GRCh38
rs982828507 Chr.22: 19496257-19496257 on Build GRCh38
rs567200722 Chr.22: 19496256-19496256 on Build GRCh38
rs201610895 Chr.22: 19496070-19496070 on Build GRCh38
rs753193801 Chr.22: 19496010-19496010 on Build GRCh38
rs375553186 Chr.22: 19495932-19495932 on Build GRCh38
rs370140302 Chr.22: 19495969-19495969 on Build GRCh38
rs764941393 Chr.22: 19495998-19495998 on Build GRCh38
rs993584184 Chr.22: 19496009-19496009 on Build GRCh38
rs534331332 Chr.22: 19496092-19496092 on Build GRCh38
rs111403195 Chr.22: 19495977-19495977 on Build GRCh38
rs373809427 Chr.22: 19495978-19495978 on Build GRCh38
rs373780952 Chr.22: 19496071-19496071 on Build GRCh38
rs748234011 Chr.22: 19495939-19495939 on Build GRCh38
rs566899785 Chr.22: 19496003-19496003 on Build GRCh38
rs537963411 Chr.22: 19496346-19496346 on Build GRCh38
rs967564684 Chr.22: 19496169-19496169 on Build GRCh38
rs758542881 Chr.22: 19496014-19496014 on Build GRCh38
rs148779081 Chr.22: 19496029-19496029 on Build GRCh38
rs760225957 Chr.22: 19495990-19495990 on Build GRCh38
rs568990077 Chr.22: 19496321-19496321 on Build GRCh38
rs772959198 Chr.22: 19495989-19495989 on Build GRCh38
rs909872586 Chr.22: 19496260-19496260 on Build GRCh38
rs758391220 Chr.22: 19495940-19495940 on Build GRCh38
rs776212438 Chr.22: 19496081-19496081 on Build GRCh38
rs768105134 Chr.22: 19496276-19496276 on Build GRCh38
rs376655788 Chr.22: 19496075-19496075 on Build GRCh38
rs774497688 Chr.22: 19496046-19496046 on Build GRCh38
rs762440742 Chr.22: 19496252-19496252 on Build GRCh38
rs386819699 Chr.22: 19496375-19496376 on Build GRCh38
rs941322142 Chr.22: 19496261-19496261 on Build GRCh38
rs776343481 Chr.22: 19495994-19495995 on Build GRCh38
rs778122059 Chr.22: 19495944-19495944 on Build GRCh38
rs922975510 Chr.22: 19496206-19496206 on Build GRCh38
rs111835400 Chr.22: 19496376-19496376 on Build GRCh38
rs145918922 Chr.22: 19496295-19496295 on Build GRCh38
rs769302137 Chr.22: 19496094-19496094 on Build GRCh38

View Full Product Details