Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19523711-19524198 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN5
Entrez Gene ID
7122
Gene Name
claudin 5
Gene Aliases
AWAL, BEC1, CPETRL1, TMVCF
UniGene
Hs.505337
Gene Chromosome Location
Chr.22: 19523024-19527545 on Build GRCh38
Transcripts
NM_001130861.1 NM_003277.3 XM_017028929.1 XM_017028930.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5707594 c.241C>T p.R81W Chr.22: 19524015-19524015 on Build GRCh38
6569282 c.795C>G p.L265L Chr.22: 19523716-19523716 on Build GRCh38
5393165 c.158C>T p.S53L Chr.22: 19524098-19524098 on Build GRCh38
4672697 c.240G>A p.A80A Chr.22: 19524016-19524016 on Build GRCh38
6207197 c.491C>T p.A164V Chr.22: 19524020-19524020 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771193724 Chr.22: 19523859-19523859 on Build GRCh38
rs533261799 Chr.22: 19524127-19524127 on Build GRCh38
rs777716097 Chr.22: 19523856-19523856 on Build GRCh38
rs774155075 Chr.22: 19523758-19523758 on Build GRCh38
rs373454282 Chr.22: 19524028-19524028 on Build GRCh38
rs771950465 Chr.22: 19524003-19524003 on Build GRCh38
rs371581275 Chr.22: 19523945-19523945 on Build GRCh38
rs200303099 Chr.22: 19524016-19524016 on Build GRCh38
rs769938010 Chr.22: 19523993-19523993 on Build GRCh38
rs748585889 Chr.22: 19523839-19523839 on Build GRCh38
rs112469158 Chr.22: 19523845-19523845 on Build GRCh38
rs753536874 Chr.22: 19523951-19523951 on Build GRCh38
rs749882968 Chr.22: 19523770-19523770 on Build GRCh38
rs758858377 Chr.22: 19523959-19523959 on Build GRCh38
rs150374465 Chr.22: 19524170-19524170 on Build GRCh38
rs369164427 Chr.22: 19523860-19523860 on Build GRCh38
rs758878550 Chr.22: 19523850-19523850 on Build GRCh38
rs143559030 Chr.22: 19524027-19524027 on Build GRCh38
rs930257038 Chr.22: 19524172-19524172 on Build GRCh38
rs151223315 Chr.22: 19524136-19524136 on Build GRCh38
rs766950337 Chr.22: 19523766-19523766 on Build GRCh38
rs868539380 Chr.22: 19523919-19523919 on Build GRCh38
rs764949247 Chr.22: 19523868-19523868 on Build GRCh38
rs747206310 Chr.22: 19523732-19523732 on Build GRCh38
rs775289576 Chr.22: 19523740-19523740 on Build GRCh38
rs781455520 Chr.22: 19523981-19523981 on Build GRCh38
rs888601738 Chr.22: 19523908-19523908 on Build GRCh38
rs757328212 Chr.22: 19523980-19523980 on Build GRCh38
rs750464178 Chr.22: 19524059-19524059 on Build GRCh38
rs770989138 Chr.22: 19523896-19523896 on Build GRCh38
rs990687823 Chr.22: 19523791-19523791 on Build GRCh38
rs199561265 Chr.22: 19524161-19524161 on Build GRCh38
rs34379045 Chr.22: 19523953-19523953 on Build GRCh38
rs772894288 Chr.22: 19523921-19523921 on Build GRCh38
rs11551255 Chr.22: 19523929-19523929 on Build GRCh38
rs776219658 Chr.22: 19524019-19524019 on Build GRCh38
rs764578402 Chr.22: 19524036-19524036 on Build GRCh38
rs199779842 Chr.22: 19524049-19524049 on Build GRCh38
rs756314663 Chr.22: 19524064-19524064 on Build GRCh38
rs777629886 Chr.22: 19524130-19524130 on Build GRCh38
rs1025878210 Chr.22: 19524035-19524035 on Build GRCh38
rs369422141 Chr.22: 19523958-19523958 on Build GRCh38
rs199637865 Chr.22: 19523750-19523750 on Build GRCh38
rs761662164 Chr.22: 19523920-19523920 on Build GRCh38
rs140361640 Chr.22: 19524157-19524157 on Build GRCh38
rs768431057 Chr.22: 19523871-19523871 on Build GRCh38
rs1018313006 Chr.22: 19523928-19523928 on Build GRCh38
rs576749153 Chr.22: 19523956-19523956 on Build GRCh38
rs533778709 Chr.22: 19523743-19523743 on Build GRCh38
rs774138023 Chr.22: 19523886-19523886 on Build GRCh38
rs1053613 Chr.22: 19524109-19524109 on Build GRCh38
rs754302002 Chr.22: 19523796-19523796 on Build GRCh38
rs759264044 Chr.22: 19524021-19524021 on Build GRCh38
rs780302726 Chr.22: 19524079-19524079 on Build GRCh38
rs369726801 Chr.22: 19524022-19524022 on Build GRCh38
rs746103657 Chr.22: 19523992-19523992 on Build GRCh38
rs376454295 Chr.22: 19523834-19523834 on Build GRCh38
rs771895657 Chr.22: 19524128-19524128 on Build GRCh38
rs375234645 Chr.22: 19524007-19524007 on Build GRCh38
rs772621818 Chr.22: 19523925-19523925 on Build GRCh38
rs760239571 Chr.22: 19523934-19523934 on Build GRCh38
rs755864909 Chr.22: 19523772-19523772 on Build GRCh38
rs770551409 Chr.22: 19524018-19524018 on Build GRCh38
rs752051241 Chr.22: 19524043-19524043 on Build GRCh38
rs766079245 Chr.22: 19523773-19523773 on Build GRCh38
rs747068763 Chr.22: 19523857-19523857 on Build GRCh38
rs1045780776 Chr.22: 19524060-19524060 on Build GRCh38
rs751634461 Chr.22: 19523978-19523978 on Build GRCh38
rs749152398 Chr.22: 19523862-19523862 on Build GRCh38
rs778324400 Chr.22: 19523976-19523976 on Build GRCh38
rs1029596000 Chr.22: 19523974-19523974 on Build GRCh38
rs372805287 Chr.22: 19523807-19523807 on Build GRCh38
rs568221772 Chr.22: 19523742-19523742 on Build GRCh38
rs879930785 Chr.22: 19523923-19523923 on Build GRCh38
rs866581567 Chr.22: 19523799-19523799 on Build GRCh38
rs368493240 Chr.22: 19523733-19523733 on Build GRCh38
rs759065120 Chr.22: 19524158-19524158 on Build GRCh38
rs201059401 Chr.22: 19523997-19523997 on Build GRCh38
rs778606058 Chr.22: 19523995-19523995 on Build GRCh38

View Full Product Details