Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19801911-19802418 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNB1L
Entrez Gene ID
54584
Gene Name
G protein subunit beta 1 like
Gene Aliases
DGCRK3, FKSG1, GY2, WDR14, WDVCF
UniGene
Hs.105642
Gene Chromosome Location
Chr.22: 19788411-19854939 on Build GRCh38
Transcripts
NM_053004.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
341879 c.587C>T p.S196F Chr.22: 19802146-19802146 on Build GRCh38
3424038 c.550G>A p.G184S Chr.22: 19802183-19802183 on Build GRCh38
6395946 c.721C>T p.Q241* Chr.22: 19802012-19802012 on Build GRCh38
1032140 c.592C>T p.Q198* Chr.22: 19802141-19802141 on Build GRCh38
6410104 c.717G>A p.W239* Chr.22: 19802016-19802016 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs566852346 Chr.22: 19802340-19802340 on Build GRCh38
rs368405667 Chr.22: 19802347-19802347 on Build GRCh38
rs776926205 Chr.22: 19802024-19802024 on Build GRCh38
rs372247382 Chr.22: 19802135-19802135 on Build GRCh38
rs768609078 Chr.22: 19802114-19802114 on Build GRCh38
rs564960241 Chr.22: 19802216-19802216 on Build GRCh38
rs373727341 Chr.22: 19802371-19802371 on Build GRCh38
rs376821599 Chr.22: 19802167-19802167 on Build GRCh38
rs778831517 Chr.22: 19802193-19802193 on Build GRCh38
rs779421335 Chr.22: 19801988-19801988 on Build GRCh38
rs767099084 Chr.22: 19802049-19802049 on Build GRCh38
rs771906189 Chr.22: 19802343-19802343 on Build GRCh38
rs370517613 Chr.22: 19802080-19802080 on Build GRCh38
rs566824805 Chr.22: 19802067-19802067 on Build GRCh38
rs762384270 Chr.22: 19802243-19802243 on Build GRCh38
rs759926151 Chr.22: 19802065-19802065 on Build GRCh38
rs777975099 Chr.22: 19802097-19802097 on Build GRCh38
rs777693457 Chr.22: 19802015-19802015 on Build GRCh38
rs994463734 Chr.22: 19802385-19802385 on Build GRCh38
rs747052578 Chr.22: 19802088-19802099 on Build GRCh38
rs550410178 Chr.22: 19802234-19802234 on Build GRCh38
rs1008941921 Chr.22: 19802159-19802159 on Build GRCh38
rs770924275 Chr.22: 19802131-19802131 on Build GRCh38
rs376426350 Chr.22: 19802052-19802052 on Build GRCh38
rs899094510 Chr.22: 19802351-19802351 on Build GRCh38
rs933416301 Chr.22: 19802278-19802278 on Build GRCh38
rs746935334 Chr.22: 19802206-19802206 on Build GRCh38
rs898430475 Chr.22: 19802377-19802377 on Build GRCh38
rs200059079 Chr.22: 19802104-19802104 on Build GRCh38
rs374782970 Chr.22: 19801981-19801981 on Build GRCh38
rs767760872 Chr.22: 19802255-19802255 on Build GRCh38
rs758819568 Chr.22: 19802093-19802093 on Build GRCh38
rs748171822 Chr.22: 19802123-19802123 on Build GRCh38
rs766895083 Chr.22: 19801955-19801955 on Build GRCh38
rs907604710 Chr.22: 19802330-19802330 on Build GRCh38
rs755280805 Chr.22: 19801985-19801985 on Build GRCh38
rs530248368 Chr.22: 19802273-19802273 on Build GRCh38
rs1010230380 Chr.22: 19801976-19801976 on Build GRCh38
rs547245567 Chr.22: 19802274-19802274 on Build GRCh38
rs753443329 Chr.22: 19802063-19802063 on Build GRCh38
rs752054535 Chr.22: 19802169-19802169 on Build GRCh38
rs201659733 Chr.22: 19802051-19802051 on Build GRCh38
rs979748061 Chr.22: 19802238-19802238 on Build GRCh38
rs139155499 Chr.22: 19802098-19802098 on Build GRCh38
rs369472841 Chr.22: 19802129-19802130 on Build GRCh38
rs5748437 Chr.22: 19801963-19801963 on Build GRCh38
rs538631985 Chr.22: 19802341-19802341 on Build GRCh38
rs2073770 Chr.22: 19802018-19802018 on Build GRCh38
rs972428672 Chr.22: 19802073-19802073 on Build GRCh38
rs572852392 Chr.22: 19802201-19802201 on Build GRCh38
rs5748440 Chr.22: 19802061-19802061 on Build GRCh38
rs748616036 Chr.22: 19801992-19801992 on Build GRCh38
rs771053171 Chr.22: 19802021-19802021 on Build GRCh38
rs561494727 Chr.22: 19802272-19802272 on Build GRCh38
rs758548638 Chr.22: 19801993-19801993 on Build GRCh38
rs201224032 Chr.22: 19802200-19802200 on Build GRCh38
rs1050499096 Chr.22: 19802301-19802301 on Build GRCh38
rs771911578 Chr.22: 19802125-19802125 on Build GRCh38
rs778824018 Chr.22: 19802121-19802121 on Build GRCh38
rs753000516 Chr.22: 19802151-19802151 on Build GRCh38
rs899063242 Chr.22: 19802099-19802099 on Build GRCh38
rs775257953 Chr.22: 19802370-19802370 on Build GRCh38
rs765557200 Chr.22: 19802072-19802072 on Build GRCh38
rs569937118 Chr.22: 19802033-19802033 on Build GRCh38
rs375516094 Chr.22: 19802221-19802221 on Build GRCh38
rs749106756 Chr.22: 19802040-19802040 on Build GRCh38
rs774458770 Chr.22: 19802044-19802044 on Build GRCh38
rs545054042 Chr.22: 19802214-19802214 on Build GRCh38
rs533256667 Chr.22: 19801952-19801952 on Build GRCh38
rs766534417 Chr.22: 19802268-19802268 on Build GRCh38
rs756207620 Chr.22: 19802267-19802267 on Build GRCh38
rs147844556 Chr.22: 19802041-19802041 on Build GRCh38
rs901814437 Chr.22: 19802246-19802246 on Build GRCh38
rs538670673 Chr.22: 19802092-19802092 on Build GRCh38
rs752833737 Chr.22: 19801948-19801948 on Build GRCh38
rs754210431 Chr.22: 19801983-19801983 on Build GRCh38
rs779281672 Chr.22: 19802285-19802285 on Build GRCh38
rs200780846 Chr.22: 19802126-19802126 on Build GRCh38
rs1050417575 Chr.22: 19802368-19802368 on Build GRCh38
rs573762517 Chr.22: 19802177-19802177 on Build GRCh38
rs5748439 Chr.22: 19802031-19802031 on Build GRCh38
rs865899122 Chr.22: 19802026-19802026 on Build GRCh38
rs757443043 Chr.22: 19802172-19802172 on Build GRCh38
rs750524013 Chr.22: 19802263-19802263 on Build GRCh38
rs996060622 Chr.22: 19802367-19802367 on Build GRCh38
rs558606059 Chr.22: 19802102-19802102 on Build GRCh38
rs781585413 Chr.22: 19802103-19802103 on Build GRCh38
rs201962212 Chr.22: 19802184-19802184 on Build GRCh38
rs926942416 Chr.22: 19802244-19802244 on Build GRCh38
rs770713977 Chr.22: 19802183-19802183 on Build GRCh38
rs971018279 Chr.22: 19802394-19802394 on Build GRCh38
rs764479352 Chr.22: 19802224-19802224 on Build GRCh38
rs766799639 Chr.22: 19801982-19801982 on Build GRCh38
rs776306803 Chr.22: 19802141-19802141 on Build GRCh38
rs1025294699 Chr.22: 19802387-19802387 on Build GRCh38
rs775148749 Chr.22: 19802220-19802220 on Build GRCh38
rs763310294 Chr.22: 19802161-19802161 on Build GRCh38
rs750126240 Chr.22: 19801959-19801959 on Build GRCh38
rs5748436 Chr.22: 19801950-19801950 on Build GRCh38
rs760127944 Chr.22: 19802132-19802132 on Build GRCh38
rs879229900 Chr.22: 19802136-19802138 on Build GRCh38
rs574576970 Chr.22: 19802050-19802050 on Build GRCh38
rs927489306 Chr.22: 19802197-19802197 on Build GRCh38
rs5748441 Chr.22: 19802189-19802189 on Build GRCh38
rs893165025 Chr.22: 19801958-19801958 on Build GRCh38
rs761621740 Chr.22: 19802046-19802046 on Build GRCh38
rs777605606 Chr.22: 19802203-19802203 on Build GRCh38
rs374236553 Chr.22: 19802150-19802150 on Build GRCh38
rs775567324 Chr.22: 19802228-19802228 on Build GRCh38

View Full Product Details