Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19962443-19962950 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COMT
Entrez Gene ID
1312
Gene Name
catechol-O-methyltransferase
Gene Aliases
HEL-S-98n
UniGene
Hs.370408
Gene Chromosome Location
Chr.22: 19941740-19969975 on Build GRCh38
Transcripts
NM_000754.3 NM_001135161.1 NM_001135162.1 NM_007310.2 XM_011529886.1 XM_017028594.1 XM_017028595.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4596424 c.115A>G p.N39D Chr.22: 19962641-19962641 on Build GRCh38
3552221 c.177C>T p.I59I Chr.22: 19962703-19962703 on Build GRCh38
4137325 c.109G>T p.G37C Chr.22: 19962635-19962635 on Build GRCh38
310281 c.172C>T p.R58C Chr.22: 19962698-19962698 on Build GRCh38
6354285 c.214G>T p.A72S Chr.22: 19962740-19962740 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373611092 Chr.22: 19962794-19962794 on Build GRCh38
rs781461677 Chr.22: 19962587-19962587 on Build GRCh38
rs755825980 Chr.22: 19962799-19962799 on Build GRCh38
rs764392202 Chr.22: 19962565-19962565 on Build GRCh38
rs34819182 Chr.22: 19962868-19962868 on Build GRCh38
rs905166477 Chr.22: 19962730-19962730 on Build GRCh38
rs377353154 Chr.22: 19962499-19962499 on Build GRCh38
rs772880300 Chr.22: 19962525-19962525 on Build GRCh38
rs552979711 Chr.22: 19962775-19962775 on Build GRCh38
rs776094728 Chr.22: 19962642-19962642 on Build GRCh38
rs745542046 Chr.22: 19962733-19962733 on Build GRCh38
rs747016982 Chr.22: 19962780-19962782 on Build GRCh38
rs759181324 Chr.22: 19962533-19962533 on Build GRCh38
rs781266226 Chr.22: 19962503-19962503 on Build GRCh38
rs999200429 Chr.22: 19962749-19962749 on Build GRCh38
rs75012854 Chr.22: 19962641-19962641 on Build GRCh38
rs199755426 Chr.22: 19962517-19962517 on Build GRCh38
rs752620960 Chr.22: 19962701-19962701 on Build GRCh38
rs768578574 Chr.22: 19962505-19962505 on Build GRCh38
rs377574270 Chr.22: 19962846-19962846 on Build GRCh38
rs3218737 Chr.22: 19962824-19962824 on Build GRCh38
rs368616155 Chr.22: 19962532-19962532 on Build GRCh38
rs774847933 Chr.22: 19962797-19962797 on Build GRCh38
rs530456300 Chr.22: 19962899-19962899 on Build GRCh38
rs740602 Chr.22: 19962745-19962745 on Build GRCh38
rs777507390 Chr.22: 19962623-19962624 on Build GRCh38
rs761093019 Chr.22: 19962827-19962827 on Build GRCh38
rs561449386 Chr.22: 19962886-19962886 on Build GRCh38
rs4633 Chr.22: 19962712-19962712 on Build GRCh38
rs760899191 Chr.22: 19962753-19962753 on Build GRCh38
rs528311784 Chr.22: 19962807-19962807 on Build GRCh38
rs918618565 Chr.22: 19962625-19962625 on Build GRCh38
rs781018433 Chr.22: 19962856-19962857 on Build GRCh38
rs771850671 Chr.22: 19962629-19962629 on Build GRCh38
rs998414059 Chr.22: 19962475-19962475 on Build GRCh38
rs776957402 Chr.22: 19962839-19962839 on Build GRCh38
rs774277271 Chr.22: 19962720-19962720 on Build GRCh38
rs750490382 Chr.22: 19962599-19962599 on Build GRCh38
rs758271838 Chr.22: 19962704-19962704 on Build GRCh38
rs746764639 Chr.22: 19962714-19962714 on Build GRCh38
rs138628382 Chr.22: 19962620-19962620 on Build GRCh38
rs547048962 Chr.22: 19962577-19962577 on Build GRCh38
rs139227383 Chr.22: 19962614-19962614 on Build GRCh38
rs1033358503 Chr.22: 19962504-19962504 on Build GRCh38
rs777441922 Chr.22: 19962632-19962632 on Build GRCh38
rs773373201 Chr.22: 19962750-19962750 on Build GRCh38
rs765914859 Chr.22: 19962484-19962484 on Build GRCh38
rs11544670 Chr.22: 19962553-19962553 on Build GRCh38
rs753820115 Chr.22: 19962560-19962561 on Build GRCh38
rs746604009 Chr.22: 19962634-19962634 on Build GRCh38
rs72563160 Chr.22: 19962497-19962497 on Build GRCh38
rs13306281 Chr.22: 19962800-19962800 on Build GRCh38
rs921623583 Chr.22: 19962579-19962587 on Build GRCh38
rs544846648 Chr.22: 19962832-19962832 on Build GRCh38
rs375558228 Chr.22: 19962727-19962727 on Build GRCh38
rs376114841 Chr.22: 19962518-19962518 on Build GRCh38
rs749158838 Chr.22: 19962504-19962504 on Build GRCh38
rs778881400 Chr.22: 19962509-19962509 on Build GRCh38
rs767705110 Chr.22: 19962676-19962676 on Build GRCh38
rs757427295 Chr.22: 19962585-19962585 on Build GRCh38
rs6270 Chr.22: 19962627-19962627 on Build GRCh38
rs559479480 Chr.22: 19962584-19962584 on Build GRCh38
rs768254194 Chr.22: 19962805-19962805 on Build GRCh38
rs776476521 Chr.22: 19962637-19962637 on Build GRCh38
rs765385973 Chr.22: 19962842-19962842 on Build GRCh38
rs191204865 Chr.22: 19962787-19962787 on Build GRCh38
rs386819748 Chr.22: 19962855-19962857 on Build GRCh38
rs74745580 Chr.22: 19962643-19962643 on Build GRCh38
rs180991698 Chr.22: 19962550-19962550 on Build GRCh38
rs148620887 Chr.22: 19962773-19962773 on Build GRCh38
rs761850855 Chr.22: 19962847-19962847 on Build GRCh38
rs2239393 Chr.22: 19962905-19962905 on Build GRCh38
rs766377984 Chr.22: 19962761-19962761 on Build GRCh38
rs753856814 Chr.22: 19962762-19962762 on Build GRCh38
rs76384862 Chr.22: 19962739-19962739 on Build GRCh38
rs984098043 Chr.22: 19962644-19962644 on Build GRCh38
rs759091417 Chr.22: 19962639-19962639 on Build GRCh38
rs901434028 Chr.22: 19962474-19962474 on Build GRCh38
rs778030113 Chr.22: 19962498-19962498 on Build GRCh38
rs6267 Chr.22: 19962740-19962740 on Build GRCh38
rs773569271 Chr.22: 19962826-19962826 on Build GRCh38
rs373404261 Chr.22: 19962825-19962825 on Build GRCh38
rs762297267 Chr.22: 19962663-19962663 on Build GRCh38
rs756145905 Chr.22: 19962695-19962695 on Build GRCh38
rs569294134 Chr.22: 19962678-19962678 on Build GRCh38
rs757163626 Chr.22: 19962726-19962726 on Build GRCh38
rs745940378 Chr.22: 19962898-19962898 on Build GRCh38
rs551312068 Chr.22: 19962531-19962531 on Build GRCh38
rs970645223 Chr.22: 19962594-19962594 on Build GRCh38
rs750684179 Chr.22: 19962682-19962682 on Build GRCh38
rs753104108 Chr.22: 19962487-19962487 on Build GRCh38
rs748066830 Chr.22: 19962621-19962621 on Build GRCh38
rs770700364 Chr.22: 19962635-19962635 on Build GRCh38
rs149909767 Chr.22: 19962734-19962734 on Build GRCh38
rs753302283 Chr.22: 19962546-19962546 on Build GRCh38
rs750232064 Chr.22: 19962858-19962858 on Build GRCh38
rs759968047 Chr.22: 19962482-19962482 on Build GRCh38
rs779175593 Chr.22: 19962883-19962883 on Build GRCh38
rs755100597 Chr.22: 19962580-19962581 on Build GRCh38
rs767632109 Chr.22: 19962849-19962849 on Build GRCh38
rs760266742 Chr.22: 19962527-19962527 on Build GRCh38
rs1017155239 Chr.22: 19962514-19962514 on Build GRCh38
rs76452330 Chr.22: 19962806-19962806 on Build GRCh38
rs757073571 Chr.22: 19962781-19962781 on Build GRCh38
rs757034165 Chr.22: 19962784-19962785 on Build GRCh38
rs931653147 Chr.22: 19962619-19962619 on Build GRCh38
rs972188532 Chr.22: 19962929-19962929 on Build GRCh38
rs199690157 Chr.22: 19962776-19962776 on Build GRCh38
rs766469681 Chr.22: 19962699-19962699 on Build GRCh38
rs561536243 Chr.22: 19962541-19962541 on Build GRCh38
rs756235784 Chr.22: 19962602-19962602 on Build GRCh38

View Full Product Details