Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 20091777-20092275 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGCR8
Entrez Gene ID
54487
Gene Name
DGCR8 microprocessor complex subunit
Gene Aliases
C22orf12, DGCRK6, Gy1, pasha
UniGene
Hs.643452
Gene Chromosome Location
Chr.22: 20080232-20111877 on Build GRCh38
Transcripts
NM_001190326.1 NM_022720.6 XM_006724268.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3371859 c.1552G>A p.E518K Chr.22: 20091916-20091916 on Build GRCh38
4102654 c.1579C>T p.R527C Chr.22: 20091943-20091943 on Build GRCh38
4677165 c.1599T>C p.F533F Chr.22: 20091963-20091963 on Build GRCh38
1203436 c.1596C>A p.F532L Chr.22: 20091960-20091960 on Build GRCh38
1203437 c.1576G>A p.V526I Chr.22: 20091940-20091940 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs989829546 Chr.22: 20092139-20092139 on Build GRCh38
rs749186827 Chr.22: 20091870-20091870 on Build GRCh38
rs1022262348 Chr.22: 20092102-20092102 on Build GRCh38
rs771156181 Chr.22: 20091853-20091853 on Build GRCh38
rs757717070 Chr.22: 20092051-20092051 on Build GRCh38
rs771572387 Chr.22: 20091871-20091872 on Build GRCh38
rs1047636663 Chr.22: 20092002-20092002 on Build GRCh38
rs372312511 Chr.22: 20092052-20092052 on Build GRCh38
rs995069883 Chr.22: 20092029-20092029 on Build GRCh38
rs144592551 Chr.22: 20091897-20091897 on Build GRCh38
rs753002278 Chr.22: 20091950-20091950 on Build GRCh38
rs12628703 Chr.22: 20092028-20092028 on Build GRCh38
rs775526389 Chr.22: 20091865-20091865 on Build GRCh38
rs371062233 Chr.22: 20091963-20091963 on Build GRCh38
rs79279088 Chr.22: 20092059-20092059 on Build GRCh38
rs370653105 Chr.22: 20091828-20091828 on Build GRCh38
rs924355806 Chr.22: 20091809-20091809 on Build GRCh38
rs957105029 Chr.22: 20092137-20092137 on Build GRCh38
rs942123457 Chr.22: 20092047-20092047 on Build GRCh38
rs761731183 Chr.22: 20091891-20091891 on Build GRCh38
rs928567664 Chr.22: 20092225-20092225 on Build GRCh38
rs765008728 Chr.22: 20092011-20092011 on Build GRCh38
rs201043772 Chr.22: 20091985-20091985 on Build GRCh38
rs999644472 Chr.22: 20092097-20092097 on Build GRCh38
rs760132322 Chr.22: 20091906-20091906 on Build GRCh38
rs369009223 Chr.22: 20092020-20092020 on Build GRCh38
rs538045714 Chr.22: 20092239-20092239 on Build GRCh38
rs749814406 Chr.22: 20091948-20091948 on Build GRCh38
rs758875827 Chr.22: 20091960-20091960 on Build GRCh38
rs887845314 Chr.22: 20092006-20092006 on Build GRCh38
rs867118698 Chr.22: 20092033-20092033 on Build GRCh38
rs150083562 Chr.22: 20092172-20092172 on Build GRCh38
rs765939851 Chr.22: 20092020-20092021 on Build GRCh38
rs773999657 Chr.22: 20091888-20091888 on Build GRCh38
rs11089328 Chr.22: 20092080-20092080 on Build GRCh38
rs534655918 Chr.22: 20091915-20091915 on Build GRCh38
rs1038675097 Chr.22: 20091977-20091977 on Build GRCh38
rs747189726 Chr.22: 20092238-20092238 on Build GRCh38
rs768440089 Chr.22: 20091883-20091883 on Build GRCh38
rs528434922 Chr.22: 20092087-20092087 on Build GRCh38
rs372197987 Chr.22: 20092007-20092007 on Build GRCh38
rs909020038 Chr.22: 20092253-20092253 on Build GRCh38
rs929148321 Chr.22: 20091921-20091921 on Build GRCh38
rs551944306 Chr.22: 20092223-20092223 on Build GRCh38
rs1032494959 Chr.22: 20092110-20092110 on Build GRCh38
rs1045613329 Chr.22: 20091909-20091909 on Build GRCh38
rs969717112 Chr.22: 20092181-20092181 on Build GRCh38
rs77524092 Chr.22: 20092249-20092249 on Build GRCh38
rs201890587 Chr.22: 20091831-20091831 on Build GRCh38
rs532539327 Chr.22: 20092031-20092031 on Build GRCh38
rs780692891 Chr.22: 20091900-20091900 on Build GRCh38
rs530685579 Chr.22: 20092074-20092074 on Build GRCh38
rs755455097 Chr.22: 20091918-20091918 on Build GRCh38
rs747263923 Chr.22: 20091994-20091994 on Build GRCh38
rs112629076 Chr.22: 20092165-20092165 on Build GRCh38
rs765909332 Chr.22: 20091913-20091913 on Build GRCh38
rs550599362 Chr.22: 20092209-20092209 on Build GRCh38
rs746208324 Chr.22: 20092073-20092073 on Build GRCh38
rs373148087 Chr.22: 20091842-20091842 on Build GRCh38
rs529745584 Chr.22: 20092135-20092135 on Build GRCh38
rs569147373 Chr.22: 20092229-20092229 on Build GRCh38
rs772862570 Chr.22: 20091872-20091873 on Build GRCh38

View Full Product Details