Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 20314466-20314973 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGCR6L
Entrez Gene ID
85359
Gene Name
DiGeorge syndrome critical region gene 6-like
Gene Aliases
DGCR6
UniGene
Hs.410965
Gene Chromosome Location
Chr.22: 20314238-20320105 on Build GRCh38
Transcripts
NM_033257.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
319870 c.544C>G p.L182V Chr.22: 20314794-20314794 on Build GRCh38
6050611 c.587C>T p.A196V Chr.22: 20314751-20314751 on Build GRCh38
4501535 c.591C>T p.A197A Chr.22: 20314747-20314747 on Build GRCh38
5444637 c.564G>A p.Q188Q Chr.22: 20314774-20314774 on Build GRCh38
5645465 c.572G>T p.C191F Chr.22: 20314766-20314766 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768848553 Chr.22: 20314733-20314733 on Build GRCh38
rs772769914 Chr.22: 20314912-20314912 on Build GRCh38
rs982273332 Chr.22: 20314599-20314599 on Build GRCh38
rs751027658 Chr.22: 20314664-20314664 on Build GRCh38
rs748105829 Chr.22: 20314748-20314748 on Build GRCh38
rs367906227 Chr.22: 20314627-20314627 on Build GRCh38
rs767088606 Chr.22: 20314801-20314801 on Build GRCh38
rs755338081 Chr.22: 20314822-20314822 on Build GRCh38
rs376912694 Chr.22: 20314671-20314671 on Build GRCh38
rs193920891 Chr.22: 20314730-20314730 on Build GRCh38
rs372042388 Chr.22: 20314666-20314666 on Build GRCh38
rs780593246 Chr.22: 20314819-20314819 on Build GRCh38
rs556178343 Chr.22: 20314656-20314656 on Build GRCh38
rs761498447 Chr.22: 20314905-20314905 on Build GRCh38
rs529605190 Chr.22: 20314923-20314923 on Build GRCh38
rs557927205 Chr.22: 20314846-20314846 on Build GRCh38
rs1056804 Chr.22: 20314753-20314753 on Build GRCh38
rs571530199 Chr.22: 20314606-20314606 on Build GRCh38
rs973457691 Chr.22: 20314551-20314551 on Build GRCh38
rs928140480 Chr.22: 20314944-20314944 on Build GRCh38
rs953966119 Chr.22: 20314488-20314488 on Build GRCh38
rs767128006 Chr.22: 20314911-20314911 on Build GRCh38
rs187047145 Chr.22: 20314841-20314841 on Build GRCh38
rs143957631 Chr.22: 20314763-20314763 on Build GRCh38
rs1056808 Chr.22: 20314786-20314786 on Build GRCh38
rs183700583 Chr.22: 20314881-20314881 on Build GRCh38
rs950806525 Chr.22: 20314885-20314885 on Build GRCh38
rs1056803 Chr.22: 20314741-20314741 on Build GRCh38
rs368248559 Chr.22: 20314674-20314674 on Build GRCh38
rs755384737 Chr.22: 20314840-20314841 on Build GRCh38
rs752358018 Chr.22: 20314653-20314653 on Build GRCh38
rs149702805 Chr.22: 20314703-20314703 on Build GRCh38
rs140892524 Chr.22: 20314720-20314720 on Build GRCh38
rs1007245094 Chr.22: 20314492-20314492 on Build GRCh38
rs191150933 Chr.22: 20314861-20314861 on Build GRCh38
rs1056805 Chr.22: 20314764-20314764 on Build GRCh38
rs1017548177 Chr.22: 20314513-20314513 on Build GRCh38
rs748423243 Chr.22: 20314835-20314835 on Build GRCh38
rs770766978 Chr.22: 20314624-20314624 on Build GRCh38
rs749034261 Chr.22: 20314888-20314888 on Build GRCh38
rs775573494 Chr.22: 20314876-20314876 on Build GRCh38
rs754194034 Chr.22: 20314821-20314821 on Build GRCh38
rs963383406 Chr.22: 20314547-20314547 on Build GRCh38
rs373175771 Chr.22: 20314827-20314827 on Build GRCh38
rs928140896 Chr.22: 20314600-20314600 on Build GRCh38
rs374728220 Chr.22: 20314591-20314591 on Build GRCh38
rs879561869 Chr.22: 20314734-20314734 on Build GRCh38
rs751239135 Chr.22: 20314787-20314787 on Build GRCh38
rs765145232 Chr.22: 20314641-20314641 on Build GRCh38
rs761182237 Chr.22: 20314794-20314794 on Build GRCh38
rs756595242 Chr.22: 20314811-20314811 on Build GRCh38
rs535612937 Chr.22: 20314721-20314721 on Build GRCh38
rs756433690 Chr.22: 20314718-20314718 on Build GRCh38
rs765373109 Chr.22: 20314860-20314861 on Build GRCh38
rs946403833 Chr.22: 20314623-20314623 on Build GRCh38
rs543408799 Chr.22: 20314891-20314891 on Build GRCh38
rs769849362 Chr.22: 20314872-20314872 on Build GRCh38
rs779259627 Chr.22: 20314737-20314737 on Build GRCh38
rs1055412939 Chr.22: 20314607-20314607 on Build GRCh38
rs763898486 Chr.22: 20314662-20314662 on Build GRCh38
rs914901310 Chr.22: 20314613-20314613 on Build GRCh38
rs754103164 Chr.22: 20314637-20314639 on Build GRCh38
rs950871085 Chr.22: 20314591-20314592 on Build GRCh38
rs775412122 Chr.22: 20314785-20314785 on Build GRCh38
rs747134952 Chr.22: 20314855-20314855 on Build GRCh38
rs749945281 Chr.22: 20314805-20314805 on Build GRCh38
rs919304886 Chr.22: 20314562-20314562 on Build GRCh38
rs367818045 Chr.22: 20314634-20314634 on Build GRCh38
rs868667788 Chr.22: 20314729-20314729 on Build GRCh38
rs1056802 Chr.22: 20314738-20314738 on Build GRCh38
rs563788433 Chr.22: 20314916-20314916 on Build GRCh38

View Full Product Details