Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 21632716-21633222 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC116
Entrez Gene ID
164592
Gene Name
coiled-coil domain containing 116
Gene Aliases
-
UniGene
Hs.131615
Gene Chromosome Location
Chr.22: 21632121-21637327 on Build GRCh38
Transcripts
NM_152612.2 XM_006724159.2 XM_011529984.2 XM_011529985.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6355967 c.36C>T p.A12A Chr.22: 21633217-21633217 on Build GRCh38
7417862 c.37G>A p.D13N Chr.22: 21633218-21633218 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs557330925 Chr.22: 21633085-21633085 on Build GRCh38
rs1025835520 Chr.22: 21633145-21633145 on Build GRCh38
rs866745369 Chr.22: 21633044-21633044 on Build GRCh38
rs770248199 Chr.22: 21633037-21633037 on Build GRCh38
rs916799473 Chr.22: 21632984-21632984 on Build GRCh38
rs900333769 Chr.22: 21632918-21632918 on Build GRCh38
rs915313797 Chr.22: 21632751-21632751 on Build GRCh38
rs746466917 Chr.22: 21632990-21632990 on Build GRCh38
rs535121121 Chr.22: 21632742-21632742 on Build GRCh38
rs977778573 Chr.22: 21632962-21632962 on Build GRCh38
rs1002482034 Chr.22: 21633153-21633153 on Build GRCh38
rs751664140 Chr.22: 21633086-21633086 on Build GRCh38
rs747548931 Chr.22: 21633115-21633115 on Build GRCh38
rs771621127 Chr.22: 21632953-21632953 on Build GRCh38
rs957665776 Chr.22: 21632981-21632981 on Build GRCh38
rs537245242 Chr.22: 21633076-21633076 on Build GRCh38
rs534395083 Chr.22: 21632802-21632802 on Build GRCh38
rs938828985 Chr.22: 21633198-21633198 on Build GRCh38
rs942677846 Chr.22: 21633101-21633101 on Build GRCh38
rs193114164 Chr.22: 21632901-21632901 on Build GRCh38
rs1006107649 Chr.22: 21632903-21632903 on Build GRCh38
rs535404870 Chr.22: 21632743-21632743 on Build GRCh38
rs1034010266 Chr.22: 21633169-21633169 on Build GRCh38
rs949556855 Chr.22: 21632808-21632808 on Build GRCh38
rs772340766 Chr.22: 21633035-21633035 on Build GRCh38
rs994328067 Chr.22: 21632819-21632819 on Build GRCh38
rs915449470 Chr.22: 21632909-21632909 on Build GRCh38
rs1046919836 Chr.22: 21632818-21632818 on Build GRCh38
rs982362950 Chr.22: 21632860-21632860 on Build GRCh38
rs573966878 Chr.22: 21633194-21633194 on Build GRCh38
rs150595883 Chr.22: 21632764-21632764 on Build GRCh38
rs946782053 Chr.22: 21632960-21632960 on Build GRCh38
rs993904781 Chr.22: 21633130-21633130 on Build GRCh38
rs778835689 Chr.22: 21633174-21633174 on Build GRCh38
rs991034536 Chr.22: 21633203-21633203 on Build GRCh38
rs34414420 Chr.22: 21632994-21632995 on Build GRCh38
rs1051117861 Chr.22: 21633004-21633004 on Build GRCh38
rs771595725 Chr.22: 21633007-21633007 on Build GRCh38
rs933888286 Chr.22: 21632776-21632776 on Build GRCh38
rs760094449 Chr.22: 21633024-21633024 on Build GRCh38
rs778136755 Chr.22: 21633061-21633061 on Build GRCh38
rs957695059 Chr.22: 21633195-21633195 on Build GRCh38
rs936889987 Chr.22: 21632747-21632747 on Build GRCh38
rs946824731 Chr.22: 21632759-21632759 on Build GRCh38
rs1030526437 Chr.22: 21632950-21632950 on Build GRCh38
rs991499794 Chr.22: 21632982-21632982 on Build GRCh38
rs763238058 Chr.22: 21633083-21633083 on Build GRCh38
rs767557841 Chr.22: 21633124-21633124 on Build GRCh38
rs930013368 Chr.22: 21632866-21632866 on Build GRCh38
rs898247888 Chr.22: 21632817-21632817 on Build GRCh38
rs1039368465 Chr.22: 21633112-21633112 on Build GRCh38
rs754021904 Chr.22: 21632888-21632888 on Build GRCh38
rs1035218157 Chr.22: 21632864-21632864 on Build GRCh38
rs897213116 Chr.22: 21632847-21632847 on Build GRCh38
rs906976268 Chr.22: 21633151-21633151 on Build GRCh38
rs942624978 Chr.22: 21633047-21633047 on Build GRCh38
rs888464791 Chr.22: 21633065-21633065 on Build GRCh38
rs748390541 Chr.22: 21632929-21632929 on Build GRCh38
rs867353228 Chr.22: 21633129-21633129 on Build GRCh38
rs777316606 Chr.22: 21633015-21633015 on Build GRCh38
rs966445438 Chr.22: 21632958-21632958 on Build GRCh38
rs1017496764 Chr.22: 21632915-21632915 on Build GRCh38
rs577714097 Chr.22: 21632976-21632976 on Build GRCh38
rs753821344 Chr.22: 21633160-21633160 on Build GRCh38
rs889692637 Chr.22: 21633029-21633029 on Build GRCh38
rs754859285 Chr.22: 21633161-21633161 on Build GRCh38
rs775709983 Chr.22: 21633052-21633052 on Build GRCh38
rs1051019707 Chr.22: 21632796-21632797 on Build GRCh38
rs930047834 Chr.22: 21633023-21633023 on Build GRCh38
rs918711739 Chr.22: 21633009-21633009 on Build GRCh38
rs1025276961 Chr.22: 21632830-21632830 on Build GRCh38
rs1016803662 Chr.22: 21632812-21632812 on Build GRCh38
rs781248267 Chr.22: 21633116-21633116 on Build GRCh38
rs779072057 Chr.22: 21632925-21632925 on Build GRCh38

View Full Product Details