Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 21636954-21637396 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC116
Entrez Gene ID
164592
Gene Name
coiled-coil domain containing 116
Gene Aliases
-
UniGene
Hs.131615
Gene Chromosome Location
Chr.22: 21632121-21637327 on Build GRCh38
Transcripts
NM_152612.2 XM_006724159.2 XM_011529984.2 XM_011529985.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7359704 c.1733C>A p.S578Y Chr.22: 21636961-21636961 on Build GRCh38
6161751 c.1839C>A p.V613V Chr.22: 21637067-21637067 on Build GRCh38
5858986 c.1766C>T p.A589V Chr.22: 21636994-21636994 on Build GRCh38
4854146 c.1804G>A p.E602K Chr.22: 21637032-21637032 on Build GRCh38
4472046 c.1756C>T p.R586C Chr.22: 21636984-21636984 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs570799372 Chr.22: 21637365-21637365 on Build GRCh38
rs985455011 Chr.22: 21637360-21637360 on Build GRCh38
rs763434723 Chr.22: 21637072-21637072 on Build GRCh38
rs764497269 Chr.22: 21637077-21637077 on Build GRCh38
rs751784450 Chr.22: 21636989-21636989 on Build GRCh38
rs866523353 Chr.22: 21636994-21636994 on Build GRCh38
rs903826335 Chr.22: 21637211-21637211 on Build GRCh38
rs1037672294 Chr.22: 21637146-21637146 on Build GRCh38
rs185677785 Chr.22: 21637112-21637112 on Build GRCh38
rs368909911 Chr.22: 21637063-21637063 on Build GRCh38
rs746604543 Chr.22: 21637059-21637059 on Build GRCh38
rs754857590 Chr.22: 21637008-21637008 on Build GRCh38
rs1002034996 Chr.22: 21637200-21637200 on Build GRCh38
rs763309538 Chr.22: 21636984-21636984 on Build GRCh38
rs933609190 Chr.22: 21637346-21637346 on Build GRCh38
rs1009597578 Chr.22: 21637160-21637160 on Build GRCh38
rs370189885 Chr.22: 21637096-21637096 on Build GRCh38
rs200688012 Chr.22: 21637073-21637078 on Build GRCh38
rs758199431 Chr.22: 21637116-21637116 on Build GRCh38
rs138232832 Chr.22: 21637068-21637068 on Build GRCh38
rs1024031743 Chr.22: 21637246-21637246 on Build GRCh38
rs897841974 Chr.22: 21637147-21637147 on Build GRCh38
rs531575551 Chr.22: 21637092-21637092 on Build GRCh38
rs546212895 Chr.22: 21637140-21637140 on Build GRCh38
rs766839776 Chr.22: 21637128-21637128 on Build GRCh38
rs1000521787 Chr.22: 21637225-21637225 on Build GRCh38
rs778817130 Chr.22: 21637011-21637011 on Build GRCh38
rs562099758 Chr.22: 21637240-21637240 on Build GRCh38
rs534017416 Chr.22: 21637270-21637270 on Build GRCh38
rs775996084 Chr.22: 21637062-21637062 on Build GRCh38
rs970647143 Chr.22: 21637181-21637181 on Build GRCh38
rs913578918 Chr.22: 21637039-21637039 on Build GRCh38
rs555243931 Chr.22: 21637069-21637069 on Build GRCh38
rs777294772 Chr.22: 21637116-21637118 on Build GRCh38
rs949759061 Chr.22: 21636986-21636986 on Build GRCh38
rs539042211 Chr.22: 21636991-21636991 on Build GRCh38
rs2303399 Chr.22: 21637061-21637061 on Build GRCh38
rs141503507 Chr.22: 21637000-21637000 on Build GRCh38
rs199576042 Chr.22: 21637145-21637145 on Build GRCh38
rs548178257 Chr.22: 21637094-21637094 on Build GRCh38
rs919548153 Chr.22: 21636996-21636996 on Build GRCh38
rs986278169 Chr.22: 21637366-21637366 on Build GRCh38
rs548777273 Chr.22: 21637182-21637182 on Build GRCh38
rs923510777 Chr.22: 21637338-21637338 on Build GRCh38
rs547821107 Chr.22: 21637362-21637362 on Build GRCh38
rs971107189 Chr.22: 21637269-21637269 on Build GRCh38
rs781356113 Chr.22: 21636995-21636995 on Build GRCh38
rs1021384696 Chr.22: 21637170-21637170 on Build GRCh38
rs777423951 Chr.22: 21637044-21637044 on Build GRCh38
rs753880873 Chr.22: 21637004-21637004 on Build GRCh38
rs913547613 Chr.22: 21637323-21637323 on Build GRCh38
rs150451119 Chr.22: 21637018-21637018 on Build GRCh38
rs1006719807 Chr.22: 21637135-21637135 on Build GRCh38
rs750447299 Chr.22: 21637100-21637100 on Build GRCh38
rs752580182 Chr.22: 21637113-21637113 on Build GRCh38
rs368046807 Chr.22: 21636992-21636992 on Build GRCh38
rs563008474 Chr.22: 21637154-21637154 on Build GRCh38
rs768723979 Chr.22: 21637334-21637334 on Build GRCh38
rs966349097 Chr.22: 21637328-21637328 on Build GRCh38
rs1033968838 Chr.22: 21637231-21637231 on Build GRCh38
rs527989539 Chr.22: 21637337-21637337 on Build GRCh38
rs762817553 Chr.22: 21637244-21637244 on Build GRCh38
rs930288793 Chr.22: 21637358-21637358 on Build GRCh38
rs764444464 Chr.22: 21636985-21636985 on Build GRCh38
rs560544337 Chr.22: 21637101-21637101 on Build GRCh38
rs1032893764 Chr.22: 21637234-21637234 on Build GRCh38
rs531808607 Chr.22: 21637173-21637173 on Build GRCh38
rs751835624 Chr.22: 21637080-21637080 on Build GRCh38
rs1040518747 Chr.22: 21637099-21637099 on Build GRCh38

View Full Product Details