Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 23124468-23125004 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNAZ
Entrez Gene ID
2781
Gene Name
G protein subunit alpha z
Gene Aliases
-
UniGene
Hs.584760
Gene Chromosome Location
Chr.22: 23070483-23125037 on Build GRCh38
Transcripts
NM_002073.2 NM_014433.2 XM_011530149.2 XM_011530150.1 XM_011530151.2 XM_011530152.2 XM_011530154.2 XM_017028774.1

Gene Symbol
RSPH14
Entrez Gene ID
27156
Gene Name
radial spoke head 14 homolog
Gene Aliases
RTDR1
UniGene
Hs.526920
Gene Chromosome Location
Chr.22: 23059408-23180048 on Build GRCh38
Transcripts
NM_002073.2 NM_014433.2 XM_011530149.2 XM_011530150.1 XM_011530151.2 XM_011530152.2 XM_011530154.2 XM_017028774.1

SNP Detail

SNP ID Chromosome Location
rs889051831 Chr.22: 23124816-23124816 on Build GRCh38
rs775449475 Chr.22: 23124926-23124926 on Build GRCh38
rs75987287 Chr.22: 23124545-23124545 on Build GRCh38
rs941287767 Chr.22: 23124935-23124935 on Build GRCh38
rs5751586 Chr.22: 23124619-23124619 on Build GRCh38
rs535987574 Chr.22: 23124617-23124617 on Build GRCh38
rs770849102 Chr.22: 23124865-23124865 on Build GRCh38
rs11703490 Chr.22: 23124782-23124782 on Build GRCh38
rs148043323 Chr.22: 23124880-23124880 on Build GRCh38
rs372215395 Chr.22: 23124960-23124960 on Build GRCh38
rs544114955 Chr.22: 23124794-23124794 on Build GRCh38
rs980361488 Chr.22: 23124814-23124814 on Build GRCh38
rs957656892 Chr.22: 23124835-23124835 on Build GRCh38
rs897685651 Chr.22: 23124711-23124711 on Build GRCh38
rs990277888 Chr.22: 23124849-23124849 on Build GRCh38
rs1023915770 Chr.22: 23124768-23124768 on Build GRCh38
rs746726450 Chr.22: 23124705-23124705 on Build GRCh38
rs923280609 Chr.22: 23124888-23124888 on Build GRCh38
rs1051891 Chr.22: 23124831-23124831 on Build GRCh38
rs780032386 Chr.22: 23124849-23124850 on Build GRCh38
rs993884244 Chr.22: 23124767-23124767 on Build GRCh38
rs901506478 Chr.22: 23124837-23124837 on Build GRCh38
rs190849942 Chr.22: 23124509-23124509 on Build GRCh38
rs917215885 Chr.22: 23124670-23124670 on Build GRCh38
rs13407 Chr.22: 23124893-23124893 on Build GRCh38
rs906855152 Chr.22: 23124968-23124968 on Build GRCh38
rs990237943 Chr.22: 23124600-23124600 on Build GRCh38
rs913410467 Chr.22: 23124853-23124853 on Build GRCh38
rs927480490 Chr.22: 23124547-23124547 on Build GRCh38
rs959215394 Chr.22: 23124952-23124952 on Build GRCh38
rs897026888 Chr.22: 23124945-23124945 on Build GRCh38
rs1020641401 Chr.22: 23124877-23124877 on Build GRCh38
rs980795373 Chr.22: 23124529-23124529 on Build GRCh38
rs950120678 Chr.22: 23124672-23124672 on Build GRCh38
rs1038132728 Chr.22: 23124943-23124943 on Build GRCh38
rs917295237 Chr.22: 23124958-23124958 on Build GRCh38
rs879225921 Chr.22: 23124517-23124517 on Build GRCh38
rs775113413 Chr.22: 23124944-23124944 on Build GRCh38
rs937578269 Chr.22: 23124638-23124638 on Build GRCh38
rs1040365339 Chr.22: 23124671-23124671 on Build GRCh38
rs773928497 Chr.22: 23124490-23124490 on Build GRCh38
rs933922801 Chr.22: 23124683-23124683 on Build GRCh38
rs1016739725 Chr.22: 23124744-23124744 on Build GRCh38
rs558447704 Chr.22: 23124762-23124762 on Build GRCh38
rs1023926949 Chr.22: 23124519-23124519 on Build GRCh38
rs73878651 Chr.22: 23124722-23124722 on Build GRCh38
rs967514540 Chr.22: 23124510-23124510 on Build GRCh38
rs566912917 Chr.22: 23124597-23124597 on Build GRCh38
rs971637643 Chr.22: 23124528-23124528 on Build GRCh38
rs183047051 Chr.22: 23124809-23124809 on Build GRCh38
rs936190707 Chr.22: 23124572-23124572 on Build GRCh38
rs2727 Chr.22: 23124493-23124493 on Build GRCh38
rs1056216908 Chr.22: 23124650-23124650 on Build GRCh38
rs769882030 Chr.22: 23124913-23124913 on Build GRCh38
rs991721926 Chr.22: 23124957-23124957 on Build GRCh38
rs1007439469 Chr.22: 23124819-23124819 on Build GRCh38
rs534872630 Chr.22: 23124724-23124724 on Build GRCh38
rs932006302 Chr.22: 23124894-23124894 on Build GRCh38
rs913433849 Chr.22: 23124495-23124495 on Build GRCh38
rs552603708 Chr.22: 23124645-23124645 on Build GRCh38
rs1036445485 Chr.22: 23124673-23124673 on Build GRCh38
rs779119158 Chr.22: 23124506-23124506 on Build GRCh38

View Full Product Details