Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 23180548-23181057 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCR
Entrez Gene ID
613
Gene Name
BCR, RhoGEF and GTPase activating protein
Gene Aliases
ALL, BCR1, CML, D22S11, D22S662, PHL
UniGene
Hs.517461
Gene Chromosome Location
Chr.22: 23180365-23318037 on Build GRCh38
Transcripts
NM_004327.3 NM_021574.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4171068 c.86G>A p.G29D Chr.22: 23181046-23181046 on Build GRCh38
4171065 c.77G>A p.R26H Chr.22: 23181037-23181037 on Build GRCh38
5417597 c.72G>A p.E24E Chr.22: 23181032-23181032 on Build GRCh38
6052196 c.81A>G p.S27S Chr.22: 23181041-23181041 on Build GRCh38
5657848 c.67A>G p.M23V Chr.22: 23181027-23181027 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373742833 Chr.22: 23180592-23180593 on Build GRCh38
rs964034925 Chr.22: 23180691-23180691 on Build GRCh38
rs139121414 Chr.22: 23180952-23180952 on Build GRCh38
rs761634532 Chr.22: 23180976-23180976 on Build GRCh38
rs868681508 Chr.22: 23180737-23180737 on Build GRCh38
rs537413302 Chr.22: 23181032-23181032 on Build GRCh38
rs554517491 Chr.22: 23180849-23180850 on Build GRCh38
rs1004233456 Chr.22: 23180769-23180780 on Build GRCh38
rs573152643 Chr.22: 23180600-23180600 on Build GRCh38
rs774090412 Chr.22: 23181024-23181024 on Build GRCh38
rs746347186 Chr.22: 23181018-23181018 on Build GRCh38
rs757563933 Chr.22: 23181015-23181015 on Build GRCh38
rs908285863 Chr.22: 23180858-23180858 on Build GRCh38
rs368527107 Chr.22: 23180960-23180960 on Build GRCh38
rs1046826941 Chr.22: 23180733-23180733 on Build GRCh38
rs199561166 Chr.22: 23180998-23180998 on Build GRCh38
rs998867996 Chr.22: 23180636-23180636 on Build GRCh38
rs771287353 Chr.22: 23180942-23180942 on Build GRCh38
rs370385572 Chr.22: 23180886-23180887 on Build GRCh38
rs529512434 Chr.22: 23180794-23180794 on Build GRCh38
rs551631242 Chr.22: 23181022-23181022 on Build GRCh38
rs530488003 Chr.22: 23180719-23180719 on Build GRCh38
rs987519870 Chr.22: 23180685-23180685 on Build GRCh38
rs779080917 Chr.22: 23180922-23180922 on Build GRCh38
rs747241983 Chr.22: 23181014-23181014 on Build GRCh38
rs765207801 Chr.22: 23181007-23181007 on Build GRCh38
rs931012980 Chr.22: 23180572-23180572 on Build GRCh38
rs931150318 Chr.22: 23180729-23180729 on Build GRCh38
rs756116107 Chr.22: 23180996-23180996 on Build GRCh38
rs1032161132 Chr.22: 23180773-23180773 on Build GRCh38
rs769150219 Chr.22: 23180953-23180953 on Build GRCh38
rs1002564967 Chr.22: 23180580-23180580 on Build GRCh38
rs866141429 Chr.22: 23180833-23180833 on Build GRCh38
rs750346210 Chr.22: 23180990-23180990 on Build GRCh38
rs908313145 Chr.22: 23180739-23180739 on Build GRCh38
rs938404671 Chr.22: 23180862-23180862 on Build GRCh38
rs777061768 Chr.22: 23180947-23180947 on Build GRCh38
rs988982933 Chr.22: 23180754-23180754 on Build GRCh38
rs112064774 Chr.22: 23180617-23180617 on Build GRCh38
rs1043271505 Chr.22: 23180964-23180964 on Build GRCh38
rs954853708 Chr.22: 23180776-23180776 on Build GRCh38
rs879125164 Chr.22: 23180592-23180592 on Build GRCh38
rs867957216 Chr.22: 23180848-23180848 on Build GRCh38
rs1009312614 Chr.22: 23180791-23180791 on Build GRCh38
rs774852686 Chr.22: 23181020-23181020 on Build GRCh38
rs372973719 Chr.22: 23181008-23181008 on Build GRCh38
rs564887525 Chr.22: 23180647-23180647 on Build GRCh38
rs762494209 Chr.22: 23180956-23180956 on Build GRCh38
rs767433795 Chr.22: 23180983-23180983 on Build GRCh38
rs879548982 Chr.22: 23180585-23180592 on Build GRCh38
rs968652992 Chr.22: 23180802-23180802 on Build GRCh38
rs1013088369 Chr.22: 23180764-23180765 on Build GRCh38
rs753721385 Chr.22: 23181036-23181036 on Build GRCh38
rs199698427 Chr.22: 23180895-23180896 on Build GRCh38
rs781572099 Chr.22: 23181017-23181017 on Build GRCh38
rs919862749 Chr.22: 23180821-23180821 on Build GRCh38
rs768462153 Chr.22: 23180593-23180594 on Build GRCh38
rs375737541 Chr.22: 23180999-23180999 on Build GRCh38
rs1057292290 Chr.22: 23180903-23180903 on Build GRCh38
rs773189108 Chr.22: 23181029-23181029 on Build GRCh38
rs904813538 Chr.22: 23181035-23181035 on Build GRCh38
rs1046858519 Chr.22: 23180573-23180573 on Build GRCh38
rs975302139 Chr.22: 23180819-23180819 on Build GRCh38
rs180778730 Chr.22: 23180703-23180703 on Build GRCh38
rs768563588 Chr.22: 23180591-23180592 on Build GRCh38
rs767388787 Chr.22: 23181026-23181026 on Build GRCh38
rs749045362 Chr.22: 23181013-23181014 on Build GRCh38
rs954856026 Chr.22: 23180675-23180675 on Build GRCh38
rs566067977 Chr.22: 23180938-23180938 on Build GRCh38
rs571311446 Chr.22: 23181031-23181031 on Build GRCh38
rs866563446 Chr.22: 23180589-23180589 on Build GRCh38
rs912462454 Chr.22: 23180687-23180687 on Build GRCh38
rs866164558 Chr.22: 23180759-23180759 on Build GRCh38
rs370530698 Chr.22: 23180927-23180927 on Build GRCh38
rs886749187 Chr.22: 23180576-23180576 on Build GRCh38
rs758542346 Chr.22: 23181011-23181011 on Build GRCh38
rs952509258 Chr.22: 23180826-23180826 on Build GRCh38
rs796688570 Chr.22: 23180591-23180592 on Build GRCh38
rs1021709037 Chr.22: 23180616-23180616 on Build GRCh38
rs777901995 Chr.22: 23181012-23181012 on Build GRCh38
rs549535838 Chr.22: 23180795-23180795 on Build GRCh38
rs867644920 Chr.22: 23180583-23180583 on Build GRCh38
rs75695389 Chr.22: 23180765-23180765 on Build GRCh38
rs894760656 Chr.22: 23180599-23180599 on Build GRCh38
rs780142795 Chr.22: 23180570-23180571 on Build GRCh38
rs1031774483 Chr.22: 23180649-23180649 on Build GRCh38
rs914042531 Chr.22: 23180766-23180766 on Build GRCh38
rs560994400 Chr.22: 23180767-23180767 on Build GRCh38
rs777984327 Chr.22: 23180936-23180936 on Build GRCh38
rs3138696 Chr.22: 23180602-23180604 on Build GRCh38
rs938337410 Chr.22: 23180751-23180751 on Build GRCh38
rs999003792 Chr.22: 23180771-23180771 on Build GRCh38
rs376532812 Chr.22: 23181025-23181025 on Build GRCh38
rs75147926 Chr.22: 23180623-23180623 on Build GRCh38
rs1057501009 Chr.22: 23180585-23180585 on Build GRCh38
rs201435938 Chr.22: 23180852-23180853 on Build GRCh38
rs772384988 Chr.22: 23180932-23180932 on Build GRCh38
rs1056838268 Chr.22: 23180754-23180755 on Build GRCh38

View Full Product Details